Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 154027935-154028438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM114A2
Entrez Gene ID
10827
Gene Name
family with sequence similarity 114 member A2
Gene Aliases
133K02, C5orf3
UniGene
Hs.166551
Gene Chromosome Location
Chr.5: 153990131-154038938 on Build GRCh38
Transcripts
NM_001317993.1 NM_001317994.1 NM_001317995.1 NM_018691.3 XM_005268360.4 XM_005268361.4 XM_011537540.2 XM_011537541.2 XM_011537542.2 XM_017008967.1 XM_017008968.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6169904 c.496-1G>T p.? Chr.5: 154028284-154028284 on Build GRCh38
4553859 c.603G>A p.L201L Chr.5: 154028176-154028176 on Build GRCh38
1435402 c.548delA p.K183fs*5 Chr.5: 154028231-154028231 on Build GRCh38
6658112 c.611G>A p.R204Q Chr.5: 154028168-154028168 on Build GRCh38
420818 c.566C>T p.A189V Chr.5: 154028213-154028213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370798035 Chr.5: 154028169-154028169 on Build GRCh38
rs541018198 Chr.5: 154028031-154028031 on Build GRCh38
rs986177233 Chr.5: 154028039-154028039 on Build GRCh38
rs769595669 Chr.5: 154028193-154028193 on Build GRCh38
rs757546488 Chr.5: 154028244-154028244 on Build GRCh38
rs147017359 Chr.5: 154028264-154028264 on Build GRCh38
rs778394677 Chr.5: 154028241-154028241 on Build GRCh38
rs141421700 Chr.5: 154028160-154028160 on Build GRCh38
rs758017033 Chr.5: 154028103-154028103 on Build GRCh38
rs763279667 Chr.5: 154028345-154028345 on Build GRCh38
rs766115411 Chr.5: 154028210-154028210 on Build GRCh38
rs920492572 Chr.5: 154028056-154028060 on Build GRCh38
rs781782618 Chr.5: 154028137-154028137 on Build GRCh38
rs530693291 Chr.5: 154028350-154028350 on Build GRCh38
rs147781886 Chr.5: 154028157-154028157 on Build GRCh38
rs773016496 Chr.5: 154028197-154028197 on Build GRCh38
rs773011900 Chr.5: 154028294-154028294 on Build GRCh38
rs754436402 Chr.5: 154028131-154028131 on Build GRCh38
rs754280437 Chr.5: 154028242-154028242 on Build GRCh38
rs77279974 Chr.5: 154027968-154027968 on Build GRCh38
rs543566221 Chr.5: 154027990-154027990 on Build GRCh38
rs911764767 Chr.5: 154028028-154028028 on Build GRCh38
rs780055579 Chr.5: 154028274-154028274 on Build GRCh38
rs879200684 Chr.5: 154028319-154028321 on Build GRCh38
rs576550624 Chr.5: 154027982-154027982 on Build GRCh38
rs765834980 Chr.5: 154028104-154028104 on Build GRCh38
rs776515953 Chr.5: 154028044-154028044 on Build GRCh38
rs776676028 Chr.5: 154028260-154028261 on Build GRCh38
rs774070536 Chr.5: 154028330-154028330 on Build GRCh38
rs762875989 Chr.5: 154028299-154028299 on Build GRCh38
rs765355725 Chr.5: 154028313-154028315 on Build GRCh38
rs937954235 Chr.5: 154028233-154028233 on Build GRCh38
rs779259950 Chr.5: 154028261-154028261 on Build GRCh38
rs370349510 Chr.5: 154028146-154028146 on Build GRCh38
rs772152341 Chr.5: 154028271-154028271 on Build GRCh38
rs866027522 Chr.5: 154027961-154027961 on Build GRCh38
rs200198235 Chr.5: 154028202-154028202 on Build GRCh38
rs377171595 Chr.5: 154028163-154028163 on Build GRCh38
rs775865472 Chr.5: 154028327-154028328 on Build GRCh38
rs896508282 Chr.5: 154028270-154028270 on Build GRCh38
rs988800352 Chr.5: 154027959-154027959 on Build GRCh38
rs180926126 Chr.5: 154028159-154028159 on Build GRCh38
rs377447910 Chr.5: 154028334-154028334 on Build GRCh38
rs753279745 Chr.5: 154028234-154028234 on Build GRCh38
rs34738549 Chr.5: 154028247-154028247 on Build GRCh38
rs1039390974 Chr.5: 154028053-154028053 on Build GRCh38
rs1015666896 Chr.5: 154028312-154028312 on Build GRCh38
rs769864208 Chr.5: 154028240-154028240 on Build GRCh38
rs759215466 Chr.5: 154028333-154028333 on Build GRCh38
rs199529104 Chr.5: 154028155-154028155 on Build GRCh38
rs374213195 Chr.5: 154028158-154028158 on Build GRCh38
rs748113001 Chr.5: 154028281-154028281 on Build GRCh38
rs529318628 Chr.5: 154028030-154028030 on Build GRCh38
rs200664638 Chr.5: 154028222-154028222 on Build GRCh38
rs200243840 Chr.5: 154028226-154028226 on Build GRCh38
rs759717974 Chr.5: 154028280-154028281 on Build GRCh38
rs759094509 Chr.5: 154028223-154028223 on Build GRCh38
rs748602939 Chr.5: 154028139-154028139 on Build GRCh38
rs1010656642 Chr.5: 154028288-154028288 on Build GRCh38
rs974290142 Chr.5: 154027981-154027981 on Build GRCh38
rs771227234 Chr.5: 154028152-154028152 on Build GRCh38
rs866738698 Chr.5: 154028259-154028260 on Build GRCh38
rs913727847 Chr.5: 154028168-154028168 on Build GRCh38
rs762622396 Chr.5: 154028207-154028207 on Build GRCh38
rs946497331 Chr.5: 154028203-154028203 on Build GRCh38
rs368390978 Chr.5: 154028328-154028328 on Build GRCh38
rs562166107 Chr.5: 154028027-154028027 on Build GRCh38
rs749504863 Chr.5: 154028151-154028151 on Build GRCh38
rs751132745 Chr.5: 154028114-154028114 on Build GRCh38
rs770821931 Chr.5: 154028134-154028143 on Build GRCh38
rs962891195 Chr.5: 154027970-154027970 on Build GRCh38
rs747079046 Chr.5: 154028292-154028292 on Build GRCh38

View Full Product Details