Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 96879833-96880363 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPQ
Entrez Gene ID
10404
Gene Name
carboxypeptidase Q
Gene Aliases
LDP, PGCP
UniGene
Hs.156178
Gene Chromosome Location
Chr.8: 96645227-97143503 on Build GRCh38
Transcripts
NM_016134.3 XM_017012953.1 XM_017012954.1 XM_017012955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6047879 c.694G>T p.A232S Chr.8: 96879850-96879850 on Build GRCh38
3835190 c.707T>G p.V236G Chr.8: 96879863-96879863 on Build GRCh38
5007811 c.797C>A p.T266N Chr.8: 96879953-96879953 on Build GRCh38
6634756 c.700A>G p.I234V Chr.8: 96879856-96879856 on Build GRCh38
3731755 c.705G>A p.T235T Chr.8: 96879861-96879861 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771260509 Chr.8: 96879946-96879946 on Build GRCh38
rs748699520 Chr.8: 96879917-96879917 on Build GRCh38
rs774757454 Chr.8: 96879968-96879968 on Build GRCh38
rs955252527 Chr.8: 96880295-96880295 on Build GRCh38
rs756363859 Chr.8: 96879935-96879935 on Build GRCh38
rs753447649 Chr.8: 96879889-96879889 on Build GRCh38
rs774599864 Chr.8: 96879863-96879863 on Build GRCh38
rs186228301 Chr.8: 96880155-96880155 on Build GRCh38
rs112681566 Chr.8: 96879899-96879899 on Build GRCh38
rs569944091 Chr.8: 96880344-96880344 on Build GRCh38
rs761690836 Chr.8: 96880293-96880293 on Build GRCh38
rs141107654 Chr.8: 96879891-96879891 on Build GRCh38
rs547116623 Chr.8: 96880220-96880220 on Build GRCh38
rs368218633 Chr.8: 96879854-96879854 on Build GRCh38
rs375182115 Chr.8: 96879986-96879986 on Build GRCh38
rs372113350 Chr.8: 96879861-96879861 on Build GRCh38
rs1017938669 Chr.8: 96880330-96880330 on Build GRCh38
rs977174043 Chr.8: 96879882-96879882 on Build GRCh38
rs151033390 Chr.8: 96879898-96879898 on Build GRCh38
rs986479684 Chr.8: 96880327-96880327 on Build GRCh38
rs1024526578 Chr.8: 96880267-96880267 on Build GRCh38
rs556155987 Chr.8: 96880294-96880294 on Build GRCh38
rs781535494 Chr.8: 96879911-96879911 on Build GRCh38
rs1040265324 Chr.8: 96880065-96880065 on Build GRCh38
rs771615579 Chr.8: 96879989-96879989 on Build GRCh38
rs560500593 Chr.8: 96879896-96879896 on Build GRCh38
rs567331653 Chr.8: 96880222-96880222 on Build GRCh38
rs896706656 Chr.8: 96880140-96880140 on Build GRCh38
rs897671902 Chr.8: 96880097-96880097 on Build GRCh38
rs778316900 Chr.8: 96879937-96879937 on Build GRCh38
rs140416726 Chr.8: 96879973-96879973 on Build GRCh38
rs898322489 Chr.8: 96880252-96880252 on Build GRCh38
rs550967978 Chr.8: 96880024-96880024 on Build GRCh38
rs749474745 Chr.8: 96879945-96879945 on Build GRCh38
rs28452385 Chr.8: 96880120-96880120 on Build GRCh38
rs993064538 Chr.8: 96879856-96879856 on Build GRCh38
rs201646330 Chr.8: 96879883-96879883 on Build GRCh38
rs776386723 Chr.8: 96880006-96880006 on Build GRCh38
rs925456050 Chr.8: 96880106-96880106 on Build GRCh38
rs1013540248 Chr.8: 96880146-96880146 on Build GRCh38
rs76533568 Chr.8: 96880263-96880263 on Build GRCh38
rs149951331 Chr.8: 96880003-96880003 on Build GRCh38
rs183452020 Chr.8: 96880022-96880022 on Build GRCh38
rs775566173 Chr.8: 96879877-96879877 on Build GRCh38
rs775562223 Chr.8: 96879983-96879983 on Build GRCh38
rs750129395 Chr.8: 96880037-96880037 on Build GRCh38
rs772251568 Chr.8: 96879970-96879970 on Build GRCh38
rs935697062 Chr.8: 96879916-96879916 on Build GRCh38
rs200913578 Chr.8: 96879910-96879910 on Build GRCh38
rs888501102 Chr.8: 96880168-96880168 on Build GRCh38
rs1005624140 Chr.8: 96880177-96880177 on Build GRCh38
rs1017220819 Chr.8: 96880187-96880187 on Build GRCh38
rs745938647 Chr.8: 96879969-96879969 on Build GRCh38
rs763963550 Chr.8: 96879886-96879886 on Build GRCh38
rs139595101 Chr.8: 96879860-96879860 on Build GRCh38
rs955032340 Chr.8: 96879966-96879966 on Build GRCh38
rs935621010 Chr.8: 96880109-96880109 on Build GRCh38
rs972675589 Chr.8: 96879908-96879908 on Build GRCh38
rs759260946 Chr.8: 96879884-96879885 on Build GRCh38
rs905297892 Chr.8: 96880165-96880165 on Build GRCh38
rs1044312635 Chr.8: 96880089-96880089 on Build GRCh38
rs578138575 Chr.8: 96879865-96879865 on Build GRCh38
rs778966186 Chr.8: 96879887-96879887 on Build GRCh38

View Full Product Details