Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 108228597-108229109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NBPF4
Entrez Gene ID
148545
Gene Name
neuroblastoma breakpoint family member 4
Gene Aliases
-
UniGene
Hs.511849
Gene Chromosome Location
Chr.1: 108222464-108244081 on Build GRCh38
Transcripts
NM_001143989.2 XM_011540763.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3355887 c.1472G>A p.S491N Chr.1: 108229108-108229108 on Build GRCh38
5467953 c.1588C>A p.Q530K Chr.1: 108228992-108228992 on Build GRCh38
4004653 c.1492C>A p.Q498K Chr.1: 108229088-108229088 on Build GRCh38
6389408 c.1630G>T p.A544S Chr.1: 108228950-108228950 on Build GRCh38
4467953 c.1517C>T p.P506L Chr.1: 108229063-108229063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963261650 Chr.1: 108228945-108228945 on Build GRCh38
rs867510594 Chr.1: 108228806-108228806 on Build GRCh38
rs1026132624 Chr.1: 108229017-108229017 on Build GRCh38
rs879240199 Chr.1: 108228683-108228683 on Build GRCh38
rs992700747 Chr.1: 108229077-108229077 on Build GRCh38
rs369137709 Chr.1: 108229071-108229071 on Build GRCh38
rs915609991 Chr.1: 108229086-108229086 on Build GRCh38
rs2260438 Chr.1: 108229085-108229085 on Build GRCh38
rs77009338 Chr.1: 108228727-108228727 on Build GRCh38
rs901456043 Chr.1: 108228684-108228684 on Build GRCh38
rs904694270 Chr.1: 108228655-108228655 on Build GRCh38
rs760936384 Chr.1: 108229002-108229002 on Build GRCh38
rs866894835 Chr.1: 108229056-108229056 on Build GRCh38
rs61799399 Chr.1: 108228902-108228902 on Build GRCh38
rs773693443 Chr.1: 108229001-108229001 on Build GRCh38
rs777478410 Chr.1: 108229049-108229049 on Build GRCh38
rs779091594 Chr.1: 108228953-108228953 on Build GRCh38
rs972279286 Chr.1: 108228967-108228967 on Build GRCh38
rs889784332 Chr.1: 108228879-108228879 on Build GRCh38
rs1028368755 Chr.1: 108228858-108228858 on Build GRCh38
rs879104426 Chr.1: 108228995-108228995 on Build GRCh38
rs576531087 Chr.1: 108228653-108228653 on Build GRCh38
rs561956445 Chr.1: 108229036-108229036 on Build GRCh38
rs9435349 Chr.1: 108228756-108228756 on Build GRCh38
rs768890940 Chr.1: 108228947-108228947 on Build GRCh38
rs751304825 Chr.1: 108228876-108228876 on Build GRCh38
rs1005646808 Chr.1: 108228904-108228904 on Build GRCh38
rs556098844 Chr.1: 108228905-108228905 on Build GRCh38
rs879098065 Chr.1: 108228836-108228836 on Build GRCh38
rs368581022 Chr.1: 108229088-108229088 on Build GRCh38
rs541795275 Chr.1: 108228988-108228988 on Build GRCh38
rs772640925 Chr.1: 108228989-108228989 on Build GRCh38
rs539477752 Chr.1: 108228682-108228682 on Build GRCh38
rs576165846 Chr.1: 108228906-108228906 on Build GRCh38
rs376298071 Chr.1: 108229042-108229042 on Build GRCh38
rs759138893 Chr.1: 108229037-108229037 on Build GRCh38
rs867621671 Chr.1: 108228668-108228668 on Build GRCh38
rs934803871 Chr.1: 108228666-108228666 on Build GRCh38
rs1017394222 Chr.1: 108228933-108228933 on Build GRCh38
rs770667387 Chr.1: 108229006-108229006 on Build GRCh38
rs541440618 Chr.1: 108229089-108229089 on Build GRCh38
rs878914492 Chr.1: 108228855-108228855 on Build GRCh38
rs959969171 Chr.1: 108229064-108229064 on Build GRCh38

View Full Product Details