Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 112917919-112918448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC16A1
Entrez Gene ID
6566
Gene Name
solute carrier family 16 member 1
Gene Aliases
HHF7, MCT, MCT1, MCT1D
UniGene
Hs.75231
Gene Chromosome Location
Chr.1: 112911847-112956353 on Build GRCh38
Transcripts
NM_001166496.1 NM_003051.3 XM_011542026.2 XM_011542027.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4434493 c.474C>G p.L158L Chr.1: 112917932-112917932 on Build GRCh38
4434492 c.474C>G p.L158L Chr.1: 112917932-112917932 on Build GRCh38
4617099 c.432A>G p.P144P Chr.1: 112917974-112917974 on Build GRCh38
674295 c.383T>G p.L128W Chr.1: 112918023-112918023 on Build GRCh38
2120071 c.457G>T p.G153C Chr.1: 112917949-112917949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1018853094 Chr.1: 112918384-112918386 on Build GRCh38
rs756350441 Chr.1: 112918089-112918089 on Build GRCh38
rs149491709 Chr.1: 112918055-112918058 on Build GRCh38
rs781581888 Chr.1: 112918087-112918087 on Build GRCh38
rs549385060 Chr.1: 112918276-112918276 on Build GRCh38
rs1000886236 Chr.1: 112918214-112918214 on Build GRCh38
rs767161177 Chr.1: 112918086-112918088 on Build GRCh38
rs542005502 Chr.1: 112918073-112918073 on Build GRCh38
rs903904171 Chr.1: 112918182-112918182 on Build GRCh38
rs780430486 Chr.1: 112918092-112918092 on Build GRCh38
rs3849175 Chr.1: 112918406-112918406 on Build GRCh38
rs201021807 Chr.1: 112918065-112918065 on Build GRCh38
rs4084029 Chr.1: 112918395-112918395 on Build GRCh38
rs909762879 Chr.1: 112918105-112918107 on Build GRCh38
rs973785442 Chr.1: 112918420-112918420 on Build GRCh38
rs752724375 Chr.1: 112918239-112918239 on Build GRCh38
rs745794640 Chr.1: 112918298-112918298 on Build GRCh38
rs1020486548 Chr.1: 112918033-112918033 on Build GRCh38
rs569374211 Chr.1: 112918278-112918278 on Build GRCh38
rs762269279 Chr.1: 112918055-112918074 on Build GRCh38
rs370336565 Chr.1: 112918265-112918265 on Build GRCh38
rs763576399 Chr.1: 112918097-112918104 on Build GRCh38
rs752144977 Chr.1: 112918090-112918090 on Build GRCh38
rs3849174 Chr.1: 112918295-112918295 on Build GRCh38
rs1045263166 Chr.1: 112918114-112918114 on Build GRCh38
rs575973104 Chr.1: 112918096-112918103 on Build GRCh38
rs199817200 Chr.1: 112918055-112918062 on Build GRCh38
rs1344429 Chr.1: 112918061-112918061 on Build GRCh38
rs777192182 Chr.1: 112918055-112918066 on Build GRCh38
rs546303132 Chr.1: 112918042-112918042 on Build GRCh38
rs917787878 Chr.1: 112918154-112918154 on Build GRCh38
rs537084285 Chr.1: 112918210-112918210 on Build GRCh38
rs765615393 Chr.1: 112918055-112918078 on Build GRCh38
rs747535406 Chr.1: 112917996-112917996 on Build GRCh38
rs774344077 Chr.1: 112918063-112918063 on Build GRCh38
rs368776923 Chr.1: 112917973-112917973 on Build GRCh38
rs555135475 Chr.1: 112918251-112918251 on Build GRCh38
rs1017987999 Chr.1: 112918405-112918405 on Build GRCh38
rs201699455 Chr.1: 112918077-112918077 on Build GRCh38
rs752085204 Chr.1: 112918081-112918081 on Build GRCh38
rs201450570 Chr.1: 112918085-112918085 on Build GRCh38
rs1045174277 Chr.1: 112918158-112918158 on Build GRCh38
rs941222403 Chr.1: 112918106-112918106 on Build GRCh38
rs746074147 Chr.1: 112918024-112918024 on Build GRCh38
rs1052839832 Chr.1: 112918248-112918248 on Build GRCh38
rs577209023 Chr.1: 112917979-112917979 on Build GRCh38
rs1029313324 Chr.1: 112918408-112918408 on Build GRCh38
rs773587590 Chr.1: 112918057-112918058 on Build GRCh38
rs548325659 Chr.1: 112918294-112918294 on Build GRCh38
rs768676136 Chr.1: 112918055-112918070 on Build GRCh38
rs778256326 Chr.1: 112917984-112917984 on Build GRCh38
rs1057192932 Chr.1: 112918231-112918231 on Build GRCh38
rs922270290 Chr.1: 112918101-112918104 on Build GRCh38
rs759147371 Chr.1: 112918058-112918059 on Build GRCh38
rs547712239 Chr.1: 112918165-112918165 on Build GRCh38
rs982651247 Chr.1: 112918104-112918104 on Build GRCh38
rs200417465 Chr.1: 112918069-112918069 on Build GRCh38
rs755603597 Chr.1: 112918095-112918101 on Build GRCh38
rs947864742 Chr.1: 112918156-112918156 on Build GRCh38
rs746243243 Chr.1: 112918088-112918088 on Build GRCh38
rs964102941 Chr.1: 112918099-112918101 on Build GRCh38
rs1020544803 Chr.1: 112918287-112918287 on Build GRCh38
rs781459840 Chr.1: 112918019-112918019 on Build GRCh38
rs767307620 Chr.1: 112918067-112918067 on Build GRCh38
rs757628694 Chr.1: 112918016-112918016 on Build GRCh38
rs749394895 Chr.1: 112918052-112918052 on Build GRCh38
rs867955609 Chr.1: 112918336-112918336 on Build GRCh38
rs905954420 Chr.1: 112918117-112918117 on Build GRCh38
rs757744686 Chr.1: 112918083-112918083 on Build GRCh38
rs755757976 Chr.1: 112917961-112917961 on Build GRCh38
rs571384818 Chr.1: 112918259-112918259 on Build GRCh38
rs756177576 Chr.1: 112918285-112918285 on Build GRCh38
rs140544033 Chr.1: 112918100-112918100 on Build GRCh38
rs547887211 Chr.1: 112918340-112918340 on Build GRCh38
rs749593174 Chr.1: 112918093-112918093 on Build GRCh38
rs772909432 Chr.1: 112918068-112918068 on Build GRCh38
rs563913396 Chr.1: 112918178-112918178 on Build GRCh38
rs149383630 Chr.1: 112917971-112917971 on Build GRCh38
rs535183835 Chr.1: 112918286-112918286 on Build GRCh38
rs929920171 Chr.1: 112918101-112918102 on Build GRCh38
rs764710681 Chr.1: 112918078-112918078 on Build GRCh38
rs770173083 Chr.1: 112918040-112918040 on Build GRCh38
rs150246870 Chr.1: 112917965-112917965 on Build GRCh38
rs781480885 Chr.1: 112918170-112918170 on Build GRCh38

View Full Product Details