Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 81263686-81264229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STON2
Entrez Gene ID
85439
Gene Name
stonin 2
Gene Aliases
STN2, STNB, STNB2
UniGene
Hs.14248
Gene Chromosome Location
Chr.14: 81260650-81427404 on Build GRCh38
Transcripts
NM_001256430.1 NM_033104.3 XM_006720279.1 XM_011537240.1 XM_011537242.1

SNP Detail

SNP ID Chromosome Location
rs974436004 Chr.14: 81264059-81264059 on Build GRCh38
rs946182179 Chr.14: 81264195-81264195 on Build GRCh38
rs578155271 Chr.14: 81264200-81264200 on Build GRCh38
rs776517644 Chr.14: 81263987-81263987 on Build GRCh38
rs941636813 Chr.14: 81263718-81263718 on Build GRCh38
rs143587183 Chr.14: 81264089-81264089 on Build GRCh38
rs148728781 Chr.14: 81263936-81263936 on Build GRCh38
rs528383532 Chr.14: 81263767-81263767 on Build GRCh38
rs1019361825 Chr.14: 81263758-81263758 on Build GRCh38
rs926159773 Chr.14: 81263895-81263895 on Build GRCh38
rs952676559 Chr.14: 81263780-81263780 on Build GRCh38
rs750489706 Chr.14: 81263982-81263982 on Build GRCh38
rs909000547 Chr.14: 81264005-81264005 on Build GRCh38
rs556591224 Chr.14: 81264194-81264194 on Build GRCh38
rs983201729 Chr.14: 81264004-81264004 on Build GRCh38
rs768609504 Chr.14: 81263955-81263955 on Build GRCh38
rs927105985 Chr.14: 81263788-81263788 on Build GRCh38
rs1026652926 Chr.14: 81264130-81264130 on Build GRCh38
rs561709535 Chr.14: 81263831-81263831 on Build GRCh38
rs556073185 Chr.14: 81264116-81264116 on Build GRCh38
rs75913204 Chr.14: 81264192-81264192 on Build GRCh38
rs545397609 Chr.14: 81263749-81263749 on Build GRCh38
rs551816807 Chr.14: 81263993-81263993 on Build GRCh38
rs946009460 Chr.14: 81263859-81263859 on Build GRCh38
rs566907766 Chr.14: 81264068-81264068 on Build GRCh38
rs564010637 Chr.14: 81263751-81263751 on Build GRCh38
rs533339818 Chr.14: 81263984-81263984 on Build GRCh38
rs913346295 Chr.14: 81263849-81263849 on Build GRCh38
rs771027099 Chr.14: 81264025-81264025 on Build GRCh38
rs921596670 Chr.14: 81264147-81264147 on Build GRCh38
rs749373762 Chr.14: 81264017-81264017 on Build GRCh38
rs1007923315 Chr.14: 81263719-81263719 on Build GRCh38
rs941750518 Chr.14: 81264045-81264045 on Build GRCh38
rs1043080714 Chr.14: 81263883-81263883 on Build GRCh38
rs1041178280 Chr.14: 81264030-81264030 on Build GRCh38
rs569379043 Chr.14: 81263977-81263977 on Build GRCh38
rs745530918 Chr.14: 81263888-81263888 on Build GRCh38
rs1038716494 Chr.14: 81263737-81263737 on Build GRCh38
rs1010292870 Chr.14: 81263824-81263824 on Build GRCh38
rs961314983 Chr.14: 81263764-81263764 on Build GRCh38
rs938535800 Chr.14: 81263793-81263793 on Build GRCh38
rs540552967 Chr.14: 81263776-81263776 on Build GRCh38
rs992896293 Chr.14: 81263812-81263812 on Build GRCh38
rs904536844 Chr.14: 81263880-81263880 on Build GRCh38
rs950497443 Chr.14: 81263994-81263994 on Build GRCh38
rs189158709 Chr.14: 81263908-81263908 on Build GRCh38
rs774499327 Chr.14: 81264035-81264035 on Build GRCh38
rs994212414 Chr.14: 81264128-81264128 on Build GRCh38
rs757922464 Chr.14: 81263898-81263898 on Build GRCh38
rs979890944 Chr.14: 81263781-81263781 on Build GRCh38
rs890619588 Chr.14: 81263939-81263939 on Build GRCh38
rs574851925 Chr.14: 81263727-81263727 on Build GRCh38
rs933586392 Chr.14: 81264168-81264168 on Build GRCh38
rs181976980 Chr.14: 81264153-81264153 on Build GRCh38
rs371991868 Chr.14: 81263947-81263947 on Build GRCh38

View Full Product Details