Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 23002916-23003352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF728
Entrez Gene ID
388523
Gene Name
zinc finger protein 728
Gene Aliases
-
UniGene
Hs.616766
Gene Chromosome Location
Chr.19: 22974883-23003208 on Build GRCh38
Transcripts
NM_001267716.1

SNP Detail

SNP ID Chromosome Location
rs1007219059 Chr.19: 23003043-23003043 on Build GRCh38
rs760992529 Chr.19: 23002991-23002991 on Build GRCh38
rs771299298 Chr.19: 23003064-23003064 on Build GRCh38
rs950060393 Chr.19: 23002971-23002971 on Build GRCh38
rs1049186477 Chr.19: 23003234-23003234 on Build GRCh38
rs111427747 Chr.19: 23003048-23003048 on Build GRCh38
rs201357331 Chr.19: 23003329-23003329 on Build GRCh38
rs777030720 Chr.19: 23003070-23003070 on Build GRCh38
rs562228189 Chr.19: 23003107-23003107 on Build GRCh38
rs569736505 Chr.19: 23003195-23003195 on Build GRCh38
rs998064039 Chr.19: 23003312-23003312 on Build GRCh38
rs529499967 Chr.19: 23003133-23003133 on Build GRCh38
rs958926164 Chr.19: 23003131-23003131 on Build GRCh38
rs1019908700 Chr.19: 23003117-23003117 on Build GRCh38
rs535148860 Chr.19: 23003316-23003316 on Build GRCh38
rs551391319 Chr.19: 23003232-23003232 on Build GRCh38
rs369090986 Chr.19: 23003145-23003145 on Build GRCh38
rs115347083 Chr.19: 23003054-23003054 on Build GRCh38
rs747196524 Chr.19: 23003062-23003062 on Build GRCh38
rs556205331 Chr.19: 23003149-23003149 on Build GRCh38
rs180943528 Chr.19: 23003272-23003272 on Build GRCh38
rs535690855 Chr.19: 23003324-23003324 on Build GRCh38
rs549922920 Chr.19: 23003055-23003055 on Build GRCh38
rs917478060 Chr.19: 23003139-23003139 on Build GRCh38
rs769405811 Chr.19: 23003275-23003275 on Build GRCh38
rs777073089 Chr.19: 23002945-23002945 on Build GRCh38
rs775634137 Chr.19: 23003160-23003160 on Build GRCh38
rs1000334976 Chr.19: 23003330-23003330 on Build GRCh38
rs748247387 Chr.19: 23003051-23003051 on Build GRCh38
rs967496099 Chr.19: 23003119-23003119 on Build GRCh38
rs1019898839 Chr.19: 23003313-23003313 on Build GRCh38
rs760038754 Chr.19: 23003001-23003001 on Build GRCh38
rs566404167 Chr.19: 23003177-23003177 on Build GRCh38
rs759854146 Chr.19: 23003072-23003072 on Build GRCh38
rs777387077 Chr.19: 23003017-23003017 on Build GRCh38
rs752220171 Chr.19: 23003011-23003011 on Build GRCh38
rs774557093 Chr.19: 23003197-23003197 on Build GRCh38
rs540890773 Chr.19: 23003214-23003214 on Build GRCh38
rs772420406 Chr.19: 23003126-23003126 on Build GRCh38
rs765712024 Chr.19: 23003002-23003002 on Build GRCh38
rs756051934 Chr.19: 23003046-23003046 on Build GRCh38
rs772420251 Chr.19: 23002982-23002982 on Build GRCh38
rs897889746 Chr.19: 23002994-23002994 on Build GRCh38
rs143565543 Chr.19: 23003207-23003207 on Build GRCh38
rs766784823 Chr.19: 23002993-23002993 on Build GRCh38
rs564152302 Chr.19: 23003037-23003037 on Build GRCh38
rs773627102 Chr.19: 23002988-23002988 on Build GRCh38
rs555021336 Chr.19: 23003317-23003317 on Build GRCh38
rs1039527986 Chr.19: 23003285-23003285 on Build GRCh38
rs888261301 Chr.19: 23003241-23003241 on Build GRCh38
rs978568143 Chr.19: 23003120-23003120 on Build GRCh38
rs754183096 Chr.19: 23002998-23002998 on Build GRCh38
rs917235358 Chr.19: 23002967-23002967 on Build GRCh38
rs751152431 Chr.19: 23003025-23003025 on Build GRCh38
rs902887122 Chr.19: 23003322-23003322 on Build GRCh38
rs867332229 Chr.19: 23003094-23003094 on Build GRCh38
rs537114846 Chr.19: 23003294-23003294 on Build GRCh38
rs919268690 Chr.19: 23003203-23003203 on Build GRCh38
rs1056089187 Chr.19: 23002951-23002951 on Build GRCh38
rs547726984 Chr.19: 23003143-23003143 on Build GRCh38
rs545820766 Chr.19: 23003012-23003012 on Build GRCh38
rs745702376 Chr.19: 23003042-23003042 on Build GRCh38
rs571940426 Chr.19: 23002941-23002941 on Build GRCh38
rs558452009 Chr.19: 23003036-23003036 on Build GRCh38
rs773089784 Chr.19: 23003309-23003309 on Build GRCh38

View Full Product Details