Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 126465576-126466091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100128554
Entrez Gene ID
100128554
Gene Name
uncharacterized LOC100128554
Gene Aliases
-
UniGene
Hs.408255
Gene Chromosome Location
Chr.12: 126442481-126472785 on Build GRCh38
Transcripts
NR_130748.1

SNP Detail

SNP ID Chromosome Location
rs12307292 Chr.12: 126465971-126465971 on Build GRCh38
rs770957621 Chr.12: 126465809-126465809 on Build GRCh38
rs574557914 Chr.12: 126465636-126465636 on Build GRCh38
rs1050215896 Chr.12: 126465736-126465736 on Build GRCh38
rs754122273 Chr.12: 126466013-126466013 on Build GRCh38
rs548848158 Chr.12: 126465962-126465962 on Build GRCh38
rs897836715 Chr.12: 126465804-126465804 on Build GRCh38
rs905392387 Chr.12: 126465652-126465652 on Build GRCh38
rs1035103419 Chr.12: 126465724-126465724 on Build GRCh38
rs527846058 Chr.12: 126465972-126465972 on Build GRCh38
rs923010966 Chr.12: 126465718-126465718 on Build GRCh38
rs777698297 Chr.12: 126465712-126465712 on Build GRCh38
rs116170628 Chr.12: 126465841-126465841 on Build GRCh38
rs933157245 Chr.12: 126465722-126465722 on Build GRCh38
rs963902984 Chr.12: 126466051-126466051 on Build GRCh38
rs545767814 Chr.12: 126465810-126465810 on Build GRCh38
rs1009677566 Chr.12: 126465982-126465982 on Build GRCh38
rs988153386 Chr.12: 126465733-126465733 on Build GRCh38
rs552598737 Chr.12: 126465979-126465979 on Build GRCh38
rs756350826 Chr.12: 126466026-126466026 on Build GRCh38
rs976627685 Chr.12: 126466056-126466056 on Build GRCh38
rs929758236 Chr.12: 126465863-126465863 on Build GRCh38
rs564363020 Chr.12: 126465839-126465839 on Build GRCh38
rs149214113 Chr.12: 126465911-126465911 on Build GRCh38
rs910366503 Chr.12: 126465989-126465989 on Build GRCh38
rs115201623 Chr.12: 126465988-126465988 on Build GRCh38
rs1037729588 Chr.12: 126465768-126465768 on Build GRCh38
rs1003730017 Chr.12: 126465965-126465965 on Build GRCh38
rs528976888 Chr.12: 126465945-126465945 on Build GRCh38
rs919149784 Chr.12: 126466035-126466035 on Build GRCh38
rs572349727 Chr.12: 126465781-126465781 on Build GRCh38
rs10847164 Chr.12: 126465756-126465756 on Build GRCh38
rs780669218 Chr.12: 126466028-126466028 on Build GRCh38
rs929303008 Chr.12: 126466036-126466036 on Build GRCh38
rs543410320 Chr.12: 126465876-126465876 on Build GRCh38
rs1033841974 Chr.12: 126465842-126465842 on Build GRCh38
rs922464805 Chr.12: 126466058-126466058 on Build GRCh38
rs889065394 Chr.12: 126465744-126465744 on Build GRCh38
rs956752837 Chr.12: 126465732-126465732 on Build GRCh38
rs11058670 Chr.12: 126465618-126465618 on Build GRCh38
rs911096902 Chr.12: 126465783-126465783 on Build GRCh38
rs571159577 Chr.12: 126466011-126466011 on Build GRCh38
rs565243909 Chr.12: 126465806-126465806 on Build GRCh38
rs551180653 Chr.12: 126465831-126465831 on Build GRCh38
rs977075283 Chr.12: 126465958-126465958 on Build GRCh38
rs541655828 Chr.12: 126465755-126465755 on Build GRCh38
rs73416858 Chr.12: 126466066-126466066 on Build GRCh38
rs990121651 Chr.12: 126465918-126465918 on Build GRCh38
rs371816566 Chr.12: 126465940-126465940 on Build GRCh38
rs1024303347 Chr.12: 126465642-126465642 on Build GRCh38
rs941842735 Chr.12: 126465990-126465990 on Build GRCh38

View Full Product Details