Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 30876365-30876793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COCH
Entrez Gene ID
1690
Gene Name
cochlin
Gene Aliases
COCH-5B2, COCH5B2, DFNA9
UniGene
Hs.21016
Gene Chromosome Location
Chr.14: 30874496-30890618 on Build GRCh38
Transcripts
NM_001135058.1 NM_004086.2 XM_017021071.1 XM_017021072.1 NR_038356.1

Gene Symbol
LOC100506071
Entrez Gene ID
100506071
Gene Name
uncharacterized LOC100506071
Gene Aliases
-
UniGene
Hs.729195
Gene Chromosome Location
Chr.14: 30876179-30889808 on Build GRCh38
Transcripts
NM_001135058.1 NM_004086.2 XM_017021071.1 XM_017021072.1 NR_038356.1

SNP Detail

SNP ID Chromosome Location
rs923892156 Chr.14: 30876650-30876650 on Build GRCh38
rs1045086496 Chr.14: 30876485-30876485 on Build GRCh38
rs568873512 Chr.14: 30876721-30876721 on Build GRCh38
rs139143424 Chr.14: 30876508-30876508 on Build GRCh38
rs993366627 Chr.14: 30876497-30876497 on Build GRCh38
rs1053998548 Chr.14: 30876422-30876422 on Build GRCh38
rs952479923 Chr.14: 30876583-30876583 on Build GRCh38
rs986141747 Chr.14: 30876600-30876600 on Build GRCh38
rs142247487 Chr.14: 30876685-30876685 on Build GRCh38
rs772425844 Chr.14: 30876460-30876460 on Build GRCh38
rs1001186634 Chr.14: 30876415-30876415 on Build GRCh38
rs1027192014 Chr.14: 30876393-30876393 on Build GRCh38
rs140224407 Chr.14: 30876703-30876704 on Build GRCh38
rs568627216 Chr.14: 30876602-30876602 on Build GRCh38
rs147616570 Chr.14: 30876536-30876536 on Build GRCh38
rs534267228 Chr.14: 30876584-30876584 on Build GRCh38
rs541815818 Chr.14: 30876400-30876400 on Build GRCh38
rs995029162 Chr.14: 30876538-30876538 on Build GRCh38
rs563702017 Chr.14: 30876436-30876436 on Build GRCh38
rs935209072 Chr.14: 30876670-30876670 on Build GRCh38
rs760788682 Chr.14: 30876683-30876683 on Build GRCh38
rs547238332 Chr.14: 30876550-30876550 on Build GRCh38
rs943936416 Chr.14: 30876639-30876639 on Build GRCh38
rs539130116 Chr.14: 30876733-30876733 on Build GRCh38
rs145194337 Chr.14: 30876535-30876535 on Build GRCh38
rs557457734 Chr.14: 30876765-30876765 on Build GRCh38
rs967984665 Chr.14: 30876530-30876530 on Build GRCh38
rs976799219 Chr.14: 30876612-30876612 on Build GRCh38
rs556638896 Chr.14: 30876712-30876712 on Build GRCh38
rs373492607 Chr.14: 30876389-30876389 on Build GRCh38
rs948175810 Chr.14: 30876700-30876700 on Build GRCh38
rs8005851 Chr.14: 30876477-30876477 on Build GRCh38
rs965054809 Chr.14: 30876610-30876610 on Build GRCh38
rs1004233198 Chr.14: 30876517-30876517 on Build GRCh38

View Full Product Details