Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 58265317-58265822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMA3
Entrez Gene ID
5684
Gene Name
proteasome subunit alpha 3
Gene Aliases
HC8, PSC3
UniGene
Hs.558799
Gene Chromosome Location
Chr.14: 58244805-58272009 on Build GRCh38
Transcripts
NM_002788.3 NM_152132.2 NR_038123.1 NR_029434.1 NR_029435.1

Gene Symbol
PSMA3-AS1
Entrez Gene ID
379025
Gene Name
PSMA3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.531089
Gene Chromosome Location
Chr.14: 58265365-58298137 on Build GRCh38
Transcripts
NM_002788.3 NM_152132.2 NR_038123.1 NR_029434.1 NR_029435.1

SNP Detail

SNP ID Chromosome Location
rs555317436 Chr.14: 58265402-58265402 on Build GRCh38
rs747562558 Chr.14: 58265404-58265404 on Build GRCh38
rs1010100942 Chr.14: 58265757-58265757 on Build GRCh38
rs781722280 Chr.14: 58265384-58265387 on Build GRCh38
rs796739380 Chr.14: 58265582-58265582 on Build GRCh38
rs1047292522 Chr.14: 58265583-58265583 on Build GRCh38
rs1033171646 Chr.14: 58265732-58265732 on Build GRCh38
rs920409668 Chr.14: 58265561-58265562 on Build GRCh38
rs1001726061 Chr.14: 58265707-58265707 on Build GRCh38
rs148619470 Chr.14: 58265747-58265747 on Build GRCh38
rs758892962 Chr.14: 58265350-58265350 on Build GRCh38
rs113929164 Chr.14: 58265690-58265691 on Build GRCh38
rs141195663 Chr.14: 58265753-58265753 on Build GRCh38
rs747333587 Chr.14: 58265347-58265357 on Build GRCh38
rs748345712 Chr.14: 58265691-58265691 on Build GRCh38
rs913171239 Chr.14: 58265437-58265437 on Build GRCh38
rs930756826 Chr.14: 58265587-58265587 on Build GRCh38
rs943262385 Chr.14: 58265552-58265552 on Build GRCh38
rs781156324 Chr.14: 58265519-58265519 on Build GRCh38
rs773538281 Chr.14: 58265730-58265730 on Build GRCh38
rs771207981 Chr.14: 58265644-58265648 on Build GRCh38
rs186234912 Chr.14: 58265460-58265460 on Build GRCh38
rs769796548 Chr.14: 58265714-58265714 on Build GRCh38
rs538150018 Chr.14: 58265521-58265521 on Build GRCh38
rs989623497 Chr.14: 58265518-58265518 on Build GRCh38
rs34096389 Chr.14: 58265545-58265546 on Build GRCh38
rs574026089 Chr.14: 58265469-58265469 on Build GRCh38
rs971790266 Chr.14: 58265449-58265449 on Build GRCh38
rs1032779676 Chr.14: 58265744-58265744 on Build GRCh38
rs1047751304 Chr.14: 58265369-58265370 on Build GRCh38
rs1014176197 Chr.14: 58265670-58265670 on Build GRCh38
rs987391484 Chr.14: 58265522-58265522 on Build GRCh38
rs930170227 Chr.14: 58265558-58265558 on Build GRCh38
rs556766466 Chr.14: 58265638-58265638 on Build GRCh38
rs1048250840 Chr.14: 58265602-58265602 on Build GRCh38
rs566589816 Chr.14: 58265345-58265345 on Build GRCh38
rs533951985 Chr.14: 58265401-58265401 on Build GRCh38
rs1008975581 Chr.14: 58265641-58265641 on Build GRCh38
rs987172369 Chr.14: 58265796-58265796 on Build GRCh38
rs988701703 Chr.14: 58265427-58265427 on Build GRCh38
rs1033140601 Chr.14: 58265441-58265441 on Build GRCh38
rs1010676477 Chr.14: 58265750-58265750 on Build GRCh38
rs868137304 Chr.14: 58265517-58265517 on Build GRCh38
rs543108340 Chr.14: 58265740-58265740 on Build GRCh38
rs920071870 Chr.14: 58265540-58265540 on Build GRCh38
rs966476924 Chr.14: 58265793-58265793 on Build GRCh38
rs143871576 Chr.14: 58265700-58265700 on Build GRCh38
rs899287422 Chr.14: 58265346-58265349 on Build GRCh38
rs557837444 Chr.14: 58265344-58265344 on Build GRCh38
rs1001691798 Chr.14: 58265410-58265410 on Build GRCh38
rs1019100143 Chr.14: 58265393-58265393 on Build GRCh38
rs911843890 Chr.14: 58265541-58265541 on Build GRCh38
rs755047275 Chr.14: 58265477-58265477 on Build GRCh38
rs1039998410 Chr.14: 58265655-58265655 on Build GRCh38
rs867134902 Chr.14: 58265568-58265568 on Build GRCh38
rs964529695 Chr.14: 58265395-58265395 on Build GRCh38
rs981505832 Chr.14: 58265457-58265457 on Build GRCh38

View Full Product Details