Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70761052-70761560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VAC14
Entrez Gene ID
55697
Gene Name
Vac14, PIKFYVE complex component
Gene Aliases
ArPIKfyve, TAX1BP2, TRX
UniGene
Hs.445061
Gene Chromosome Location
Chr.16: 70687439-70801169 on Build GRCh38
Transcripts
NM_018052.3 XM_005256038.3 XM_005256041.3 XM_011523223.2 XM_011523224.2 XM_011523225.2 XM_017023441.1 XM_017023442.1 NR_034083.2

Gene Symbol
VAC14-AS1
Entrez Gene ID
100130894
Gene Name
VAC14 antisense RNA 1
Gene Aliases
-
UniGene
Hs.678899
Gene Chromosome Location
Chr.16: 70755060-70773251 on Build GRCh38
Transcripts
NM_018052.3 XM_005256038.3 XM_005256041.3 XM_011523223.2 XM_011523224.2 XM_011523225.2 XM_017023441.1 XM_017023442.1 NR_034083.2

SNP Detail

SNP ID Chromosome Location
rs111280326 Chr.16: 70761520-70761520 on Build GRCh38
rs923320723 Chr.16: 70761187-70761187 on Build GRCh38
rs577469642 Chr.16: 70761225-70761225 on Build GRCh38
rs1026561705 Chr.16: 70761287-70761287 on Build GRCh38
rs950681681 Chr.16: 70761096-70761096 on Build GRCh38
rs978290259 Chr.16: 70761448-70761448 on Build GRCh38
rs748613929 Chr.16: 70761080-70761080 on Build GRCh38
rs368808482 Chr.16: 70761198-70761198 on Build GRCh38
rs113712448 Chr.16: 70761444-70761444 on Build GRCh38
rs1016148871 Chr.16: 70761332-70761332 on Build GRCh38
rs934048646 Chr.16: 70761397-70761397 on Build GRCh38
rs779346337 Chr.16: 70761202-70761202 on Build GRCh38
rs1011082558 Chr.16: 70761174-70761174 on Build GRCh38
rs974833450 Chr.16: 70761182-70761182 on Build GRCh38
rs777654745 Chr.16: 70761389-70761389 on Build GRCh38
rs924100234 Chr.16: 70761460-70761460 on Build GRCh38
rs890068821 Chr.16: 70761099-70761099 on Build GRCh38
rs974717290 Chr.16: 70761390-70761390 on Build GRCh38
rs950702335 Chr.16: 70761323-70761323 on Build GRCh38
rs540839956 Chr.16: 70761121-70761121 on Build GRCh38
rs201751546 Chr.16: 70761111-70761111 on Build GRCh38
rs544900349 Chr.16: 70761286-70761286 on Build GRCh38
rs562010118 Chr.16: 70761443-70761443 on Build GRCh38
rs145956487 Chr.16: 70761381-70761381 on Build GRCh38
rs943345142 Chr.16: 70761179-70761179 on Build GRCh38
rs373523336 Chr.16: 70761271-70761271 on Build GRCh38
rs551725955 Chr.16: 70761195-70761195 on Build GRCh38
rs1002692903 Chr.16: 70761276-70761276 on Build GRCh38
rs911225470 Chr.16: 70761427-70761427 on Build GRCh38
rs959320929 Chr.16: 70761281-70761281 on Build GRCh38
rs1026214238 Chr.16: 70761090-70761090 on Build GRCh38
rs756360509 Chr.16: 70761127-70761127 on Build GRCh38
rs563285483 Chr.16: 70761343-70761343 on Build GRCh38
rs749701971 Chr.16: 70761162-70761162 on Build GRCh38
rs555153778 Chr.16: 70761078-70761078 on Build GRCh38
rs985116513 Chr.16: 70761327-70761327 on Build GRCh38
rs895377728 Chr.16: 70761493-70761493 on Build GRCh38
rs184706307 Chr.16: 70761420-70761420 on Build GRCh38
rs1054451984 Chr.16: 70761485-70761485 on Build GRCh38
rs1022689789 Chr.16: 70761266-70761266 on Build GRCh38
rs933378521 Chr.16: 70761428-70761428 on Build GRCh38
rs762773837 Chr.16: 70761513-70761513 on Build GRCh38
rs906945927 Chr.16: 70761272-70761272 on Build GRCh38
rs892794050 Chr.16: 70761247-70761247 on Build GRCh38
rs771414896 Chr.16: 70761292-70761292 on Build GRCh38
rs1054670039 Chr.16: 70761139-70761139 on Build GRCh38
rs756222450 Chr.16: 70761164-70761164 on Build GRCh38
rs1054164957 Chr.16: 70761236-70761236 on Build GRCh38
rs1007132488 Chr.16: 70761135-70761135 on Build GRCh38
rs573708646 Chr.16: 70761085-70761085 on Build GRCh38
rs12917867 Chr.16: 70761109-70761109 on Build GRCh38
rs748701528 Chr.16: 70761285-70761285 on Build GRCh38
rs936908722 Chr.16: 70761234-70761234 on Build GRCh38
rs1012500949 Chr.16: 70761250-70761250 on Build GRCh38
rs529407279 Chr.16: 70761487-70761487 on Build GRCh38
rs769436053 Chr.16: 70761473-70761473 on Build GRCh38
rs376564770 Chr.16: 70761161-70761161 on Build GRCh38
rs7188712 Chr.16: 70761499-70761499 on Build GRCh38
rs1044029276 Chr.16: 70761500-70761500 on Build GRCh38
rs962535106 Chr.16: 70761335-70761335 on Build GRCh38
rs1042544429 Chr.16: 70761482-70761482 on Build GRCh38
rs966797752 Chr.16: 70761189-70761189 on Build GRCh38
rs750580360 Chr.16: 70761110-70761110 on Build GRCh38

View Full Product Details