Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1267706-1268210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIRBP-AS1
Entrez Gene ID
148046
Gene Name
CIRBP antisense RNA 1
Gene Aliases
C19orf23
UniGene
Hs.438829
Gene Chromosome Location
Chr.19: 1267471-1270260 on Build GRCh38
Transcripts
NR_027271.1

SNP Detail

SNP ID Chromosome Location
rs1043034299 Chr.19: 1268042-1268042 on Build GRCh38
rs998992516 Chr.19: 1267809-1267809 on Build GRCh38
rs968461959 Chr.19: 1268185-1268185 on Build GRCh38
rs933878930 Chr.19: 1267743-1267743 on Build GRCh38
rs1006805892 Chr.19: 1268088-1268088 on Build GRCh38
rs532216542 Chr.19: 1267819-1267822 on Build GRCh38
rs1018227976 Chr.19: 1268128-1268128 on Build GRCh38
rs184814656 Chr.19: 1268104-1268104 on Build GRCh38
rs767740524 Chr.19: 1267758-1267758 on Build GRCh38
rs934424202 Chr.19: 1267819-1267820 on Build GRCh38
rs12975875 Chr.19: 1268179-1268179 on Build GRCh38
rs933932297 Chr.19: 1267974-1267974 on Build GRCh38
rs563583611 Chr.19: 1267886-1267886 on Build GRCh38
rs1014389760 Chr.19: 1268191-1268191 on Build GRCh38
rs979774478 Chr.19: 1267963-1267963 on Build GRCh38
rs770517477 Chr.19: 1267769-1267769 on Build GRCh38
rs993483972 Chr.19: 1267839-1267839 on Build GRCh38
rs150588322 Chr.19: 1267734-1267734 on Build GRCh38
rs988654213 Chr.19: 1268172-1268172 on Build GRCh38
rs1050117956 Chr.19: 1267979-1267979 on Build GRCh38
rs923120233 Chr.19: 1267913-1267913 on Build GRCh38
rs952697214 Chr.19: 1267903-1267903 on Build GRCh38
rs374824458 Chr.19: 1267905-1267905 on Build GRCh38
rs956093855 Chr.19: 1267929-1267929 on Build GRCh38
rs1022663366 Chr.19: 1267889-1267889 on Build GRCh38
rs28468475 Chr.19: 1268075-1268075 on Build GRCh38
rs906128198 Chr.19: 1267881-1267881 on Build GRCh38
rs983762558 Chr.19: 1267901-1267901 on Build GRCh38
rs914340608 Chr.19: 1267947-1267947 on Build GRCh38
rs1053255187 Chr.19: 1268124-1268124 on Build GRCh38
rs889109717 Chr.19: 1267751-1267751 on Build GRCh38
rs960525116 Chr.19: 1268187-1268187 on Build GRCh38
rs922428503 Chr.19: 1267954-1267954 on Build GRCh38
rs988558454 Chr.19: 1267931-1267931 on Build GRCh38
rs914225709 Chr.19: 1267829-1267829 on Build GRCh38
rs776293953 Chr.19: 1268106-1268106 on Build GRCh38
rs947057729 Chr.19: 1267840-1267840 on Build GRCh38
rs575563109 Chr.19: 1267910-1267910 on Build GRCh38
rs748266180 Chr.19: 1267821-1267822 on Build GRCh38
rs888537877 Chr.19: 1268072-1268072 on Build GRCh38
rs764498871 Chr.19: 1268033-1268033 on Build GRCh38
rs528666863 Chr.19: 1267883-1267883 on Build GRCh38
rs564400559 Chr.19: 1267789-1267789 on Build GRCh38
rs961494815 Chr.19: 1267765-1267765 on Build GRCh38
rs994255763 Chr.19: 1267892-1267892 on Build GRCh38
rs1053310986 Chr.19: 1267824-1267824 on Build GRCh38
rs976443786 Chr.19: 1267911-1267911 on Build GRCh38
rs1017440270 Chr.19: 1267907-1267907 on Build GRCh38
rs960303635 Chr.19: 1267818-1267818 on Build GRCh38
rs997427491 Chr.19: 1268144-1268144 on Build GRCh38
rs761007322 Chr.19: 1267950-1267950 on Build GRCh38
rs1057304892 Chr.19: 1267890-1267890 on Build GRCh38
rs577742551 Chr.19: 1267797-1267797 on Build GRCh38
rs979889359 Chr.19: 1268189-1268189 on Build GRCh38
rs889671263 Chr.19: 1267992-1267992 on Build GRCh38
rs955874358 Chr.19: 1268157-1268157 on Build GRCh38
rs938946206 Chr.19: 1267971-1267971 on Build GRCh38
rs984659974 Chr.19: 1267796-1267796 on Build GRCh38
rs546790355 Chr.19: 1268027-1268027 on Build GRCh38
rs909640177 Chr.19: 1267904-1267904 on Build GRCh38
rs930160946 Chr.19: 1268053-1268053 on Build GRCh38
rs1027057073 Chr.19: 1267900-1267901 on Build GRCh38
rs1031788155 Chr.19: 1267813-1267813 on Build GRCh38
rs901203310 Chr.19: 1268132-1268132 on Build GRCh38
rs545188116 Chr.19: 1267850-1267850 on Build GRCh38
rs1026985398 Chr.19: 1267785-1267785 on Build GRCh38
rs1044646761 Chr.19: 1267843-1267843 on Build GRCh38
rs994537690 Chr.19: 1267782-1267782 on Build GRCh38
rs117823231 Chr.19: 1268071-1268071 on Build GRCh38
rs943383209 Chr.19: 1268031-1268031 on Build GRCh38
rs1030684991 Chr.19: 1268152-1268152 on Build GRCh38
rs1051812784 Chr.19: 1267749-1267749 on Build GRCh38
rs904593779 Chr.19: 1267792-1267792 on Build GRCh38
rs970134414 Chr.19: 1267891-1267891 on Build GRCh38

View Full Product Details