Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38450211-38450706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNHG11
Entrez Gene ID
128439
Gene Name
small nucleolar RNA host gene 11
Gene Aliases
C20orf198, LINC00101, NCRNA00101
UniGene
Hs.16936
Gene Chromosome Location
Chr.20: 38446654-38450921 on Build GRCh38
Transcripts
NR_003239.1

SNP Detail

SNP ID Chromosome Location
rs948398086 Chr.20: 38450566-38450566 on Build GRCh38
rs866818537 Chr.20: 38450341-38450341 on Build GRCh38
rs559543102 Chr.20: 38450424-38450424 on Build GRCh38
rs923324233 Chr.20: 38450323-38450323 on Build GRCh38
rs937540529 Chr.20: 38450636-38450636 on Build GRCh38
rs1017501816 Chr.20: 38450634-38450634 on Build GRCh38
rs141926008 Chr.20: 38450595-38450595 on Build GRCh38
rs541533532 Chr.20: 38450504-38450504 on Build GRCh38
rs551823541 Chr.20: 38450619-38450619 on Build GRCh38
rs765981222 Chr.20: 38450275-38450275 on Build GRCh38
rs906933152 Chr.20: 38450588-38450588 on Build GRCh38
rs969761531 Chr.20: 38450652-38450652 on Build GRCh38
rs1011488888 Chr.20: 38450289-38450289 on Build GRCh38
rs1045391420 Chr.20: 38450567-38450567 on Build GRCh38
rs776177409 Chr.20: 38450291-38450291 on Build GRCh38
rs892961656 Chr.20: 38450614-38450614 on Build GRCh38
rs978739907 Chr.20: 38450350-38450350 on Build GRCh38
rs963919650 Chr.20: 38450420-38450420 on Build GRCh38
rs1045445502 Chr.20: 38450237-38450237 on Build GRCh38
rs147597373 Chr.20: 38450436-38450436 on Build GRCh38
rs12193 Chr.20: 38450387-38450387 on Build GRCh38
rs540943852 Chr.20: 38450423-38450423 on Build GRCh38
rs916849051 Chr.20: 38450492-38450492 on Build GRCh38
rs186797557 Chr.20: 38450407-38450407 on Build GRCh38
rs1031478957 Chr.20: 38450274-38450274 on Build GRCh38
rs906078064 Chr.20: 38450613-38450613 on Build GRCh38
rs933363981 Chr.20: 38450453-38450453 on Build GRCh38
rs1055375517 Chr.20: 38450487-38450487 on Build GRCh38
rs1019935241 Chr.20: 38450265-38450265 on Build GRCh38
rs1011852574 Chr.20: 38450628-38450628 on Build GRCh38
rs765148166 Chr.20: 38450321-38450321 on Build GRCh38
rs909381285 Chr.20: 38450432-38450432 on Build GRCh38
rs906987121 Chr.20: 38450252-38450252 on Build GRCh38
rs12397 Chr.20: 38450594-38450594 on Build GRCh38
rs368419694 Chr.20: 38450320-38450320 on Build GRCh38
rs551721056 Chr.20: 38450633-38450633 on Build GRCh38
rs1037993628 Chr.20: 38450611-38450611 on Build GRCh38
rs1057197342 Chr.20: 38450646-38450646 on Build GRCh38
rs762703683 Chr.20: 38450373-38450373 on Build GRCh38
rs965005225 Chr.20: 38450648-38450648 on Build GRCh38
rs998622695 Chr.20: 38450272-38450272 on Build GRCh38

View Full Product Details