Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49549031-49549529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSN-AS2
Entrez Gene ID
100132677
Gene Name
BSN antisense RNA 2 (head to head)
Gene Aliases
-
UniGene
Hs.435651
Gene Chromosome Location
Chr.3: 49549306-49554366 on Build GRCh38
Transcripts
NR_038866.1

SNP Detail

SNP ID Chromosome Location
rs898615935 Chr.3: 49549156-49549157 on Build GRCh38
rs1028711456 Chr.3: 49549280-49549280 on Build GRCh38
rs774742680 Chr.3: 49549401-49549401 on Build GRCh38
rs779858862 Chr.3: 49549060-49549060 on Build GRCh38
rs890159176 Chr.3: 49549468-49549468 on Build GRCh38
rs902671964 Chr.3: 49549404-49549404 on Build GRCh38
rs75470186 Chr.3: 49549267-49549267 on Build GRCh38
rs554690242 Chr.3: 49549079-49549079 on Build GRCh38
rs920390644 Chr.3: 49549194-49549194 on Build GRCh38
rs780997661 Chr.3: 49549162-49549162 on Build GRCh38
rs142853520 Chr.3: 49549107-49549107 on Build GRCh38
rs940246876 Chr.3: 49549445-49549445 on Build GRCh38
rs995597901 Chr.3: 49549157-49549157 on Build GRCh38
rs999184489 Chr.3: 49549240-49549240 on Build GRCh38
rs1031817791 Chr.3: 49549464-49549464 on Build GRCh38
rs973230409 Chr.3: 49549156-49549156 on Build GRCh38
rs925074052 Chr.3: 49549085-49549085 on Build GRCh38
rs932811204 Chr.3: 49549128-49549128 on Build GRCh38
rs769631435 Chr.3: 49549210-49549210 on Build GRCh38
rs921436242 Chr.3: 49549125-49549125 on Build GRCh38
rs529814586 Chr.3: 49549396-49549396 on Build GRCh38
rs148961138 Chr.3: 49549063-49549063 on Build GRCh38
rs541306555 Chr.3: 49549238-49549238 on Build GRCh38
rs964571119 Chr.3: 49549145-49549145 on Build GRCh38
rs910459622 Chr.3: 49549124-49549124 on Build GRCh38
rs928026475 Chr.3: 49549218-49549218 on Build GRCh38
rs1024865148 Chr.3: 49549058-49549058 on Build GRCh38
rs760049023 Chr.3: 49549426-49549426 on Build GRCh38
rs564850754 Chr.3: 49549164-49549164 on Build GRCh38
rs754747169 Chr.3: 49549111-49549111 on Build GRCh38
rs946911843 Chr.3: 49549360-49549360 on Build GRCh38
rs554803996 Chr.3: 49549228-49549228 on Build GRCh38
rs1043923837 Chr.3: 49549403-49549403 on Build GRCh38
rs1055173575 Chr.3: 49549169-49549169 on Build GRCh38
rs912798175 Chr.3: 49549138-49549138 on Build GRCh38
rs1051795220 Chr.3: 49549129-49549129 on Build GRCh38
rs986966115 Chr.3: 49549339-49549339 on Build GRCh38
rs569221710 Chr.3: 49549195-49549195 on Build GRCh38
rs777565809 Chr.3: 49549237-49549237 on Build GRCh38
rs968457263 Chr.3: 49549056-49549056 on Build GRCh38
rs146557783 Chr.3: 49549109-49549109 on Build GRCh38
rs972945909 Chr.3: 49549452-49549452 on Build GRCh38
rs919926238 Chr.3: 49549500-49549500 on Build GRCh38
rs940037845 Chr.3: 49549140-49549140 on Build GRCh38
rs1031978845 Chr.3: 49549260-49549260 on Build GRCh38
rs748815478 Chr.3: 49549321-49549321 on Build GRCh38
rs895417280 Chr.3: 49549293-49549293 on Build GRCh38

View Full Product Details