Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 3183101-3183628 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100507194
Entrez Gene ID
100507194
Gene Name
uncharacterized LOC100507194
Gene Aliases
-
UniGene
Hs.678053
Gene Chromosome Location
Chr.6: 3182817-3195767 on Build GRCh38
Transcripts
NR_038295.1 NR_038296.1

SNP Detail

SNP ID Chromosome Location
rs992734989 Chr.6: 3183434-3183434 on Build GRCh38
rs1018489413 Chr.6: 3183567-3183567 on Build GRCh38
rs535375175 Chr.6: 3183163-3183163 on Build GRCh38
rs908025201 Chr.6: 3183133-3183133 on Build GRCh38
rs376797981 Chr.6: 3183328-3183328 on Build GRCh38
rs939497272 Chr.6: 3183137-3183137 on Build GRCh38
rs935303451 Chr.6: 3183521-3183521 on Build GRCh38
rs77870136 Chr.6: 3183140-3183140 on Build GRCh38
rs1008468382 Chr.6: 3183267-3183267 on Build GRCh38
rs1008314141 Chr.6: 3183565-3183565 on Build GRCh38
rs547724355 Chr.6: 3183488-3183488 on Build GRCh38
rs773802270 Chr.6: 3183129-3183129 on Build GRCh38
rs920952435 Chr.6: 3183132-3183132 on Build GRCh38
rs550194207 Chr.6: 3183589-3183589 on Build GRCh38
rs1031112458 Chr.6: 3183231-3183231 on Build GRCh38
rs914775072 Chr.6: 3183395-3183395 on Build GRCh38
rs1026840730 Chr.6: 3183597-3183597 on Build GRCh38
rs986464462 Chr.6: 3183484-3183484 on Build GRCh38
rs187488977 Chr.6: 3183546-3183546 on Build GRCh38
rs899677989 Chr.6: 3183570-3183570 on Build GRCh38
rs567993170 Chr.6: 3183499-3183502 on Build GRCh38
rs191872905 Chr.6: 3183253-3183253 on Build GRCh38
rs572090535 Chr.6: 3183247-3183247 on Build GRCh38
rs576554076 Chr.6: 3183174-3183174 on Build GRCh38
rs903834197 Chr.6: 3183492-3183492 on Build GRCh38
rs990300747 Chr.6: 3183391-3183391 on Build GRCh38
rs543347812 Chr.6: 3183347-3183347 on Build GRCh38
rs1051024318 Chr.6: 3183148-3183148 on Build GRCh38
rs139888036 Chr.6: 3183432-3183432 on Build GRCh38
rs539650535 Chr.6: 3183130-3183130 on Build GRCh38
rs916758695 Chr.6: 3183442-3183442 on Build GRCh38
rs78292071 Chr.6: 3183296-3183296 on Build GRCh38
rs765247150 Chr.6: 3183448-3183448 on Build GRCh38
rs540168860 Chr.6: 3183588-3183588 on Build GRCh38
rs928360215 Chr.6: 3183406-3183406 on Build GRCh38
rs11753978 Chr.6: 3183273-3183273 on Build GRCh38
rs563538833 Chr.6: 3183352-3183352 on Build GRCh38
rs527689279 Chr.6: 3183455-3183455 on Build GRCh38
rs555659102 Chr.6: 3183266-3183266 on Build GRCh38
rs73343586 Chr.6: 3183425-3183425 on Build GRCh38
rs1013015837 Chr.6: 3183294-3183294 on Build GRCh38
rs898571539 Chr.6: 3183275-3183275 on Build GRCh38
rs555477739 Chr.6: 3183200-3183200 on Build GRCh38
rs1018143973 Chr.6: 3183292-3183292 on Build GRCh38
rs895380421 Chr.6: 3183144-3183144 on Build GRCh38
rs903865079 Chr.6: 3183198-3183198 on Build GRCh38
rs145954686 Chr.6: 3183175-3183175 on Build GRCh38
rs117429828 Chr.6: 3183362-3183362 on Build GRCh38
rs1052468136 Chr.6: 3183524-3183524 on Build GRCh38
rs760508832 Chr.6: 3183316-3183316 on Build GRCh38
rs182497682 Chr.6: 3183360-3183360 on Build GRCh38
rs76581686 Chr.6: 3183296-3183296 on Build GRCh38

View Full Product Details