Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27324447-27325030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTG1P20
Entrez Gene ID
644961
Gene Name
actin gamma 1 pseudogene 20
Gene Aliases
-
UniGene
Hs.660887
Gene Chromosome Location
Chr.1: 27323874-27326525 on Build GRCh38
Transcripts
NR_033926.1 NM_032125.2 NR_037576.1 NR_037577.1 NR_037580.1

Gene Symbol
TMEM222
Entrez Gene ID
84065
Gene Name
transmembrane protein 222
Gene Aliases
C1orf160
UniGene
Hs.469171
Gene Chromosome Location
Chr.1: 27322145-27336400 on Build GRCh38
Transcripts
NR_033926.1 NM_032125.2 NR_037576.1 NR_037577.1 NR_037580.1

SNP Detail

SNP ID Chromosome Location
rs74416667 Chr.1: 27324540-27324540 on Build GRCh38
rs373655769 Chr.1: 27324596-27324596 on Build GRCh38
rs57922300 Chr.1: 27324891-27324891 on Build GRCh38
rs911604860 Chr.1: 27324753-27324753 on Build GRCh38
rs760600489 Chr.1: 27324626-27324626 on Build GRCh38
rs1007520403 Chr.1: 27324976-27324976 on Build GRCh38
rs752368263 Chr.1: 27324959-27324959 on Build GRCh38
rs776231453 Chr.1: 27324771-27324771 on Build GRCh38
rs991437710 Chr.1: 27324848-27324848 on Build GRCh38
rs906751592 Chr.1: 27324884-27324884 on Build GRCh38
rs994866373 Chr.1: 27324923-27324923 on Build GRCh38
rs908255651 Chr.1: 27324925-27324925 on Build GRCh38
rs796274590 Chr.1: 27324470-27324470 on Build GRCh38
rs772115284 Chr.1: 27324545-27324545 on Build GRCh38
rs965782107 Chr.1: 27324762-27324762 on Build GRCh38
rs536859200 Chr.1: 27324883-27324883 on Build GRCh38
rs916959488 Chr.1: 27324523-27324523 on Build GRCh38
rs1021015919 Chr.1: 27324709-27324709 on Build GRCh38
rs544746902 Chr.1: 27324568-27324568 on Build GRCh38
rs867251779 Chr.1: 27324818-27324818 on Build GRCh38
rs377659882 Chr.1: 27324792-27324792 on Build GRCh38
rs997056052 Chr.1: 27324625-27324625 on Build GRCh38
rs887255134 Chr.1: 27324555-27324555 on Build GRCh38
rs971074587 Chr.1: 27324870-27324870 on Build GRCh38
rs377249816 Chr.1: 27324948-27324948 on Build GRCh38
rs900057429 Chr.1: 27324607-27324607 on Build GRCh38
rs1029611904 Chr.1: 27324630-27324630 on Build GRCh38
rs539008113 Chr.1: 27324482-27324482 on Build GRCh38
rs991113978 Chr.1: 27324514-27324514 on Build GRCh38
rs528491774 Chr.1: 27324754-27324754 on Build GRCh38
rs374366252 Chr.1: 27324602-27324602 on Build GRCh38
rs1018780297 Chr.1: 27324989-27324989 on Build GRCh38
rs573488278 Chr.1: 27324915-27324915 on Build GRCh38
rs371541973 Chr.1: 27324872-27324872 on Build GRCh38
rs925864571 Chr.1: 27324476-27324476 on Build GRCh38
rs1038496475 Chr.1: 27324953-27324953 on Build GRCh38
rs759397790 Chr.1: 27324851-27324851 on Build GRCh38
rs1045101417 Chr.1: 27324517-27324517 on Build GRCh38
rs1046840281 Chr.1: 27324548-27324548 on Build GRCh38
rs770904543 Chr.1: 27324700-27324700 on Build GRCh38
rs370480091 Chr.1: 27324973-27324973 on Build GRCh38
rs1012432775 Chr.1: 27324480-27324480 on Build GRCh38
rs937250589 Chr.1: 27324494-27324494 on Build GRCh38
rs968551367 Chr.1: 27324769-27324769 on Build GRCh38
rs921470118 Chr.1: 27324986-27324986 on Build GRCh38
rs995149997 Chr.1: 27324620-27324620 on Build GRCh38
rs958609445 Chr.1: 27324841-27324841 on Build GRCh38
rs891157442 Chr.1: 27324676-27324676 on Build GRCh38
rs1038955620 Chr.1: 27324581-27324581 on Build GRCh38
rs146112325 Chr.1: 27324834-27324834 on Build GRCh38
rs962103973 Chr.1: 27324616-27324616 on Build GRCh38
rs370997967 Chr.1: 27324804-27324804 on Build GRCh38
rs893945357 Chr.1: 27324471-27324471 on Build GRCh38
rs935702134 Chr.1: 27324793-27324793 on Build GRCh38
rs985969474 Chr.1: 27324747-27324747 on Build GRCh38

View Full Product Details