Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9229222-9229776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
A2MP1
Entrez Gene ID
3
Gene Name
alpha-2-macroglobulin pseudogene 1
Gene Aliases
A2MP
UniGene
Hs.569125
Gene Chromosome Location
Chr.12: 9228533-9234207 on Build GRCh38
Transcripts
NR_040112.1

SNP Detail

SNP ID Chromosome Location
rs963119363 Chr.12: 9229655-9229655 on Build GRCh38
rs1001314932 Chr.12: 9229444-9229444 on Build GRCh38
rs999781667 Chr.12: 9229350-9229350 on Build GRCh38
rs192407919 Chr.12: 9229502-9229502 on Build GRCh38
rs559440578 Chr.12: 9229515-9229515 on Build GRCh38
rs117646963 Chr.12: 9229343-9229343 on Build GRCh38
rs1053079100 Chr.12: 9229357-9229357 on Build GRCh38
rs776464082 Chr.12: 9229339-9229339 on Build GRCh38
rs766497264 Chr.12: 9229578-9229578 on Build GRCh38
rs551385012 Chr.12: 9229630-9229630 on Build GRCh38
rs141530373 Chr.12: 9229629-9229629 on Build GRCh38
rs1011220282 Chr.12: 9229377-9229377 on Build GRCh38
rs765345810 Chr.12: 9229682-9229704 on Build GRCh38
rs150870555 Chr.12: 9229672-9229672 on Build GRCh38
rs779013850 Chr.12: 9229683-9229683 on Build GRCh38
rs908961592 Chr.12: 9229654-9229654 on Build GRCh38
rs751854791 Chr.12: 9229524-9229524 on Build GRCh38
rs905594564 Chr.12: 9229392-9229392 on Build GRCh38
rs1052631614 Chr.12: 9229706-9229706 on Build GRCh38
rs564897572 Chr.12: 9229671-9229671 on Build GRCh38
rs765658472 Chr.12: 9229369-9229369 on Build GRCh38
rs753154990 Chr.12: 9229420-9229420 on Build GRCh38
rs946977191 Chr.12: 9229716-9229716 on Build GRCh38
rs76128776 Chr.12: 9229301-9229301 on Build GRCh38
rs750856820 Chr.12: 9229383-9229383 on Build GRCh38
rs930112594 Chr.12: 9229705-9229705 on Build GRCh38
rs890646042 Chr.12: 9229327-9229327 on Build GRCh38
rs1021319789 Chr.12: 9229389-9229389 on Build GRCh38
rs980952561 Chr.12: 9229304-9229304 on Build GRCh38
rs775128255 Chr.12: 9229322-9229322 on Build GRCh38
rs57548405 Chr.12: 9229246-9229246 on Build GRCh38
rs1024829076 Chr.12: 9229612-9229612 on Build GRCh38
rs960184287 Chr.12: 9229467-9229467 on Build GRCh38
rs1040207676 Chr.12: 9229484-9229484 on Build GRCh38
rs747687431 Chr.12: 9229718-9229718 on Build GRCh38
rs140984867 Chr.12: 9229267-9229267 on Build GRCh38
rs921958861 Chr.12: 9229624-9229624 on Build GRCh38
rs774852073 Chr.12: 9229419-9229419 on Build GRCh38
rs918835227 Chr.12: 9229367-9229367 on Build GRCh38
rs574698925 Chr.12: 9229473-9229473 on Build GRCh38
rs891158511 Chr.12: 9229397-9229397 on Build GRCh38
rs1001110863 Chr.12: 9229523-9229523 on Build GRCh38
rs1035521085 Chr.12: 9229460-9229460 on Build GRCh38
rs1013484500 Chr.12: 9229603-9229603 on Build GRCh38
rs1051367064 Chr.12: 9229707-9229707 on Build GRCh38
rs930248473 Chr.12: 9229368-9229368 on Build GRCh38
rs1015220947 Chr.12: 9229475-9229475 on Build GRCh38
rs61918378 Chr.12: 9229276-9229276 on Build GRCh38
rs911985073 Chr.12: 9229635-9229635 on Build GRCh38
rs956531825 Chr.12: 9229600-9229600 on Build GRCh38
rs1001282284 Chr.12: 9229725-9229725 on Build GRCh38
rs763528905 Chr.12: 9229457-9229457 on Build GRCh38
rs117876726 Chr.12: 9229363-9229363 on Build GRCh38
rs992034902 Chr.12: 9229249-9229249 on Build GRCh38
rs973351250 Chr.12: 9229537-9229537 on Build GRCh38
rs969043772 Chr.12: 9229622-9229622 on Build GRCh38

View Full Product Details