Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 103860986-103861435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC41P
Entrez Gene ID
253724
Gene Name
tetratricopeptide repeat domain 41, pseudogene
Gene Aliases
GNN, GNNP
UniGene
Hs.506549
Gene Chromosome Location
Chr.12: 103843749-103930211 on Build GRCh38
Transcripts
NR_027249.1

SNP Detail

SNP ID Chromosome Location
rs764370556 Chr.12: 103861230-103861230 on Build GRCh38
rs961562523 Chr.12: 103861060-103861060 on Build GRCh38
rs753477811 Chr.12: 103861089-103861089 on Build GRCh38
rs751822853 Chr.12: 103861267-103861267 on Build GRCh38
rs960914809 Chr.12: 103861364-103861364 on Build GRCh38
rs749450550 Chr.12: 103861264-103861264 on Build GRCh38
rs1053718960 Chr.12: 103861051-103861051 on Build GRCh38
rs201156506 Chr.12: 103861312-103861312 on Build GRCh38
rs557249896 Chr.12: 103861266-103861266 on Build GRCh38
rs566644653 Chr.12: 103861201-103861201 on Build GRCh38
rs74651613 Chr.12: 103861197-103861197 on Build GRCh38
rs970858528 Chr.12: 103861360-103861360 on Build GRCh38
rs569321122 Chr.12: 103861071-103861071 on Build GRCh38
rs371881791 Chr.12: 103861058-103861058 on Build GRCh38
rs1053384588 Chr.12: 103861196-103861196 on Build GRCh38
rs757465616 Chr.12: 103861351-103861351 on Build GRCh38
rs916312126 Chr.12: 103861330-103861330 on Build GRCh38
rs376976711 Chr.12: 103861410-103861410 on Build GRCh38
rs984227186 Chr.12: 103861010-103861010 on Build GRCh38
rs528370590 Chr.12: 103861287-103861288 on Build GRCh38
rs375810638 Chr.12: 103861063-103861063 on Build GRCh38
rs992465896 Chr.12: 103861094-103861094 on Build GRCh38
rs991072951 Chr.12: 103861368-103861368 on Build GRCh38
rs553220360 Chr.12: 103861198-103861198 on Build GRCh38
rs865875499 Chr.12: 103861260-103861260 on Build GRCh38
rs924212012 Chr.12: 103861025-103861025 on Build GRCh38
rs935626306 Chr.12: 103861407-103861407 on Build GRCh38
rs1017477835 Chr.12: 103861020-103861020 on Build GRCh38
rs558403711 Chr.12: 103861327-103861327 on Build GRCh38
rs972903121 Chr.12: 103861323-103861323 on Build GRCh38
rs769824267 Chr.12: 103861229-103861229 on Build GRCh38
rs935552873 Chr.12: 103861034-103861034 on Build GRCh38
rs1042808732 Chr.12: 103861126-103861126 on Build GRCh38
rs961784970 Chr.12: 103861286-103861286 on Build GRCh38
rs774761787 Chr.12: 103861294-103861294 on Build GRCh38
rs748280233 Chr.12: 103861205-103861205 on Build GRCh38
rs552359094 Chr.12: 103861015-103861015 on Build GRCh38
rs11111817 Chr.12: 103861154-103861154 on Build GRCh38
rs948404814 Chr.12: 103861390-103861390 on Build GRCh38
rs1007421936 Chr.12: 103861061-103861061 on Build GRCh38
rs948411796 Chr.12: 103861121-103861121 on Build GRCh38
rs928185339 Chr.12: 103861078-103861078 on Build GRCh38
rs765614159 Chr.12: 103861162-103861162 on Build GRCh38
rs889784053 Chr.12: 103861252-103861252 on Build GRCh38
rs553096917 Chr.12: 103861044-103861044 on Build GRCh38
rs916873531 Chr.12: 103861375-103861375 on Build GRCh38
rs190170145 Chr.12: 103861272-103861272 on Build GRCh38
rs117881778 Chr.12: 103861093-103861093 on Build GRCh38
rs904305071 Chr.12: 103861153-103861153 on Build GRCh38
rs1047108376 Chr.12: 103861019-103861019 on Build GRCh38
rs548601419 Chr.12: 103861098-103861098 on Build GRCh38

View Full Product Details