Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 103862045-103862602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC41P
Entrez Gene ID
253724
Gene Name
tetratricopeptide repeat domain 41, pseudogene
Gene Aliases
GNN, GNNP
UniGene
Hs.506549
Gene Chromosome Location
Chr.12: 103843749-103930211 on Build GRCh38
Transcripts
NR_027249.1

SNP Detail

SNP ID Chromosome Location
rs79601998 Chr.12: 103862124-103862126 on Build GRCh38
rs918545260 Chr.12: 103862258-103862258 on Build GRCh38
rs906224359 Chr.12: 103862287-103862287 on Build GRCh38
rs78765550 Chr.12: 103862073-103862073 on Build GRCh38
rs552344524 Chr.12: 103862126-103862126 on Build GRCh38
rs1031660624 Chr.12: 103862435-103862435 on Build GRCh38
rs745955122 Chr.12: 103862117-103862117 on Build GRCh38
rs370447507 Chr.12: 103862330-103862330 on Build GRCh38
rs757037496 Chr.12: 103862276-103862276 on Build GRCh38
rs546087012 Chr.12: 103862510-103862510 on Build GRCh38
rs760241307 Chr.12: 103862204-103862204 on Build GRCh38
rs1010574271 Chr.12: 103862348-103862348 on Build GRCh38
rs985232918 Chr.12: 103862133-103862133 on Build GRCh38
rs988467243 Chr.12: 103862109-103862109 on Build GRCh38
rs151311174 Chr.12: 103862144-103862144 on Build GRCh38
rs563060135 Chr.12: 103862532-103862532 on Build GRCh38
rs780070528 Chr.12: 103862337-103862337 on Build GRCh38
rs982792288 Chr.12: 103862490-103862490 on Build GRCh38
rs908507317 Chr.12: 103862497-103862497 on Build GRCh38
rs576809641 Chr.12: 103862552-103862552 on Build GRCh38
rs898992435 Chr.12: 103862216-103862217 on Build GRCh38
rs1001077612 Chr.12: 103862402-103862402 on Build GRCh38
rs751875497 Chr.12: 103862199-103862199 on Build GRCh38
rs1003180688 Chr.12: 103862305-103862305 on Build GRCh38
rs1032637805 Chr.12: 103862085-103862085 on Build GRCh38
rs896547423 Chr.12: 103862294-103862294 on Build GRCh38
rs577466084 Chr.12: 103862365-103862365 on Build GRCh38
rs764534407 Chr.12: 103862086-103862086 on Build GRCh38
rs1029299881 Chr.12: 103862529-103862529 on Build GRCh38
rs773119160 Chr.12: 103862267-103862268 on Build GRCh38
rs929947613 Chr.12: 103862277-103862277 on Build GRCh38
rs753385944 Chr.12: 103862236-103862236 on Build GRCh38
rs533520602 Chr.12: 103862304-103862304 on Build GRCh38
rs554087929 Chr.12: 103862324-103862324 on Build GRCh38
rs758077959 Chr.12: 103862335-103862335 on Build GRCh38
rs763644444 Chr.12: 103862232-103862232 on Build GRCh38
rs964551533 Chr.12: 103862398-103862398 on Build GRCh38
rs968298471 Chr.12: 103862071-103862071 on Build GRCh38
rs904019422 Chr.12: 103862399-103862399 on Build GRCh38
rs538496864 Chr.12: 103862074-103862074 on Build GRCh38
rs1022499393 Chr.12: 103862366-103862366 on Build GRCh38
rs911054665 Chr.12: 103862136-103862136 on Build GRCh38
rs777003346 Chr.12: 103862427-103862429 on Build GRCh38
rs767787145 Chr.12: 103862493-103862493 on Build GRCh38
rs976066454 Chr.12: 103862106-103862106 on Build GRCh38
rs757572177 Chr.12: 103862331-103862331 on Build GRCh38
rs574063643 Chr.12: 103862184-103862184 on Build GRCh38
rs985180349 Chr.12: 103862545-103862545 on Build GRCh38
rs377365265 Chr.12: 103862311-103862311 on Build GRCh38
rs955582099 Chr.12: 103862441-103862441 on Build GRCh38
rs1030308731 Chr.12: 103862409-103862409 on Build GRCh38
rs756560342 Chr.12: 103862225-103862225 on Build GRCh38
rs1056501529 Chr.12: 103862289-103862289 on Build GRCh38
rs922340397 Chr.12: 103862125-103862125 on Build GRCh38
rs955894241 Chr.12: 103862090-103862090 on Build GRCh38
rs182339939 Chr.12: 103862157-103862157 on Build GRCh38
rs750283421 Chr.12: 103862327-103862327 on Build GRCh38
rs1012348394 Chr.12: 103862338-103862338 on Build GRCh38
rs1045200079 Chr.12: 103862387-103862387 on Build GRCh38
rs537213235 Chr.12: 103862183-103862183 on Build GRCh38
rs772208784 Chr.12: 103862116-103862117 on Build GRCh38
rs892744808 Chr.12: 103862469-103862469 on Build GRCh38

View Full Product Details