Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 55774239-55774788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CES1P1
Entrez Gene ID
51716
Gene Name
carboxylesterase 1 pseudogene 1
Gene Aliases
CES1A2, CES1A3, CES4, CESR, PCE-3
UniGene
Hs.721645
Gene Chromosome Location
Chr.16: 55760599-55774926 on Build GRCh38
Transcripts
NR_003276.2

SNP Detail

SNP ID Chromosome Location
rs201358847 Chr.16: 55774552-55774553 on Build GRCh38
rs760180352 Chr.16: 55774365-55774365 on Build GRCh38
rs545495628 Chr.16: 55774453-55774453 on Build GRCh38
rs28628572 Chr.16: 55774452-55774452 on Build GRCh38
rs544390165 Chr.16: 55774273-55774273 on Build GRCh38
rs574593189 Chr.16: 55774337-55774337 on Build GRCh38
rs753901108 Chr.16: 55774680-55774680 on Build GRCh38
rs532852596 Chr.16: 55774739-55774739 on Build GRCh38
rs555619076 Chr.16: 55774392-55774392 on Build GRCh38
rs767971938 Chr.16: 55774376-55774376 on Build GRCh38
rs184053530 Chr.16: 55774504-55774504 on Build GRCh38
rs556570208 Chr.16: 55774446-55774446 on Build GRCh38
rs557637154 Chr.16: 55774497-55774497 on Build GRCh38
rs139323073 Chr.16: 55774553-55774555 on Build GRCh38
rs866974833 Chr.16: 55774484-55774484 on Build GRCh38
rs375380715 Chr.16: 55774554-55774556 on Build GRCh38
rs28393348 Chr.16: 55774627-55774627 on Build GRCh38
rs562513261 Chr.16: 55774722-55774722 on Build GRCh38
rs28672783 Chr.16: 55774401-55774401 on Build GRCh38
rs557263692 Chr.16: 55774523-55774523 on Build GRCh38
rs759209617 Chr.16: 55774394-55774394 on Build GRCh38
rs563385533 Chr.16: 55774475-55774475 on Build GRCh38
rs757291229 Chr.16: 55774726-55774726 on Build GRCh38
rs3743790 Chr.16: 55774281-55774281 on Build GRCh38
rs144429530 Chr.16: 55774512-55774512 on Build GRCh38
rs28522212 Chr.16: 55774669-55774669 on Build GRCh38
rs548163880 Chr.16: 55774409-55774409 on Build GRCh38
rs561952197 Chr.16: 55774577-55774577 on Build GRCh38
rs764437931 Chr.16: 55774506-55774506 on Build GRCh38
rs28499399 Chr.16: 55774764-55774764 on Build GRCh38

View Full Product Details