Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 177672118-177672631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC202181
Entrez Gene ID
202181
Gene Name
SUMO interacting motifs containing 1 pseudogene
Gene Aliases
-
UniGene
Hs.189914
Gene Chromosome Location
Chr.5: 177618500-177672277 on Build GRCh38
Transcripts
NR_026921.1

SNP Detail

SNP ID Chromosome Location
rs767827223 Chr.5: 177672191-177672191 on Build GRCh38
rs551011239 Chr.5: 177672238-177672238 on Build GRCh38
rs759938557 Chr.5: 177672173-177672173 on Build GRCh38
rs972979608 Chr.5: 177672232-177672233 on Build GRCh38
rs746670841 Chr.5: 177672259-177672259 on Build GRCh38
rs369791757 Chr.5: 177672364-177672364 on Build GRCh38
rs935667788 Chr.5: 177672270-177672278 on Build GRCh38
rs746881741 Chr.5: 177672182-177672184 on Build GRCh38
rs865950786 Chr.5: 177672341-177672341 on Build GRCh38
rs113108733 Chr.5: 177672402-177672402 on Build GRCh38
rs868652662 Chr.5: 177672273-177672273 on Build GRCh38
rs924306677 Chr.5: 177672262-177672262 on Build GRCh38
rs531478993 Chr.5: 177672207-177672207 on Build GRCh38
rs866848831 Chr.5: 177672269-177672269 on Build GRCh38
rs866524789 Chr.5: 177672352-177672352 on Build GRCh38
rs1053991227 Chr.5: 177672517-177672517 on Build GRCh38
rs368969585 Chr.5: 177672609-177672609 on Build GRCh38
rs1041465022 Chr.5: 177672261-177672261 on Build GRCh38
rs769410651 Chr.5: 177672139-177672139 on Build GRCh38
rs200105127 Chr.5: 177672402-177672402 on Build GRCh38
rs867915892 Chr.5: 177672448-177672448 on Build GRCh38
rs536628978 Chr.5: 177672377-177672377 on Build GRCh38
rs772777052 Chr.5: 177672163-177672163 on Build GRCh38

View Full Product Details