Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 57063749-57063873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCF12
Entrez Gene ID
6938
Gene Name
transcription factor 12
Gene Aliases
CRS3, HEB, HTF4, HsT17266, TCF-12, bHLHb20
UniGene
Hs.511504
Gene Chromosome Location
Chr.15: 56918090-57291261 on Build GRCh38
Transcripts
NM_001322151.1 NM_001322152.1 NM_001322154.1 NM_001322156.1 NM_001322157.1 NM_001322158.1 NM_001322159.1 NM_001322161.1 NM_001322162.1 NM_001322164.1 NM_001322165.1 NM_003205.3 NM_207036.1 NM_207037.1 NM_207038.1 XM_011521959.2 XM_011521960.2 XM_011521961.2 XM_011521962.2 XM_011521963.2 XM_011521965.1 XM_011521967.1 XM_017022520.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3502624 c.203C>T p.P68L Chr.15: 57063804-57063804 on Build GRCh38
1493484 c.151A>T p.I51F Chr.15: 57063752-57063752 on Build GRCh38
5462468 c.149-1G>A p.? Chr.15: 57063749-57063749 on Build GRCh38
3502622 c.202C>T p.P68S Chr.15: 57063803-57063803 on Build GRCh38
5616823 c.222+9G>A p.? Chr.15: 57063832-57063832 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773012004 Chr.15: 57063810-57063810 on Build GRCh38
rs775676003 Chr.15: 57063837-57063837 on Build GRCh38
rs746120476 Chr.15: 57063819-57063819 on Build GRCh38
rs769528163 Chr.15: 57063807-57063807 on Build GRCh38
rs983063352 Chr.15: 57063812-57063812 on Build GRCh38
rs760809068 Chr.15: 57063839-57063839 on Build GRCh38
rs781060597 Chr.15: 57063792-57063792 on Build GRCh38
rs771248917 Chr.15: 57063786-57063786 on Build GRCh38
rs777050221 Chr.15: 57063842-57063842 on Build GRCh38
rs764443007 Chr.15: 57063846-57063847 on Build GRCh38
rs771423040 Chr.15: 57063845-57063845 on Build GRCh38
rs748088394 Chr.15: 57063797-57063797 on Build GRCh38
rs775722371 Chr.15: 57063841-57063842 on Build GRCh38
rs760965345 Chr.15: 57063845-57063846 on Build GRCh38
rs765547911 Chr.15: 57063846-57063846 on Build GRCh38
rs149566552 Chr.15: 57063791-57063791 on Build GRCh38
rs772223615 Chr.15: 57063831-57063831 on Build GRCh38
rs762329851 Chr.15: 57063843-57063843 on Build GRCh38

View Full Product Details