Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152325954-152326113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW7
Entrez Gene ID
55294
Gene Name
F-box and WD repeat domain containing 7
Gene Aliases
AGO, CDC4, FBW6, FBW7, FBX30, FBXO30, FBXW6, SEL-10, SEL10, hAgo, hCdc4
UniGene
Hs.561245
Gene Chromosome Location
Chr.4: 152321258-152536095 on Build GRCh38
Transcripts
NM_001013415.1 NM_018315.4 NM_033632.3 XM_011532083.1 XM_011532084.1 XM_011532085.1 XM_011532086.1 XM_011532087.1 XM_011532088.1 XM_017008362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6662544 c.1279T>C p.Y427H Chr.4: 152326017-152326017 on Build GRCh38
5490558 c.1598G>T p.C533F Chr.4: 152326052-152326052 on Build GRCh38
3721801 c.1391T>A p.V464D Chr.4: 152326019-152326019 on Build GRCh38
6921230 c.878C>T p.T293I Chr.4: 152326055-152326055 on Build GRCh38
6937181 c.1012G>T p.V338F Chr.4: 152326110-152326110 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs869059999 Chr.4: 152326079-152326079 on Build GRCh38
rs753448369 Chr.4: 152326069-152326069 on Build GRCh38
rs764317243 Chr.4: 152325991-152325991 on Build GRCh38
rs759412025 Chr.4: 152325980-152325980 on Build GRCh38
rs763590985 Chr.4: 152326024-152326024 on Build GRCh38
rs370070523 Chr.4: 152326000-152326000 on Build GRCh38
rs150453955 Chr.4: 152325993-152325993 on Build GRCh38
rs1039390062 Chr.4: 152326042-152326042 on Build GRCh38
rs141482952 Chr.4: 152325986-152325986 on Build GRCh38

View Full Product Details