Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 50245223-50245496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASAH2
Entrez Gene ID
56624
Gene Name
N-acylsphingosine amidohydrolase 2
Gene Aliases
BCDase, HNAC1, LCDase, N-CDase, NCDase
UniGene
Hs.512645
Gene Chromosome Location
Chr.10: 50184861-50251564 on Build GRCh38
Transcripts
NM_001143974.1 NM_019893.2 XM_011539970.2 XM_011539971.2 XM_011539972.2 XM_011539973.2 XM_017016435.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
378079 c.205C>A p.P69T Chr.10: 50245320-50245320 on Build GRCh38
4925987 c.291G>T p.Q97H Chr.10: 50245291-50245291 on Build GRCh38
1561303 c.173C>A p.T58N Chr.10: 50245409-50245409 on Build GRCh38
6243661 c.98C>A p.T33N Chr.10: 50245427-50245427 on Build GRCh38
6605415 c.227C>T p.T76I Chr.10: 50245355-50245355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748317315 Chr.10: 50245371-50245371 on Build GRCh38
rs891831347 Chr.10: 50245376-50245376 on Build GRCh38
rs769687313 Chr.10: 50245332-50245332 on Build GRCh38
rs763608304 Chr.10: 50245432-50245432 on Build GRCh38
rs774832323 Chr.10: 50245427-50245427 on Build GRCh38
rs199725527 Chr.10: 50245344-50245344 on Build GRCh38
rs114893748 Chr.10: 50245280-50245280 on Build GRCh38
rs867264637 Chr.10: 50245394-50245394 on Build GRCh38
rs773491153 Chr.10: 50245433-50245434 on Build GRCh38
rs149533805 Chr.10: 50245329-50245329 on Build GRCh38
rs766393311 Chr.10: 50245338-50245338 on Build GRCh38
rs765745051 Chr.10: 50245445-50245445 on Build GRCh38
rs116049719 Chr.10: 50245335-50245335 on Build GRCh38
rs867801776 Chr.10: 50245349-50245349 on Build GRCh38
rs751133831 Chr.10: 50245342-50245342 on Build GRCh38
rs766048989 Chr.10: 50245276-50245276 on Build GRCh38
rs867954994 Chr.10: 50245449-50245449 on Build GRCh38
rs750983912 Chr.10: 50245446-50245446 on Build GRCh38
rs778244882 Chr.10: 50245353-50245353 on Build GRCh38
rs192860010 Chr.10: 50245438-50245438 on Build GRCh38
rs781230478 Chr.10: 50245469-50245469 on Build GRCh38
rs7097319 Chr.10: 50245393-50245393 on Build GRCh38
rs539221623 Chr.10: 50245323-50245323 on Build GRCh38
rs769192204 Chr.10: 50245350-50245351 on Build GRCh38
rs761525494 Chr.10: 50245330-50245330 on Build GRCh38
rs538113005 Chr.10: 50245387-50245387 on Build GRCh38
rs142364715 Chr.10: 50245403-50245403 on Build GRCh38
rs747036724 Chr.10: 50245465-50245465 on Build GRCh38
rs759795155 Chr.10: 50245250-50245250 on Build GRCh38
rs767705180 Chr.10: 50245255-50245255 on Build GRCh38
rs7067625 Chr.10: 50245431-50245431 on Build GRCh38
rs772181094 Chr.10: 50245434-50245434 on Build GRCh38
rs142760278 Chr.10: 50245277-50245277 on Build GRCh38
rs780941256 Chr.10: 50245288-50245288 on Build GRCh38
rs758862345 Chr.10: 50245413-50245413 on Build GRCh38
rs773108125 Chr.10: 50245389-50245389 on Build GRCh38
rs141713369 Chr.10: 50245386-50245386 on Build GRCh38
rs749876809 Chr.10: 50245308-50245308 on Build GRCh38
rs753010670 Chr.10: 50245290-50245290 on Build GRCh38
rs750316215 Chr.10: 50245269-50245269 on Build GRCh38
rs757786642 Chr.10: 50245361-50245361 on Build GRCh38
rs755249924 Chr.10: 50245467-50245467 on Build GRCh38
rs569459344 Chr.10: 50245385-50245385 on Build GRCh38
rs80335014 Chr.10: 50245347-50245347 on Build GRCh38
rs754133961 Chr.10: 50245354-50245354 on Build GRCh38
rs139973487 Chr.10: 50245345-50245345 on Build GRCh38
rs370340088 Chr.10: 50245379-50245379 on Build GRCh38
rs762786150 Chr.10: 50245271-50245271 on Build GRCh38
rs776434478 Chr.10: 50245256-50245256 on Build GRCh38
rs188096471 Chr.10: 50245429-50245429 on Build GRCh38
rs771397769 Chr.10: 50245311-50245311 on Build GRCh38
rs779356095 Chr.10: 50245364-50245364 on Build GRCh38
rs779014422 Chr.10: 50245312-50245312 on Build GRCh38
rs374685598 Chr.10: 50245408-50245408 on Build GRCh38
rs780218782 Chr.10: 50245464-50245464 on Build GRCh38
rs772877573 Chr.10: 50245333-50245333 on Build GRCh38
rs372733179 Chr.10: 50245461-50245461 on Build GRCh38
rs1026161935 Chr.10: 50245430-50245430 on Build GRCh38
rs201282742 Chr.10: 50245297-50245297 on Build GRCh38
rs756446942 Chr.10: 50245292-50245292 on Build GRCh38
rs778943097 Chr.10: 50245351-50245368 on Build GRCh38
rs146246912 Chr.10: 50245392-50245392 on Build GRCh38
rs137937721 Chr.10: 50245328-50245328 on Build GRCh38
rs754264015 Chr.10: 50245434-50245434 on Build GRCh38
rs761627134 Chr.10: 50245259-50245259 on Build GRCh38
rs377466392 Chr.10: 50245377-50245377 on Build GRCh38
rs767204609 Chr.10: 50245415-50245415 on Build GRCh38
rs756730469 Chr.10: 50245260-50245260 on Build GRCh38

View Full Product Details