Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2339929-2340175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPPL2B
Entrez Gene ID
56928
Gene Name
signal peptide peptidase like 2B
Gene Aliases
IMP-4, IMP4, PSH4, PSL1
UniGene
Hs.744026
Gene Chromosome Location
Chr.19: 2328571-2355102 on Build GRCh38
Transcripts
NM_001077238.1 NM_152988.2 XM_011528138.2 XM_011528139.1 XM_011528140.1 XM_017027016.1 XM_017027017.1 XM_017027018.1 XM_017027019.1

SNP Detail

SNP ID Chromosome Location
rs375594733 Chr.19: 2340081-2340081 on Build GRCh38
rs770483932 Chr.19: 2340037-2340037 on Build GRCh38
rs529477585 Chr.19: 2340148-2340148 on Build GRCh38
rs775226416 Chr.19: 2340043-2340043 on Build GRCh38
rs752078524 Chr.19: 2339996-2339996 on Build GRCh38
rs750339570 Chr.19: 2340116-2340116 on Build GRCh38
rs766195711 Chr.19: 2340124-2340124 on Build GRCh38
rs990918910 Chr.19: 2340021-2340021 on Build GRCh38
rs745718368 Chr.19: 2340011-2340011 on Build GRCh38
rs374736782 Chr.19: 2340038-2340038 on Build GRCh38
rs768315832 Chr.19: 2340151-2340151 on Build GRCh38
rs201510032 Chr.19: 2340135-2340135 on Build GRCh38
rs777769674 Chr.19: 2339971-2339971 on Build GRCh38
rs371443207 Chr.19: 2340015-2340015 on Build GRCh38
rs983541821 Chr.19: 2340153-2340153 on Build GRCh38
rs200233967 Chr.19: 2339960-2339960 on Build GRCh38
rs753411001 Chr.19: 2340048-2340048 on Build GRCh38
rs755226260 Chr.19: 2340061-2340061 on Build GRCh38
rs751372403 Chr.19: 2340132-2340132 on Build GRCh38
rs376223700 Chr.19: 2339993-2339993 on Build GRCh38
rs1609752 Chr.19: 2339959-2339959 on Build GRCh38
rs775584103 Chr.19: 2339957-2339957 on Build GRCh38
rs752781433 Chr.19: 2340136-2340136 on Build GRCh38
rs117324377 Chr.19: 2339976-2339976 on Build GRCh38
rs753160287 Chr.19: 2340040-2340041 on Build GRCh38
rs748255113 Chr.19: 2340080-2340080 on Build GRCh38
rs971251929 Chr.19: 2340152-2340152 on Build GRCh38
rs3746285 Chr.19: 2339950-2339950 on Build GRCh38
rs755174070 Chr.19: 2340134-2340134 on Build GRCh38
rs978932228 Chr.19: 2340020-2340020 on Build GRCh38
rs368980475 Chr.19: 2339956-2339956 on Build GRCh38
rs748664179 Chr.19: 2340035-2340035 on Build GRCh38
rs771073240 Chr.19: 2339949-2339949 on Build GRCh38
rs1029323330 Chr.19: 2340006-2340006 on Build GRCh38
rs755946181 Chr.19: 2339970-2339970 on Build GRCh38
rs755677607 Chr.19: 2339997-2339997 on Build GRCh38
rs919611901 Chr.19: 2340067-2340067 on Build GRCh38
rs758551369 Chr.19: 2340012-2340012 on Build GRCh38
rs768759218 Chr.19: 2340028-2340028 on Build GRCh38
rs781322141 Chr.19: 2340062-2340062 on Build GRCh38
rs567396750 Chr.19: 2339992-2339992 on Build GRCh38
rs763884734 Chr.19: 2340005-2340005 on Build GRCh38
rs775376600 Chr.19: 2339979-2339979 on Build GRCh38
rs767034022 Chr.19: 2340041-2340041 on Build GRCh38
rs768250522 Chr.19: 2340105-2340105 on Build GRCh38
rs754718982 Chr.19: 2339966-2339966 on Build GRCh38
rs762464020 Chr.19: 2339982-2339982 on Build GRCh38
rs748992627 Chr.19: 2339972-2339972 on Build GRCh38
rs761346557 Chr.19: 2340111-2340111 on Build GRCh38
rs377597000 Chr.19: 2340144-2340144 on Build GRCh38
rs747228091 Chr.19: 2340059-2340059 on Build GRCh38
rs758213494 Chr.19: 2340052-2340052 on Build GRCh38
rs997040315 Chr.19: 2339994-2339994 on Build GRCh38
rs749121237 Chr.19: 2339948-2339948 on Build GRCh38
rs769004951 Chr.19: 2339980-2339980 on Build GRCh38
rs533154359 Chr.19: 2340109-2340109 on Build GRCh38
rs3746287 Chr.19: 2340047-2340047 on Build GRCh38
rs776618411 Chr.19: 2340034-2340034 on Build GRCh38
rs763412108 Chr.19: 2340039-2340039 on Build GRCh38
rs778044874 Chr.19: 2340140-2340140 on Build GRCh38
rs765670025 Chr.19: 2339989-2339989 on Build GRCh38
rs551336177 Chr.19: 2339969-2339969 on Build GRCh38
rs780847246 Chr.19: 2339967-2339967 on Build GRCh38
rs367978961 Chr.19: 2340010-2340010 on Build GRCh38
rs760370255 Chr.19: 2340104-2340104 on Build GRCh38
rs371117284 Chr.19: 2339973-2339973 on Build GRCh38
rs779762660 Chr.19: 2340054-2340054 on Build GRCh38
rs371763993 Chr.19: 2339991-2339991 on Build GRCh38
rs369330627 Chr.19: 2339978-2339978 on Build GRCh38
rs189475556 Chr.19: 2340102-2340102 on Build GRCh38
rs1005304327 Chr.19: 2340085-2340085 on Build GRCh38
rs373088449 Chr.19: 2340110-2340110 on Build GRCh38
rs762272680 Chr.19: 2339961-2339961 on Build GRCh38
rs370030049 Chr.19: 2340089-2340089 on Build GRCh38
rs771406802 Chr.19: 2340147-2340147 on Build GRCh38
rs758707222 Chr.19: 2340047-2340047 on Build GRCh38
rs561026634 Chr.19: 2340117-2340117 on Build GRCh38
rs771300915 Chr.19: 2340093-2340093 on Build GRCh38
rs766085470 Chr.19: 2339963-2339963 on Build GRCh38
rs1032235271 Chr.19: 2340137-2340137 on Build GRCh38
rs150642945 Chr.19: 2340016-2340016 on Build GRCh38
rs753791172 Chr.19: 2340008-2340008 on Build GRCh38
rs765511243 Chr.19: 2339975-2339977 on Build GRCh38
rs759559911 Chr.19: 2339953-2339953 on Build GRCh38
rs533181897 Chr.19: 2339965-2339965 on Build GRCh38
rs577983110 Chr.19: 2340017-2340017 on Build GRCh38
rs1609753 Chr.19: 2340051-2340051 on Build GRCh38
rs746229400 Chr.19: 2340150-2340150 on Build GRCh38
rs757171309 Chr.19: 2340050-2340050 on Build GRCh38

View Full Product Details