Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19131804-19132065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGCR14
Entrez Gene ID
8220
Gene Name
DiGeorge syndrome critical region gene 14
Gene Aliases
DGCR13, DGS-H, DGS-I, DGSH, DGSI, ES2, Es2el
UniGene
Hs.517407
Gene Chromosome Location
Chr.22: 19130279-19144726 on Build GRCh38
Transcripts
NM_022719.2 NR_134304.1 NM_053006.4

Gene Symbol
TSSK2
Entrez Gene ID
23617
Gene Name
testis specific serine kinase 2
Gene Aliases
DGS-G, SPOGA2, STK22B, TSK2
UniGene
Hs.694070
Gene Chromosome Location
Chr.22: 19130808-19132623 on Build GRCh38
Transcripts
NM_022719.2 NR_134304.1 NM_053006.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1032001 c.503G>A p.R168H Chr.22: 19131902-19131902 on Build GRCh38
3552106 c.612G>A p.V204V Chr.22: 19132011-19132011 on Build GRCh38
5906164 c.418G>A p.E140K Chr.22: 19131817-19131817 on Build GRCh38
725622 c.579G>T p.Q193H Chr.22: 19131978-19131978 on Build GRCh38
4135145 c.420G>C p.E140D Chr.22: 19131819-19131819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145863577 Chr.22: 19131883-19131883 on Build GRCh38
rs759371117 Chr.22: 19131921-19131921 on Build GRCh38
rs746661706 Chr.22: 19131853-19131856 on Build GRCh38
rs772250594 Chr.22: 19131994-19131994 on Build GRCh38
rs751178916 Chr.22: 19132021-19132021 on Build GRCh38
rs773470708 Chr.22: 19132001-19132001 on Build GRCh38
rs370346388 Chr.22: 19131935-19131935 on Build GRCh38
rs904529577 Chr.22: 19131853-19131853 on Build GRCh38
rs972305651 Chr.22: 19131952-19131952 on Build GRCh38
rs150218881 Chr.22: 19132036-19132036 on Build GRCh38
rs775602028 Chr.22: 19132016-19132016 on Build GRCh38
rs371638010 Chr.22: 19131886-19131886 on Build GRCh38
rs267606176 Chr.22: 19131950-19131950 on Build GRCh38
rs148566995 Chr.22: 19131927-19131927 on Build GRCh38
rs141030556 Chr.22: 19131823-19131823 on Build GRCh38
rs752323315 Chr.22: 19131867-19131867 on Build GRCh38
rs200993519 Chr.22: 19131901-19131901 on Build GRCh38
rs752627852 Chr.22: 19131928-19131928 on Build GRCh38
rs377349406 Chr.22: 19131902-19131902 on Build GRCh38
rs759077282 Chr.22: 19132025-19132025 on Build GRCh38
rs780542563 Chr.22: 19131956-19131956 on Build GRCh38
rs776331935 Chr.22: 19131911-19131911 on Build GRCh38
rs761972241 Chr.22: 19131907-19131907 on Build GRCh38
rs764088773 Chr.22: 19132020-19132020 on Build GRCh38
rs757991278 Chr.22: 19131933-19131933 on Build GRCh38
rs144966098 Chr.22: 19131877-19131877 on Build GRCh38
rs776250553 Chr.22: 19132012-19132020 on Build GRCh38
rs935955986 Chr.22: 19131891-19131891 on Build GRCh38
rs897654067 Chr.22: 19131932-19131932 on Build GRCh38
rs753837958 Chr.22: 19131831-19131831 on Build GRCh38
rs749753119 Chr.22: 19131958-19131958 on Build GRCh38
rs45604134 Chr.22: 19132032-19132032 on Build GRCh38
rs750807706 Chr.22: 19131954-19131954 on Build GRCh38
rs768744348 Chr.22: 19131974-19131974 on Build GRCh38
rs770540053 Chr.22: 19131939-19131945 on Build GRCh38
rs776048479 Chr.22: 19131897-19131897 on Build GRCh38
rs759267150 Chr.22: 19132008-19132008 on Build GRCh38
rs751088788 Chr.22: 19131940-19131940 on Build GRCh38
rs185015956 Chr.22: 19131951-19131951 on Build GRCh38
rs56279111 Chr.22: 19131989-19131989 on Build GRCh38
rs946317126 Chr.22: 19131849-19131849 on Build GRCh38
rs142906174 Chr.22: 19131977-19131977 on Build GRCh38
rs770266307 Chr.22: 19131889-19131889 on Build GRCh38
rs762938347 Chr.22: 19132018-19132018 on Build GRCh38
rs746209813 Chr.22: 19131887-19131887 on Build GRCh38
rs372941768 Chr.22: 19131852-19131852 on Build GRCh38
rs376965034 Chr.22: 19131878-19131878 on Build GRCh38
rs373815280 Chr.22: 19131864-19131864 on Build GRCh38
rs151069324 Chr.22: 19132009-19132009 on Build GRCh38
rs376174754 Chr.22: 19131855-19131855 on Build GRCh38
rs756442861 Chr.22: 19131955-19131955 on Build GRCh38
rs756466071 Chr.22: 19132043-19132043 on Build GRCh38
rs975850629 Chr.22: 19132025-19132025 on Build GRCh38

View Full Product Details