Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 4577433-4577704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf40
Entrez Gene ID
143501
Gene Name
chromosome 11 open reading frame 40
Gene Aliases
NOV1
UniGene
Hs.350556
Gene Chromosome Location
Chr.11: 4571423-4577820 on Build GRCh38
Transcripts
NM_144663.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7334420 c.136C>G p.P46A Chr.11: 4577685-4577685 on Build GRCh38
3447491 c.129C>T p.T43T Chr.11: 4577692-4577692 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs578103621 Chr.11: 4577522-4577522 on Build GRCh38
rs371188536 Chr.11: 4577633-4577633 on Build GRCh38
rs201377859 Chr.11: 4577478-4577478 on Build GRCh38
rs781328816 Chr.11: 4577637-4577637 on Build GRCh38
rs761661041 Chr.11: 4577654-4577654 on Build GRCh38
rs754415404 Chr.11: 4577494-4577494 on Build GRCh38
rs767282394 Chr.11: 4577655-4577655 on Build GRCh38
rs929136957 Chr.11: 4577506-4577506 on Build GRCh38
rs762918757 Chr.11: 4577648-4577648 on Build GRCh38
rs892863352 Chr.11: 4577482-4577482 on Build GRCh38
rs368246375 Chr.11: 4577673-4577673 on Build GRCh38
rs935705066 Chr.11: 4577459-4577459 on Build GRCh38
rs79918466 Chr.11: 4577483-4577483 on Build GRCh38
rs753182141 Chr.11: 4577679-4577679 on Build GRCh38
rs371088644 Chr.11: 4577646-4577646 on Build GRCh38
rs901705441 Chr.11: 4577499-4577499 on Build GRCh38
rs1054040193 Chr.11: 4577467-4577467 on Build GRCh38
rs376039294 Chr.11: 4577466-4577466 on Build GRCh38
rs971994574 Chr.11: 4577628-4577628 on Build GRCh38
rs920110742 Chr.11: 4577598-4577598 on Build GRCh38
rs778199032 Chr.11: 4577685-4577685 on Build GRCh38
rs1016947977 Chr.11: 4577544-4577544 on Build GRCh38
rs758913819 Chr.11: 4577680-4577680 on Build GRCh38
rs768623257 Chr.11: 4577651-4577651 on Build GRCh38
rs7123071 Chr.11: 4577647-4577647 on Build GRCh38
rs556567909 Chr.11: 4577530-4577531 on Build GRCh38
rs75618637 Chr.11: 4577672-4577672 on Build GRCh38
rs878889831 Chr.11: 4577476-4577476 on Build GRCh38
rs141200768 Chr.11: 4577488-4577488 on Build GRCh38
rs964333650 Chr.11: 4577533-4577533 on Build GRCh38
rs745952666 Chr.11: 4577643-4577643 on Build GRCh38
rs1013599190 Chr.11: 4577588-4577588 on Build GRCh38
rs1532848 Chr.11: 4577532-4577532 on Build GRCh38
rs1024546757 Chr.11: 4577618-4577618 on Build GRCh38
rs554093461 Chr.11: 4577625-4577625 on Build GRCh38
rs199528419 Chr.11: 4577575-4577575 on Build GRCh38
rs750037899 Chr.11: 4577659-4577659 on Build GRCh38
rs887184365 Chr.11: 4577528-4577528 on Build GRCh38

View Full Product Details