Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 119151060-119151285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRRM4
Entrez Gene ID
84530
Gene Name
serine/arginine repetitive matrix 4
Gene Aliases
KIAA1853, MU-MB-2.76, nSR100
UniGene
Hs.744964
Gene Chromosome Location
Chr.12: 118981495-119163051 on Build GRCh38
Transcripts
NM_194286.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2174191 c.1248C>A p.Y416* Chr.12: 119151188-119151188 on Build GRCh38
2174184 c.1488A>C p.R496R Chr.12: 119151137-119151137 on Build GRCh38
5796558 c.1507C>G p.R503G Chr.12: 119151156-119151156 on Build GRCh38
3752957 c.1508G>A p.R503Q Chr.12: 119151157-119151157 on Build GRCh38
4394482 c.1179C>T p.S393S Chr.12: 119151119-119151119 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748267299 Chr.12: 119151079-119151079 on Build GRCh38
rs749575114 Chr.12: 119151172-119151172 on Build GRCh38
rs187731686 Chr.12: 119151151-119151151 on Build GRCh38
rs764791981 Chr.12: 119151224-119151224 on Build GRCh38
rs570149154 Chr.12: 119151192-119151192 on Build GRCh38
rs947142320 Chr.12: 119151188-119151188 on Build GRCh38
rs2723880 Chr.12: 119151157-119151157 on Build GRCh38
rs200067083 Chr.12: 119151112-119151112 on Build GRCh38
rs763784419 Chr.12: 119151121-119151121 on Build GRCh38
rs1022449108 Chr.12: 119151219-119151219 on Build GRCh38
rs763862350 Chr.12: 119151198-119151198 on Build GRCh38
rs902875033 Chr.12: 119151191-119151191 on Build GRCh38
rs561694968 Chr.12: 119151152-119151152 on Build GRCh38
rs550324165 Chr.12: 119151186-119151186 on Build GRCh38
rs771567762 Chr.12: 119151085-119151085 on Build GRCh38
rs369698646 Chr.12: 119151111-119151111 on Build GRCh38
rs761572995 Chr.12: 119151206-119151206 on Build GRCh38
rs778040254 Chr.12: 119151161-119151161 on Build GRCh38
rs976776462 Chr.12: 119151133-119151133 on Build GRCh38
rs202169854 Chr.12: 119151199-119151199 on Build GRCh38
rs151299260 Chr.12: 119151250-119151250 on Build GRCh38
rs749346406 Chr.12: 119151086-119151086 on Build GRCh38
rs775343743 Chr.12: 119151190-119151190 on Build GRCh38
rs373815603 Chr.12: 119151156-119151156 on Build GRCh38
rs140861808 Chr.12: 119151214-119151214 on Build GRCh38
rs770084560 Chr.12: 119151084-119151084 on Build GRCh38
rs774425136 Chr.12: 119151178-119151178 on Build GRCh38
rs952121779 Chr.12: 119151097-119151097 on Build GRCh38
rs370821738 Chr.12: 119151093-119151093 on Build GRCh38
rs756822935 Chr.12: 119151139-119151139 on Build GRCh38
rs749982641 Chr.12: 119151245-119151245 on Build GRCh38
rs369482901 Chr.12: 119151229-119151229 on Build GRCh38
rs552602160 Chr.12: 119151214-119151214 on Build GRCh38
rs200763312 Chr.12: 119151105-119151105 on Build GRCh38
rs372438926 Chr.12: 119151247-119151247 on Build GRCh38
rs56390632 Chr.12: 119151175-119151175 on Build GRCh38
rs753604308 Chr.12: 119151136-119151136 on Build GRCh38
rs767156598 Chr.12: 119151109-119151109 on Build GRCh38
rs376733585 Chr.12: 119151187-119151187 on Build GRCh38
rs564357111 Chr.12: 119151106-119151106 on Build GRCh38
rs116238652 Chr.12: 119151248-119151248 on Build GRCh38
rs755563104 Chr.12: 119151154-119151154 on Build GRCh38

View Full Product Details