Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1456237-1456476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APC2
Entrez Gene ID
10297
Gene Name
APC2, WNT signaling pathway regulator
Gene Aliases
APCL
UniGene
Hs.446376
Gene Chromosome Location
Chr.19: 1446268-1473244 on Build GRCh38
Transcripts
NM_005883.2 XM_005259475.2 XM_006722607.2 XM_006722608.3 XM_006722609.3 XM_006722610.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
474240 c.757C>A p.P253T Chr.19: 1456345-1456345 on Build GRCh38
6602080 c.738G>A p.S246S Chr.19: 1456326-1456326 on Build GRCh38
4935249 c.729G>A p.A243A Chr.19: 1456317-1456317 on Build GRCh38
6602079 c.760G>A p.E254K Chr.19: 1456348-1456348 on Build GRCh38
5114722 c.728C>T p.A243V Chr.19: 1456316-1456316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201078362 Chr.19: 1456290-1456290 on Build GRCh38
rs758655795 Chr.19: 1456307-1456307 on Build GRCh38
rs750540977 Chr.19: 1456300-1456300 on Build GRCh38
rs955056741 Chr.19: 1456361-1456361 on Build GRCh38
rs760526553 Chr.19: 1456270-1456270 on Build GRCh38
rs371479109 Chr.19: 1456409-1456409 on Build GRCh38
rs776510776 Chr.19: 1456415-1456415 on Build GRCh38
rs771826211 Chr.19: 1456338-1456338 on Build GRCh38
rs771604904 Chr.19: 1456418-1456418 on Build GRCh38
rs760337180 Chr.19: 1456262-1456289 on Build GRCh38
rs928286038 Chr.19: 1456301-1456301 on Build GRCh38
rs570789768 Chr.19: 1456312-1456312 on Build GRCh38
rs763079355 Chr.19: 1456385-1456385 on Build GRCh38
rs969808744 Chr.19: 1456287-1456287 on Build GRCh38
rs376536547 Chr.19: 1456440-1456440 on Build GRCh38
rs776674592 Chr.19: 1456351-1456351 on Build GRCh38
rs753947911 Chr.19: 1456279-1456279 on Build GRCh38
rs146579468 Chr.19: 1456384-1456384 on Build GRCh38
rs202011073 Chr.19: 1456256-1456256 on Build GRCh38
rs779710836 Chr.19: 1456433-1456433 on Build GRCh38
rs375179902 Chr.19: 1456316-1456316 on Build GRCh38
rs142788595 Chr.19: 1456415-1456415 on Build GRCh38
rs757367544 Chr.19: 1456281-1456281 on Build GRCh38
rs367708542 Chr.19: 1456392-1456392 on Build GRCh38
rs764170426 Chr.19: 1456275-1456275 on Build GRCh38
rs555858661 Chr.19: 1456345-1456345 on Build GRCh38
rs776576610 Chr.19: 1456445-1456445 on Build GRCh38
rs375320252 Chr.19: 1456455-1456455 on Build GRCh38
rs199964203 Chr.19: 1456332-1456332 on Build GRCh38
rs751845324 Chr.19: 1456391-1456391 on Build GRCh38
rs773926328 Chr.19: 1456318-1456318 on Build GRCh38
rs759466635 Chr.19: 1456436-1456436 on Build GRCh38
rs994285941 Chr.19: 1456298-1456298 on Build GRCh38
rs956151226 Chr.19: 1456432-1456432 on Build GRCh38
rs138830892 Chr.19: 1456317-1456317 on Build GRCh38
rs746693912 Chr.19: 1456438-1456438 on Build GRCh38
rs555290470 Chr.19: 1456450-1456450 on Build GRCh38
rs766134757 Chr.19: 1456263-1456268 on Build GRCh38
rs866502550 Chr.19: 1456333-1456333 on Build GRCh38
rs781567744 Chr.19: 1456395-1456395 on Build GRCh38
rs142402932 Chr.19: 1456369-1456369 on Build GRCh38
rs764005036 Chr.19: 1456348-1456348 on Build GRCh38
rs766661462 Chr.19: 1456388-1456388 on Build GRCh38
rs373843275 Chr.19: 1456411-1456411 on Build GRCh38
rs760599390 Chr.19: 1456347-1456347 on Build GRCh38
rs1029417656 Chr.19: 1456331-1456331 on Build GRCh38
rs780267185 Chr.19: 1456309-1456309 on Build GRCh38
rs1006546896 Chr.19: 1456366-1456366 on Build GRCh38
rs931466695 Chr.19: 1456374-1456374 on Build GRCh38
rs750478030 Chr.19: 1456434-1456434 on Build GRCh38
rs753146427 Chr.19: 1456398-1456398 on Build GRCh38
rs745425258 Chr.19: 1456417-1456417 on Build GRCh38

View Full Product Details