Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 115033160-115033421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSHB
Entrez Gene ID
7252
Gene Name
thyroid stimulating hormone beta
Gene Aliases
TSH-B, TSH-BETA
UniGene
Hs.406687
Gene Chromosome Location
Chr.1: 115029824-115035935 on Build GRCh38
Transcripts
NM_000549.4 XM_011542065.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1738572 c.35G>A p.G12D Chr.1: 115033397-115033397 on Build GRCh38
894302 c.45T>C p.C15C Chr.1: 115033407-115033407 on Build GRCh38
6810399 c.54G>A p.A18A Chr.1: 115033416-115033416 on Build GRCh38
4418024 c.10C>G p.L4V Chr.1: 115033372-115033372 on Build GRCh38
228799 c.23C>T p.S8F Chr.1: 115033385-115033385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982462172 Chr.1: 115033267-115033267 on Build GRCh38
rs111630216 Chr.1: 115033315-115033315 on Build GRCh38
rs753139534 Chr.1: 115033395-115033395 on Build GRCh38
rs955244432 Chr.1: 115033249-115033249 on Build GRCh38
rs774069373 Chr.1: 115033335-115033335 on Build GRCh38
rs760078611 Chr.1: 115033372-115033372 on Build GRCh38
rs573309177 Chr.1: 115033261-115033261 on Build GRCh38
rs529049577 Chr.1: 115033281-115033281 on Build GRCh38
rs768033806 Chr.1: 115033212-115033212 on Build GRCh38
rs10776792 Chr.1: 115033402-115033402 on Build GRCh38
rs780345835 Chr.1: 115033292-115033292 on Build GRCh38
rs184627760 Chr.1: 115033223-115033223 on Build GRCh38
rs140670184 Chr.1: 115033390-115033390 on Build GRCh38
rs922502137 Chr.1: 115033188-115033188 on Build GRCh38
rs372575227 Chr.1: 115033370-115033370 on Build GRCh38
rs763009083 Chr.1: 115033332-115033332 on Build GRCh38
rs544200050 Chr.1: 115033293-115033293 on Build GRCh38
rs1049422560 Chr.1: 115033283-115033283 on Build GRCh38
rs533255695 Chr.1: 115033342-115033342 on Build GRCh38
rs767114271 Chr.1: 115033358-115033358 on Build GRCh38
rs539916727 Chr.1: 115033219-115033219 on Build GRCh38
rs562763237 Chr.1: 115033334-115033334 on Build GRCh38
rs889537835 Chr.1: 115033295-115033295 on Build GRCh38

View Full Product Details