Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127867837-127868110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK1
Entrez Gene ID
203
Gene Name
adenylate kinase 1
Gene Aliases
HTL-S-58j
UniGene
Hs.175473
Gene Chromosome Location
Chr.9: 127866480-127886749 on Build GRCh38
Transcripts
NM_000476.2 NM_001318121.1 NM_001318122.1 XM_017014427.1 XM_017014428.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6307678 c.517-1G>T p.? Chr.9: 127868077-127868077 on Build GRCh38
1330720 c.544G>C p.V182L Chr.9: 127868049-127868049 on Build GRCh38
3904309 c.574G>A p.A192T Chr.9: 127868019-127868019 on Build GRCh38
1314483 c.552C>T p.S184S Chr.9: 127868041-127868041 on Build GRCh38
6597325 c.537G>T p.V179V Chr.9: 127868056-127868056 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749438351 Chr.9: 127868020-127868020 on Build GRCh38
rs4226 Chr.9: 127867954-127867954 on Build GRCh38
rs757506900 Chr.9: 127867899-127867899 on Build GRCh38
rs374909243 Chr.9: 127867995-127867995 on Build GRCh38
rs944576937 Chr.9: 127867890-127867890 on Build GRCh38
rs773386135 Chr.9: 127868059-127868059 on Build GRCh38
rs767348248 Chr.9: 127868092-127868092 on Build GRCh38
rs780800760 Chr.9: 127868007-127868007 on Build GRCh38
rs907408724 Chr.9: 127867953-127867953 on Build GRCh38
rs190164259 Chr.9: 127868058-127868058 on Build GRCh38
rs768926123 Chr.9: 127867964-127867964 on Build GRCh38
rs559035910 Chr.9: 127867880-127867880 on Build GRCh38
rs774432775 Chr.9: 127867969-127867969 on Build GRCh38
rs369560590 Chr.9: 127868003-127868003 on Build GRCh38
rs774684518 Chr.9: 127868051-127868051 on Build GRCh38
rs903740537 Chr.9: 127867871-127867871 on Build GRCh38
rs761049374 Chr.9: 127867987-127867987 on Build GRCh38
rs760524560 Chr.9: 127867963-127867963 on Build GRCh38
rs201453879 Chr.9: 127868063-127868063 on Build GRCh38
rs768691465 Chr.9: 127868028-127868028 on Build GRCh38
rs373209848 Chr.9: 127867974-127867974 on Build GRCh38
rs112904198 Chr.9: 127868079-127868079 on Build GRCh38
rs5900774 Chr.9: 127867989-127867990 on Build GRCh38
rs369008450 Chr.9: 127867973-127867973 on Build GRCh38
rs894791772 Chr.9: 127867928-127867928 on Build GRCh38
rs780530648 Chr.9: 127868019-127868019 on Build GRCh38
rs954060658 Chr.9: 127868063-127868063 on Build GRCh38
rs370638623 Chr.9: 127868014-127868014 on Build GRCh38
rs150064330 Chr.9: 127868071-127868071 on Build GRCh38
rs773795388 Chr.9: 127867977-127867977 on Build GRCh38
rs376265763 Chr.9: 127868004-127868004 on Build GRCh38
rs756817349 Chr.9: 127868087-127868087 on Build GRCh38
rs1046139189 Chr.9: 127867934-127867934 on Build GRCh38
rs755977850 Chr.9: 127868018-127868018 on Build GRCh38
rs530826715 Chr.9: 127867992-127867992 on Build GRCh38
rs551825170 Chr.9: 127867873-127867873 on Build GRCh38
rs751837494 Chr.9: 127867889-127867889 on Build GRCh38
rs746355845 Chr.9: 127867933-127867933 on Build GRCh38
rs548371283 Chr.9: 127867900-127867900 on Build GRCh38
rs377177401 Chr.9: 127867963-127867963 on Build GRCh38
rs752194585 Chr.9: 127867994-127867994 on Build GRCh38
rs377419250 Chr.9: 127868081-127868081 on Build GRCh38
rs750245032 Chr.9: 127868093-127868093 on Build GRCh38
rs761274632 Chr.9: 127868060-127868060 on Build GRCh38
rs770770312 Chr.9: 127867973-127867974 on Build GRCh38
rs527637293 Chr.9: 127867961-127867961 on Build GRCh38
rs528235859 Chr.9: 127867885-127867885 on Build GRCh38

View Full Product Details