Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 13666515-13666742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM210A
Entrez Gene ID
125228
Gene Name
family with sequence similarity 210 member A
Gene Aliases
C18orf19, HsT2329
UniGene
Hs.13034
Gene Chromosome Location
Chr.18: 13663347-13726592 on Build GRCh38
Transcripts
NM_001098801.1 NM_152352.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
986748 c.702C>A p.D234E Chr.18: 13666597-13666597 on Build GRCh38
4854238 c.733G>A p.E245K Chr.18: 13666566-13666566 on Build GRCh38
986749 c.635C>G p.T212S Chr.18: 13666664-13666664 on Build GRCh38
377522 c.589G>T p.A197S Chr.18: 13666710-13666710 on Build GRCh38
4640148 c.738A>G p.K246K Chr.18: 13666561-13666561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757945137 Chr.18: 13666656-13666656 on Build GRCh38
rs562311629 Chr.18: 13666617-13666617 on Build GRCh38
rs959542995 Chr.18: 13666609-13666609 on Build GRCh38
rs767364628 Chr.18: 13666635-13666635 on Build GRCh38
rs35023957 Chr.18: 13666603-13666603 on Build GRCh38
rs763224814 Chr.18: 13666634-13666634 on Build GRCh38
rs778201852 Chr.18: 13666658-13666658 on Build GRCh38
rs746502672 Chr.18: 13666607-13666608 on Build GRCh38
rs764696749 Chr.18: 13666649-13666649 on Build GRCh38
rs773350410 Chr.18: 13666624-13666624 on Build GRCh38
rs928581359 Chr.18: 13666703-13666703 on Build GRCh38
rs775892022 Chr.18: 13666628-13666628 on Build GRCh38
rs779628499 Chr.18: 13666543-13666543 on Build GRCh38
rs757624460 Chr.18: 13666689-13666689 on Build GRCh38
rs753277415 Chr.18: 13666549-13666549 on Build GRCh38
rs753761934 Chr.18: 13666637-13666637 on Build GRCh38
rs778595283 Chr.18: 13666591-13666591 on Build GRCh38
rs777961735 Chr.18: 13666611-13666611 on Build GRCh38
rs138091324 Chr.18: 13666595-13666595 on Build GRCh38
rs775673770 Chr.18: 13666696-13666696 on Build GRCh38
rs754480188 Chr.18: 13666561-13666561 on Build GRCh38
rs374459586 Chr.18: 13666704-13666704 on Build GRCh38
rs760751853 Chr.18: 13666673-13666673 on Build GRCh38
rs760664524 Chr.18: 13666636-13666636 on Build GRCh38
rs576279471 Chr.18: 13666627-13666627 on Build GRCh38
rs149627810 Chr.18: 13666639-13666639 on Build GRCh38
rs59211767 Chr.18: 13666618-13666618 on Build GRCh38
rs572582049 Chr.18: 13666548-13666548 on Build GRCh38
rs529431654 Chr.18: 13666690-13666690 on Build GRCh38
rs770018973 Chr.18: 13666694-13666694 on Build GRCh38
rs752367715 Chr.18: 13666650-13666650 on Build GRCh38
rs779582741 Chr.18: 13666546-13666547 on Build GRCh38
rs747275779 Chr.18: 13666680-13666680 on Build GRCh38

View Full Product Details