Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150805804-150806061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSK
Entrez Gene ID
1513
Gene Name
cathepsin K
Gene Aliases
CTS02, CTSO, CTSO1, CTSO2, PKND, PYCD
UniGene
Hs.632466
Gene Chromosome Location
Chr.1: 150796208-150808441 on Build GRCh38
Transcripts
NM_000396.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5769502 c.567C>T p.V189V Chr.1: 150805870-150805870 on Build GRCh38
5795527 c.541C>T p.R181* Chr.1: 150805896-150805896 on Build GRCh38
895615 c.369G>T p.K123N Chr.1: 150805891-150805891 on Build GRCh38
895613 c.380T>C p.V127A Chr.1: 150805880-150805880 on Build GRCh38
1747724 c.441G>C p.Q147H Chr.1: 150805996-150805996 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907283349 Chr.1: 150805969-150805969 on Build GRCh38
rs770769840 Chr.1: 150806005-150806005 on Build GRCh38
rs1057516725 Chr.1: 150805864-150805865 on Build GRCh38
rs769029170 Chr.1: 150805853-150805853 on Build GRCh38
rs373300320 Chr.1: 150806036-150806036 on Build GRCh38
rs199802458 Chr.1: 150806040-150806040 on Build GRCh38
rs759107967 Chr.1: 150805896-150805896 on Build GRCh38
rs145693431 Chr.1: 150806016-150806016 on Build GRCh38
rs137874306 Chr.1: 150805938-150805938 on Build GRCh38
rs201667695 Chr.1: 150805909-150805909 on Build GRCh38
rs777647239 Chr.1: 150805914-150805914 on Build GRCh38
rs780461752 Chr.1: 150805959-150805959 on Build GRCh38
rs1054278735 Chr.1: 150805898-150805898 on Build GRCh38
rs779047231 Chr.1: 150805948-150805948 on Build GRCh38
rs771607501 Chr.1: 150805992-150805994 on Build GRCh38
rs912531584 Chr.1: 150805978-150805978 on Build GRCh38
rs762714840 Chr.1: 150805857-150805857 on Build GRCh38
rs772730842 Chr.1: 150805919-150805919 on Build GRCh38
rs766065317 Chr.1: 150805885-150805885 on Build GRCh38
rs752649562 Chr.1: 150805902-150805902 on Build GRCh38
rs745520467 Chr.1: 150805977-150805978 on Build GRCh38
rs200337167 Chr.1: 150805855-150805855 on Build GRCh38
rs746005364 Chr.1: 150805955-150805955 on Build GRCh38
rs188468815 Chr.1: 150805852-150805852 on Build GRCh38
rs755849677 Chr.1: 150805854-150805854 on Build GRCh38
rs773943327 Chr.1: 150805970-150805971 on Build GRCh38
rs201308534 Chr.1: 150805866-150805866 on Build GRCh38
rs770173800 Chr.1: 150805837-150805842 on Build GRCh38
rs587775407 Chr.1: 150805926-150805926 on Build GRCh38
rs768992778 Chr.1: 150805986-150805986 on Build GRCh38
rs762212949 Chr.1: 150805997-150805997 on Build GRCh38
rs587742006 Chr.1: 150805965-150805965 on Build GRCh38
rs200742781 Chr.1: 150805958-150805958 on Build GRCh38
rs201028711 Chr.1: 150806030-150806030 on Build GRCh38
rs199642127 Chr.1: 150805936-150805936 on Build GRCh38
rs142306783 Chr.1: 150805903-150805903 on Build GRCh38
rs753591279 Chr.1: 150805922-150805922 on Build GRCh38
rs201962431 Chr.1: 150805894-150805894 on Build GRCh38
rs774922287 Chr.1: 150806024-150806026 on Build GRCh38
rs112299095 Chr.1: 150806018-150806018 on Build GRCh38
rs1057516839 Chr.1: 150805859-150805859 on Build GRCh38
rs777061480 Chr.1: 150805992-150805992 on Build GRCh38

View Full Product Details