Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 77211233-77211482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE8B
Entrez Gene ID
8622
Gene Name
phosphodiesterase 8B
Gene Aliases
ADSD, PPNAD3
UniGene
Hs.584830
Gene Chromosome Location
Chr.5: 77180252-77428256 on Build GRCh38
Transcripts
NM_001029851.2 NM_001029852.2 NM_001029853.2 NM_001029854.2 NM_003719.3 XM_005248621.4 XM_005248623.4 XM_005248624.4 XM_006714725.3 XM_006714726.3 XM_011543699.2 XM_011543700.2 XM_011543701.2 XM_017010003.1 XM_017010005.1 XM_017010006.1 XM_017010007.1 XM_017010008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3074121 c.310G>A p.E104K Chr.5: 77211235-77211235 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1022918191 Chr.5: 77211285-77211285 on Build GRCh38
rs867926021 Chr.5: 77211310-77211310 on Build GRCh38
rs994673272 Chr.5: 77211417-77211417 on Build GRCh38
rs867301753 Chr.5: 77211436-77211436 on Build GRCh38
rs763086213 Chr.5: 77211305-77211308 on Build GRCh38
rs377278594 Chr.5: 77211429-77211429 on Build GRCh38
rs762032173 Chr.5: 77211292-77211292 on Build GRCh38
rs781062791 Chr.5: 77211299-77211299 on Build GRCh38
rs756962159 Chr.5: 77211295-77211295 on Build GRCh38
rs754040283 Chr.5: 77211272-77211272 on Build GRCh38
rs755791907 Chr.5: 77211303-77211303 on Build GRCh38
rs763977111 Chr.5: 77211335-77211335 on Build GRCh38
rs933904722 Chr.5: 77211461-77211461 on Build GRCh38
rs750567358 Chr.5: 77211254-77211254 on Build GRCh38
rs764174475 Chr.5: 77211279-77211279 on Build GRCh38
rs905142886 Chr.5: 77211325-77211325 on Build GRCh38
rs370564764 Chr.5: 77211453-77211453 on Build GRCh38
rs997861894 Chr.5: 77211258-77211258 on Build GRCh38
rs748851253 Chr.5: 77211312-77211312 on Build GRCh38
rs572264419 Chr.5: 77211340-77211340 on Build GRCh38
rs766500440 Chr.5: 77211268-77211268 on Build GRCh38
rs1030641299 Chr.5: 77211270-77211270 on Build GRCh38
rs758398562 Chr.5: 77211276-77211276 on Build GRCh38
rs760965908 Chr.5: 77211256-77211256 on Build GRCh38
rs942515554 Chr.5: 77211421-77211421 on Build GRCh38
rs751366731 Chr.5: 77211293-77211293 on Build GRCh38
rs773275775 Chr.5: 77211301-77211301 on Build GRCh38
rs765541325 Chr.5: 77211298-77211299 on Build GRCh38
rs192048910 Chr.5: 77211423-77211423 on Build GRCh38
rs960014346 Chr.5: 77211402-77211402 on Build GRCh38
rs762898051 Chr.5: 77211300-77211300 on Build GRCh38
rs766431297 Chr.5: 77211311-77211322 on Build GRCh38
rs919246511 Chr.5: 77211347-77211347 on Build GRCh38
rs779533364 Chr.5: 77211308-77211308 on Build GRCh38
rs984886747 Chr.5: 77211413-77211413 on Build GRCh38
rs541188344 Chr.5: 77211364-77211364 on Build GRCh38
rs1010698072 Chr.5: 77211294-77211294 on Build GRCh38

View Full Product Details