Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 54277710-54277935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLI
Entrez Gene ID
11201
Gene Name
DNA polymerase iota
Gene Aliases
RAD30B, RAD3OB
UniGene
Hs.438533
Gene Chromosome Location
Chr.18: 54269479-54321266 on Build GRCh38
Transcripts
NM_007195.2 XM_005258188.3 XM_005258192.4 XM_006722378.2 XM_011525796.2 XM_011525797.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3796450 c.484G>C p.E162Q Chr.18: 54277780-54277780 on Build GRCh38
3362554 c.551A>G p.N184S Chr.18: 54277847-54277847 on Build GRCh38
5854272 c.496C>T p.Q166* Chr.18: 54277792-54277792 on Build GRCh38
3764670 c.538G>A p.G180S Chr.18: 54277834-54277834 on Build GRCh38
988958 c.343G>T p.E115* Chr.18: 54277714-54277714 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757357753 Chr.18: 54277862-54277862 on Build GRCh38
rs771845172 Chr.18: 54277902-54277902 on Build GRCh38
rs3218785 Chr.18: 54277849-54277851 on Build GRCh38
rs752557345 Chr.18: 54277852-54277852 on Build GRCh38
rs754677413 Chr.18: 54277814-54277814 on Build GRCh38
rs372390888 Chr.18: 54277833-54277833 on Build GRCh38
rs955987774 Chr.18: 54277824-54277824 on Build GRCh38
rs142549889 Chr.18: 54277832-54277832 on Build GRCh38
rs369477870 Chr.18: 54277797-54277797 on Build GRCh38
rs150991803 Chr.18: 54277846-54277846 on Build GRCh38
rs146652056 Chr.18: 54277798-54277798 on Build GRCh38
rs745954398 Chr.18: 54277789-54277789 on Build GRCh38
rs749884527 Chr.18: 54277828-54277828 on Build GRCh38
rs140298800 Chr.18: 54277820-54277820 on Build GRCh38
rs17470325 Chr.18: 54277895-54277895 on Build GRCh38
rs771213123 Chr.18: 54277744-54277744 on Build GRCh38
rs1031004899 Chr.18: 54277874-54277874 on Build GRCh38
rs774811562 Chr.18: 54277815-54277815 on Build GRCh38
rs572459890 Chr.18: 54277796-54277796 on Build GRCh38
rs610281 Chr.18: 54277889-54277889 on Build GRCh38
rs370594980 Chr.18: 54277816-54277816 on Build GRCh38
rs778530301 Chr.18: 54277848-54277848 on Build GRCh38
rs147513004 Chr.18: 54277873-54277873 on Build GRCh38
rs774519655 Chr.18: 54277892-54277892 on Build GRCh38
rs745938963 Chr.18: 54277871-54277871 on Build GRCh38
rs748548336 Chr.18: 54277898-54277898 on Build GRCh38
rs767290352 Chr.18: 54277821-54277821 on Build GRCh38
rs753764414 Chr.18: 54277837-54277837 on Build GRCh38
rs781193036 Chr.18: 54277770-54277770 on Build GRCh38
rs375746397 Chr.18: 54277864-54277864 on Build GRCh38
rs878918641 Chr.18: 54277778-54277778 on Build GRCh38
rs143166193 Chr.18: 54277788-54277788 on Build GRCh38
rs779818486 Chr.18: 54277830-54277830 on Build GRCh38
rs750229427 Chr.18: 54277767-54277767 on Build GRCh38
rs373392550 Chr.18: 54277883-54277883 on Build GRCh38

View Full Product Details