Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 128794231-128794496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC136
Entrez Gene ID
64753
Gene Name
coiled-coil domain containing 136
Gene Aliases
NAG6
UniGene
Hs.521178
Gene Chromosome Location
Chr.7: 128791410-128822133 on Build GRCh38
Transcripts
NM_001201372.1 NM_022742.4 XM_005250534.3 XM_005250538.1 XM_011516483.1 XM_011516484.1 XM_011516485.1 XM_011516486.1 XM_011516487.2 XM_011516488.2 XM_011516489.1 XM_011516490.2 XM_017012532.1 XM_017012533.1 XM_017012534.1 XM_017012535.1 XM_017012536.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1085279 c.18G>A p.G6G Chr.7: 128794349-128794349 on Build GRCh38
5900083 c.167-4C>T p.? Chr.7: 128794344-128794344 on Build GRCh38
1085282 c.20A>G p.E7G Chr.7: 128794351-128794351 on Build GRCh38
5723895 c.409G>A p.E137K Chr.7: 128794392-128794392 on Build GRCh38
5738146 c.494G>A p.R165Q Chr.7: 128794477-128794477 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749488173 Chr.7: 128794389-128794397 on Build GRCh38
rs774361033 Chr.7: 128794401-128794403 on Build GRCh38
rs759425198 Chr.7: 128794370-128794370 on Build GRCh38
rs373132744 Chr.7: 128794313-128794313 on Build GRCh38
rs77223612 Chr.7: 128794298-128794298 on Build GRCh38
rs762696240 Chr.7: 128794417-128794417 on Build GRCh38
rs138004090 Chr.7: 128794311-128794311 on Build GRCh38
rs764371198 Chr.7: 128794420-128794420 on Build GRCh38
rs576388205 Chr.7: 128794476-128794476 on Build GRCh38
rs751367998 Chr.7: 128794392-128794403 on Build GRCh38
rs375637274 Chr.7: 128794411-128794411 on Build GRCh38
rs554434384 Chr.7: 128794335-128794335 on Build GRCh38
rs979716082 Chr.7: 128794353-128794353 on Build GRCh38
rs760167467 Chr.7: 128794339-128794339 on Build GRCh38
rs925516440 Chr.7: 128794413-128794413 on Build GRCh38
rs371358536 Chr.7: 128794453-128794453 on Build GRCh38
rs996652241 Chr.7: 128794297-128794297 on Build GRCh38
rs1013915830 Chr.7: 128794361-128794361 on Build GRCh38
rs760625719 Chr.7: 128794413-128794415 on Build GRCh38
rs901068138 Chr.7: 128794263-128794263 on Build GRCh38
rs758596458 Chr.7: 128794255-128794255 on Build GRCh38
rs369244647 Chr.7: 128794337-128794337 on Build GRCh38
rs780358741 Chr.7: 128794376-128794377 on Build GRCh38
rs770367591 Chr.7: 128794341-128794341 on Build GRCh38
rs757044539 Chr.7: 128794428-128794428 on Build GRCh38
rs751657355 Chr.7: 128794295-128794295 on Build GRCh38
rs539120350 Chr.7: 128794377-128794382 on Build GRCh38
rs377239628 Chr.7: 128794327-128794327 on Build GRCh38
rs372607897 Chr.7: 128794368-128794368 on Build GRCh38
rs376755462 Chr.7: 128794445-128794445 on Build GRCh38
rs777906366 Chr.7: 128794258-128794258 on Build GRCh38
rs143537881 Chr.7: 128794444-128794444 on Build GRCh38
rs1024578812 Chr.7: 128794366-128794366 on Build GRCh38
rs201497246 Chr.7: 128794431-128794431 on Build GRCh38
rs192348781 Chr.7: 128794397-128794397 on Build GRCh38
rs947003064 Chr.7: 128794336-128794336 on Build GRCh38
rs768765556 Chr.7: 128794395-128794397 on Build GRCh38
rs750554557 Chr.7: 128794449-128794449 on Build GRCh38
rs550457700 Chr.7: 128794296-128794296 on Build GRCh38
rs374254797 Chr.7: 128794454-128794454 on Build GRCh38
rs922525223 Chr.7: 128794439-128794439 on Build GRCh38

View Full Product Details