Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 100337065-100337308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WARS
Entrez Gene ID
7453
Gene Name
tryptophanyl-tRNA synthetase
Gene Aliases
GAMMA-2, IFI53, IFP53
UniGene
Hs.497599
Gene Chromosome Location
Chr.14: 100333788-100376343 on Build GRCh38
Transcripts
NM_004184.3 NM_173701.1 NM_213645.1 NM_213646.1 XM_005268044.3 XM_006720249.3 XM_011537133.2 XM_011537135.2 XM_011537136.2 XM_017021627.1 XM_017021628.1 XM_017021629.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6246837 c.1179C>A p.N393K Chr.14: 100337137-100337137 on Build GRCh38
3419621 c.1127C>T p.A376V Chr.14: 100337189-100337189 on Build GRCh38
5595554 c.1215C>T p.F405F Chr.14: 100337101-100337101 on Build GRCh38
6074810 c.1250G>T p.R417M Chr.14: 100337066-100337066 on Build GRCh38
4049072 c.1207C>A p.L403M Chr.14: 100337109-100337109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs909129151 Chr.14: 100337105-100337105 on Build GRCh38
rs1054605078 Chr.14: 100337088-100337088 on Build GRCh38
rs750292885 Chr.14: 100337242-100337242 on Build GRCh38
rs2234529 Chr.14: 100337086-100337086 on Build GRCh38
rs941951647 Chr.14: 100337110-100337110 on Build GRCh38
rs78582306 Chr.14: 100337278-100337278 on Build GRCh38
rs539900924 Chr.14: 100337227-100337227 on Build GRCh38
rs760155915 Chr.14: 100337189-100337189 on Build GRCh38
rs751363632 Chr.14: 100337100-100337100 on Build GRCh38
rs775081453 Chr.14: 100337206-100337206 on Build GRCh38
rs762435047 Chr.14: 100337207-100337207 on Build GRCh38
rs367840580 Chr.14: 100337225-100337225 on Build GRCh38
rs1036427682 Chr.14: 100337114-100337114 on Build GRCh38
rs373660575 Chr.14: 100337098-100337098 on Build GRCh38
rs747369344 Chr.14: 100337172-100337172 on Build GRCh38
rs755007002 Chr.14: 100337147-100337147 on Build GRCh38
rs781084155 Chr.14: 100337125-100337125 on Build GRCh38
rs150963629 Chr.14: 100337085-100337085 on Build GRCh38
rs778134498 Chr.14: 100337224-100337225 on Build GRCh38
rs752358226 Chr.14: 100337089-100337089 on Build GRCh38
rs149689884 Chr.14: 100337161-100337161 on Build GRCh38
rs757041761 Chr.14: 100337122-100337122 on Build GRCh38
rs937116227 Chr.14: 100337095-100337095 on Build GRCh38
rs191685492 Chr.14: 100337257-100337257 on Build GRCh38
rs553841509 Chr.14: 100337244-100337244 on Build GRCh38
rs1803791 Chr.14: 100337144-100337144 on Build GRCh38
rs531022944 Chr.14: 100337188-100337188 on Build GRCh38
rs763768262 Chr.14: 100337219-100337219 on Build GRCh38
rs772322833 Chr.14: 100337164-100337164 on Build GRCh38
rs574471285 Chr.14: 100337130-100337130 on Build GRCh38
rs748258652 Chr.14: 100337163-100337163 on Build GRCh38
rs758076089 Chr.14: 100337094-100337094 on Build GRCh38
rs764777357 Chr.14: 100337194-100337194 on Build GRCh38
rs771472910 Chr.14: 100337179-100337179 on Build GRCh38

View Full Product Details