Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 182728638-182728911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC10
Entrez Gene ID
54431
Gene Name
DnaJ heat shock protein family (Hsp40) member C10
Gene Aliases
ERdj5, JPDI, MTHr, PDIA19
UniGene
Hs.516632
Gene Chromosome Location
Chr.2: 182716041-182780025 on Build GRCh38
Transcripts
NM_001271581.1 NM_018981.2 NR_073365.1 NR_073366.1 NR_073367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5793475 c.504G>C p.W168C Chr.2: 182728865-182728865 on Build GRCh38
1204185 c.539G>A p.R180Q Chr.2: 182728900-182728900 on Build GRCh38
1494695 c.490T>A p.L164I Chr.2: 182728647-182728647 on Build GRCh38
3574997 c.547G>A p.A183T Chr.2: 182728908-182728908 on Build GRCh38
3673573 c.498C>A p.P166P Chr.2: 182728655-182728655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200601570 Chr.2: 182728725-182728725 on Build GRCh38
rs1043241241 Chr.2: 182728783-182728783 on Build GRCh38
rs759657664 Chr.2: 182728876-182728876 on Build GRCh38
rs763343912 Chr.2: 182728862-182728862 on Build GRCh38
rs398080876 Chr.2: 182728732-182728732 on Build GRCh38
rs770379335 Chr.2: 182728829-182728829 on Build GRCh38
rs548090700 Chr.2: 182728680-182728680 on Build GRCh38
rs543671525 Chr.2: 182728771-182728771 on Build GRCh38
rs377220389 Chr.2: 182728707-182728707 on Build GRCh38
rs778557633 Chr.2: 182728708-182728708 on Build GRCh38
rs943139108 Chr.2: 182728775-182728775 on Build GRCh38
rs1052992858 Chr.2: 182728822-182728822 on Build GRCh38
rs1055484440 Chr.2: 182728843-182728843 on Build GRCh38
rs748698791 Chr.2: 182728820-182728820 on Build GRCh38
rs3832045 Chr.2: 182728725-182728725 on Build GRCh38
rs763961102 Chr.2: 182728881-182728881 on Build GRCh38
rs774694747 Chr.2: 182728868-182728868 on Build GRCh38
rs539836910 Chr.2: 182728791-182728791 on Build GRCh38
rs749033212 Chr.2: 182728700-182728700 on Build GRCh38
rs372066243 Chr.2: 182728710-182728710 on Build GRCh38
rs752731034 Chr.2: 182728679-182728679 on Build GRCh38
rs777574339 Chr.2: 182728684-182728684 on Build GRCh38
rs755153648 Chr.2: 182728663-182728663 on Build GRCh38
rs934561156 Chr.2: 182728804-182728804 on Build GRCh38
rs375770533 Chr.2: 182728815-182728815 on Build GRCh38
rs936992734 Chr.2: 182728830-182728830 on Build GRCh38
rs911281115 Chr.2: 182728698-182728698 on Build GRCh38
rs200031184 Chr.2: 182728878-182728878 on Build GRCh38
rs762643910 Chr.2: 182728872-182728873 on Build GRCh38
rs913162270 Chr.2: 182728767-182728767 on Build GRCh38
rs193261690 Chr.2: 182728667-182728667 on Build GRCh38
rs751595885 Chr.2: 182728661-182728661 on Build GRCh38
rs752733479 Chr.2: 182728879-182728879 on Build GRCh38
rs756067875 Chr.2: 182728880-182728880 on Build GRCh38
rs570009789 Chr.2: 182728866-182728866 on Build GRCh38
rs773729343 Chr.2: 182728844-182728844 on Build GRCh38
rs989946027 Chr.2: 182728750-182728750 on Build GRCh38
rs1045441790 Chr.2: 182728811-182728811 on Build GRCh38
rs141717303 Chr.2: 182728888-182728888 on Build GRCh38

View Full Product Details