Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27777076-27777301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESCO2
Entrez Gene ID
157570
Gene Name
establishment of sister chromatid cohesion N-acetyltransferase 2
Gene Aliases
2410004I17Rik, EFO2, RBS
UniGene
Hs.99480
Gene Chromosome Location
Chr.8: 27771949-27812404 on Build GRCh38
Transcripts
NM_001017420.2 XM_011544421.2 XM_011544422.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2788148 c.798A>G p.L266L Chr.8: 27777106-27777106 on Build GRCh38
275007 c.833G>A p.S278N Chr.8: 27777141-27777141 on Build GRCh38
1319721 c.773G>A p.R258K Chr.8: 27777081-27777081 on Build GRCh38
2788149 c.858A>G p.I286M Chr.8: 27777166-27777166 on Build GRCh38
280863 c.804G>A p.E268E Chr.8: 27777112-27777112 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767361080 Chr.8: 27777209-27777209 on Build GRCh38
rs1045418056 Chr.8: 27777270-27777270 on Build GRCh38
rs143518212 Chr.8: 27777123-27777123 on Build GRCh38
rs976489585 Chr.8: 27777157-27777157 on Build GRCh38
rs759514174 Chr.8: 27777202-27777202 on Build GRCh38
rs777530905 Chr.8: 27777233-27777233 on Build GRCh38
rs761404241 Chr.8: 27777152-27777155 on Build GRCh38
rs886062859 Chr.8: 27777143-27777143 on Build GRCh38
rs755605081 Chr.8: 27777229-27777229 on Build GRCh38
rs775568427 Chr.8: 27777117-27777117 on Build GRCh38
rs766142335 Chr.8: 27777185-27777185 on Build GRCh38
rs776867846 Chr.8: 27777138-27777138 on Build GRCh38
rs966638854 Chr.8: 27777142-27777142 on Build GRCh38
rs1026428470 Chr.8: 27777252-27777252 on Build GRCh38
rs986679958 Chr.8: 27777116-27777116 on Build GRCh38
rs762060061 Chr.8: 27777141-27777141 on Build GRCh38
rs766874679 Chr.8: 27777159-27777160 on Build GRCh38
rs751370328 Chr.8: 27777191-27777191 on Build GRCh38
rs746538314 Chr.8: 27777221-27777221 on Build GRCh38
rs946638966 Chr.8: 27777265-27777265 on Build GRCh38
rs773803640 Chr.8: 27777160-27777161 on Build GRCh38
rs761040940 Chr.8: 27777214-27777215 on Build GRCh38
rs111667783 Chr.8: 27777128-27777128 on Build GRCh38
rs532061731 Chr.8: 27777109-27777109 on Build GRCh38
rs752197732 Chr.8: 27777222-27777222 on Build GRCh38
rs746369359 Chr.8: 27777096-27777096 on Build GRCh38

View Full Product Details