Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62835796-62836029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR74
Entrez Gene ID
54663
Gene Name
WD repeat domain 74
Gene Aliases
-
UniGene
Hs.654620
Gene Chromosome Location
Chr.11: 62832905-62841809 on Build GRCh38
Transcripts
NM_001307977.1 NM_018093.3 XM_005274055.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2039837 c.409C>T p.R137C Chr.11: 62835802-62835802 on Build GRCh38
429479 c.349A>T p.K117* Chr.11: 62835981-62835981 on Build GRCh38
429478 c.349A>T p.K117* Chr.11: 62835981-62835981 on Build GRCh38
2039836 c.409C>T p.R137C Chr.11: 62835802-62835802 on Build GRCh38
7268991 c.411C>T p.R137R Chr.11: 62835800-62835800 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764519354 Chr.11: 62835878-62835878 on Build GRCh38
rs749952763 Chr.11: 62835885-62835885 on Build GRCh38
rs760777558 Chr.11: 62835844-62835844 on Build GRCh38
rs776765603 Chr.11: 62835991-62835991 on Build GRCh38
rs755631894 Chr.11: 62835974-62835975 on Build GRCh38
rs776906026 Chr.11: 62835856-62835856 on Build GRCh38
rs372232991 Chr.11: 62835987-62835987 on Build GRCh38
rs779132114 Chr.11: 62835912-62835912 on Build GRCh38
rs771101254 Chr.11: 62835988-62835988 on Build GRCh38
rs754997099 Chr.11: 62835911-62835911 on Build GRCh38
rs747827129 Chr.11: 62835932-62835932 on Build GRCh38
rs780853195 Chr.11: 62835848-62835852 on Build GRCh38
rs773426645 Chr.11: 62835821-62835821 on Build GRCh38
rs367979926 Chr.11: 62835858-62835858 on Build GRCh38
rs778797002 Chr.11: 62835929-62835929 on Build GRCh38
rs772145616 Chr.11: 62835939-62835939 on Build GRCh38
rs368787398 Chr.11: 62835883-62835883 on Build GRCh38
rs979333182 Chr.11: 62835899-62835899 on Build GRCh38
rs551642087 Chr.11: 62835999-62836000 on Build GRCh38
rs745473906 Chr.11: 62835934-62835937 on Build GRCh38
rs192301669 Chr.11: 62835818-62835818 on Build GRCh38
rs201394009 Chr.11: 62835966-62835966 on Build GRCh38
rs755611722 Chr.11: 62835890-62835890 on Build GRCh38
rs771336264 Chr.11: 62835852-62835852 on Build GRCh38
rs765613667 Chr.11: 62835866-62835866 on Build GRCh38
rs754534616 Chr.11: 62835928-62835928 on Build GRCh38
rs890090790 Chr.11: 62835904-62835904 on Build GRCh38
rs767033457 Chr.11: 62835902-62835902 on Build GRCh38
rs202145747 Chr.11: 62835867-62835867 on Build GRCh38
rs375142835 Chr.11: 62835871-62835871 on Build GRCh38

View Full Product Details