Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 102821682-102821933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIVM
Entrez Gene ID
54841
Gene Name
basic, immunoglobulin-like variable motif containing
Gene Aliases
-
UniGene
Hs.288809
Gene Chromosome Location
Chr.13: 102799049-102841538 on Build GRCh38
Transcripts
NM_001159596.1 NM_017693.3 NM_001204425.1

Gene Symbol
BIVM-ERCC5
Entrez Gene ID
100533467
Gene Name
BIVM-ERCC5 readthrough
Gene Aliases
ERCC5-202
UniGene
Hs.258429
Gene Chromosome Location
Chr.13: 102807146-102876001 on Build GRCh38
Transcripts
NM_001159596.1 NM_017693.3 NM_001204425.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7357552 c.55G>T p.G19C Chr.13: 102821783-102821783 on Build GRCh38
6275930 c.84G>T p.R28S Chr.13: 102821812-102821812 on Build GRCh38
5727402 c.779C>T p.P260L Chr.13: 102821820-102821820 on Build GRCh38
5747511 c.751C>T p.P251S Chr.13: 102821792-102821792 on Build GRCh38
4558978 c.100G>A p.G34R Chr.13: 102821828-102821828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377757672 Chr.13: 102821743-102821743 on Build GRCh38
rs200740122 Chr.13: 102821845-102821845 on Build GRCh38
rs780995847 Chr.13: 102821860-102821860 on Build GRCh38
rs759053083 Chr.13: 102821720-102821720 on Build GRCh38
rs770268882 Chr.13: 102821843-102821843 on Build GRCh38
rs750446994 Chr.13: 102821785-102821785 on Build GRCh38
rs755285985 Chr.13: 102821826-102821826 on Build GRCh38
rs765095427 Chr.13: 102821768-102821768 on Build GRCh38
rs748477460 Chr.13: 102821830-102821830 on Build GRCh38
rs781573321 Chr.13: 102821827-102821827 on Build GRCh38
rs758541973 Chr.13: 102821789-102821789 on Build GRCh38
rs763844307 Chr.13: 102821751-102821751 on Build GRCh38
rs1032711909 Chr.13: 102821900-102821900 on Build GRCh38
rs771547079 Chr.13: 102821857-102821857 on Build GRCh38
rs190443957 Chr.13: 102821876-102821876 on Build GRCh38
rs1015925938 Chr.13: 102821784-102821784 on Build GRCh38
rs760367516 Chr.13: 102821744-102821744 on Build GRCh38
rs565981886 Chr.13: 102821753-102821753 on Build GRCh38
rs749852024 Chr.13: 102821848-102821848 on Build GRCh38
rs762343841 Chr.13: 102821707-102821707 on Build GRCh38
rs763673800 Chr.13: 102821869-102821869 on Build GRCh38
rs375594316 Chr.13: 102821715-102821715 on Build GRCh38
rs750368663 Chr.13: 102821896-102821896 on Build GRCh38
rs771771537 Chr.13: 102821894-102821896 on Build GRCh38
rs375916066 Chr.13: 102821874-102821874 on Build GRCh38
rs979364410 Chr.13: 102821893-102821893 on Build GRCh38
rs987274649 Chr.13: 102821891-102821891 on Build GRCh38
rs775019587 Chr.13: 102821858-102821858 on Build GRCh38
rs1025318217 Chr.13: 102821783-102821783 on Build GRCh38
rs200628135 Chr.13: 102821878-102821878 on Build GRCh38
rs994201091 Chr.13: 102821774-102821774 on Build GRCh38
rs745783186 Chr.13: 102821760-102821762 on Build GRCh38
rs773854671 Chr.13: 102821718-102821718 on Build GRCh38
rs747204924 Chr.13: 102821810-102821810 on Build GRCh38
rs1003059251 Chr.13: 102821721-102821721 on Build GRCh38
rs758453596 Chr.13: 102821897-102821897 on Build GRCh38
rs780334304 Chr.13: 102821792-102821792 on Build GRCh38
rs371238043 Chr.13: 102821875-102821875 on Build GRCh38
rs1004915672 Chr.13: 102821780-102821780 on Build GRCh38

View Full Product Details