Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49704603-49704866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPT1C
Entrez Gene ID
126129
Gene Name
carnitine palmitoyltransferase 1C
Gene Aliases
CATL1, CPT1-B, CPT1P, CPTI-B, CPTIC, SPG73
UniGene
Hs.112195
Gene Chromosome Location
Chr.19: 49690672-49713731 on Build GRCh38
Transcripts
NM_001136052.2 NM_001199752.2 NM_001199753.1 NM_152359.2 NR_108072.1 XM_005258505.2 XM_005258506.4 XM_006723009.3 XM_011526438.2 XM_011526439.2 XM_011526440.2 XM_017026265.1 XM_017026266.1 XM_017026267.1 XM_017026268.1 XM_017026269.1 XM_017026270.1 XM_017026271.1 XM_017026272.1 XM_017026273.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6634974 c.768G>A p.M256I Chr.19: 49704784-49704784 on Build GRCh38
137176 c.731C>T p.S244F Chr.19: 49704747-49704747 on Build GRCh38
4754947 c.733C>T p.R245* Chr.19: 49704749-49704749 on Build GRCh38
6306108 c.728G>T p.R243L Chr.19: 49704744-49704744 on Build GRCh38
4435644 c.741G>T p.P247P Chr.19: 49704757-49704757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028433088 Chr.19: 49704790-49704790 on Build GRCh38
rs1011670181 Chr.19: 49704777-49704777 on Build GRCh38
rs759384307 Chr.19: 49704831-49704831 on Build GRCh38
rs369066476 Chr.19: 49704834-49704834 on Build GRCh38
rs371889665 Chr.19: 49704838-49704838 on Build GRCh38
rs769337623 Chr.19: 49704701-49704701 on Build GRCh38
rs764347500 Chr.19: 49704743-49704743 on Build GRCh38
rs755374253 Chr.19: 49704766-49704766 on Build GRCh38
rs765359581 Chr.19: 49704756-49704756 on Build GRCh38
rs768458124 Chr.19: 49704685-49704685 on Build GRCh38
rs774155314 Chr.19: 49704712-49704712 on Build GRCh38
rs931122771 Chr.19: 49704662-49704662 on Build GRCh38
rs894535879 Chr.19: 49704698-49704698 on Build GRCh38
rs776554953 Chr.19: 49704631-49704631 on Build GRCh38
rs765878325 Chr.19: 49704693-49704693 on Build GRCh38
rs780478427 Chr.19: 49704678-49704678 on Build GRCh38
rs750197748 Chr.19: 49704833-49704833 on Build GRCh38
rs375954352 Chr.19: 49704836-49704836 on Build GRCh38
rs747421409 Chr.19: 49704763-49704763 on Build GRCh38
rs754045165 Chr.19: 49704749-49704749 on Build GRCh38
rs747713339 Chr.19: 49704700-49704700 on Build GRCh38
rs770939318 Chr.19: 49704819-49704819 on Build GRCh38
rs758760566 Chr.19: 49704665-49704665 on Build GRCh38
rs747802823 Chr.19: 49704801-49704801 on Build GRCh38
rs774247502 Chr.19: 49704829-49704829 on Build GRCh38
rs772792463 Chr.19: 49704810-49704810 on Build GRCh38
rs941434413 Chr.19: 49704636-49704636 on Build GRCh38
rs746377630 Chr.19: 49704659-49704659 on Build GRCh38
rs762953553 Chr.19: 49704703-49704703 on Build GRCh38
rs780381980 Chr.19: 49704762-49704762 on Build GRCh38
rs757393571 Chr.19: 49704750-49704750 on Build GRCh38
rs750408494 Chr.19: 49704757-49704757 on Build GRCh38
rs146091196 Chr.19: 49704744-49704744 on Build GRCh38
rs769264627 Chr.19: 49704806-49704806 on Build GRCh38
rs908672676 Chr.19: 49704633-49704633 on Build GRCh38
rs919831245 Chr.19: 49704847-49704847 on Build GRCh38
rs369637900 Chr.19: 49704784-49704784 on Build GRCh38
rs983231780 Chr.19: 49704630-49704630 on Build GRCh38
rs766171809 Chr.19: 49704704-49704704 on Build GRCh38
rs761907400 Chr.19: 49704661-49704661 on Build GRCh38
rs950582316 Chr.19: 49704621-49704621 on Build GRCh38
rs200029616 Chr.19: 49704816-49704816 on Build GRCh38
rs1045799752 Chr.19: 49704706-49704706 on Build GRCh38
rs144205321 Chr.19: 49704737-49704737 on Build GRCh38
rs747325421 Chr.19: 49704679-49704679 on Build GRCh38
rs565574975 Chr.19: 49704702-49704702 on Build GRCh38
rs953821911 Chr.19: 49704795-49704795 on Build GRCh38
rs762104762 Chr.19: 49704837-49704837 on Build GRCh38

View Full Product Details