Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 20619022-20619265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HS1BP3
Entrez Gene ID
64342
Gene Name
HCLS1 binding protein 3
Gene Aliases
ETM2, HS1-BP3
UniGene
Hs.531785
Gene Chromosome Location
Chr.2: 20553861-20651661 on Build GRCh38
Transcripts
NM_022460.3 XM_017004696.1 XM_017004697.1 XM_017004698.1 XM_017004699.1 XM_017004700.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
79368 c.935T>A p.L312* Chr.2: 20619231-20619231 on Build GRCh38
6090377 c.954G>T p.L318L Chr.2: 20619212-20619212 on Build GRCh38
4750937 c.1111G>A p.E371K Chr.2: 20619055-20619055 on Build GRCh38
6682192 c.1013C>T p.P338L Chr.2: 20619153-20619153 on Build GRCh38
5471400 c.1085C>T p.P362L Chr.2: 20619081-20619081 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764489015 Chr.2: 20619187-20619187 on Build GRCh38
rs754355167 Chr.2: 20619106-20619106 on Build GRCh38
rs906312817 Chr.2: 20619070-20619070 on Build GRCh38
rs921106545 Chr.2: 20619145-20619145 on Build GRCh38
rs577242302 Chr.2: 20619128-20619128 on Build GRCh38
rs1023743929 Chr.2: 20619152-20619152 on Build GRCh38
rs767897962 Chr.2: 20619153-20619153 on Build GRCh38
rs758110778 Chr.2: 20619120-20619120 on Build GRCh38
rs944491265 Chr.2: 20619088-20619088 on Build GRCh38
rs139078947 Chr.2: 20619149-20619149 on Build GRCh38
rs757841128 Chr.2: 20619211-20619211 on Build GRCh38
rs776504819 Chr.2: 20619068-20619068 on Build GRCh38
rs921793980 Chr.2: 20619098-20619098 on Build GRCh38
rs756724155 Chr.2: 20619111-20619111 on Build GRCh38
rs765630459 Chr.2: 20619213-20619213 on Build GRCh38
rs1035863225 Chr.2: 20619103-20619103 on Build GRCh38
rs974155817 Chr.2: 20619051-20619051 on Build GRCh38
rs774561974 Chr.2: 20619144-20619144 on Build GRCh38
rs775987515 Chr.2: 20619041-20619041 on Build GRCh38
rs1056689766 Chr.2: 20619127-20619127 on Build GRCh38
rs753149002 Chr.2: 20619091-20619091 on Build GRCh38
rs1021923324 Chr.2: 20619119-20619119 on Build GRCh38
rs745357270 Chr.2: 20619117-20619117 on Build GRCh38
rs779710138 Chr.2: 20619244-20619244 on Build GRCh38
rs918587712 Chr.2: 20619055-20619055 on Build GRCh38
rs759865091 Chr.2: 20619081-20619081 on Build GRCh38
rs372443182 Chr.2: 20619124-20619124 on Build GRCh38
rs754315788 Chr.2: 20619205-20619205 on Build GRCh38
rs183773608 Chr.2: 20619129-20619129 on Build GRCh38
rs775804097 Chr.2: 20619171-20619171 on Build GRCh38
rs767624861 Chr.2: 20619086-20619086 on Build GRCh38
rs374026872 Chr.2: 20619116-20619116 on Build GRCh38
rs34708707 Chr.2: 20619080-20619080 on Build GRCh38
rs866466010 Chr.2: 20619087-20619091 on Build GRCh38
rs749997504 Chr.2: 20619220-20619220 on Build GRCh38
rs756522204 Chr.2: 20619099-20619099 on Build GRCh38
rs939743128 Chr.2: 20619045-20619045 on Build GRCh38
rs898700438 Chr.2: 20619188-20619188 on Build GRCh38
rs764718066 Chr.2: 20619102-20619102 on Build GRCh38
rs757795909 Chr.2: 20619224-20619224 on Build GRCh38
rs35579164 Chr.2: 20619123-20619123 on Build GRCh38
rs761155787 Chr.2: 20619183-20619183 on Build GRCh38
rs760072015 Chr.2: 20619192-20619196 on Build GRCh38
rs111886213 Chr.2: 20619056-20619056 on Build GRCh38
rs139786951 Chr.2: 20619122-20619122 on Build GRCh38
rs983347057 Chr.2: 20619079-20619079 on Build GRCh38

View Full Product Details