Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153539987-153540254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
S100A5
Entrez Gene ID
6276
Gene Name
S100 calcium binding protein A5
Gene Aliases
S100D
UniGene
Hs.2960
Gene Chromosome Location
Chr.1: 153537114-153543514 on Build GRCh38
Transcripts
NM_002962.1 XM_011509860.2 XM_017002029.1 XM_017002030.1 XM_017002031.1 XM_017002032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5903365 c.116G>A p.G39E Chr.1: 153540130-153540130 on Build GRCh38
896357 c.56T>A p.M19K Chr.1: 153540190-153540190 on Build GRCh38
6771217 c.97A>G p.T33A Chr.1: 153540149-153540149 on Build GRCh38
5951563 c.56delT p.M19fs*9 Chr.1: 153540190-153540190 on Build GRCh38
6248182 c.46C>A p.H16N Chr.1: 153540200-153540200 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779754466 Chr.1: 153540028-153540028 on Build GRCh38
rs61731552 Chr.1: 153540117-153540117 on Build GRCh38
rs772953478 Chr.1: 153540091-153540091 on Build GRCh38
rs774141022 Chr.1: 153540163-153540163 on Build GRCh38
rs914582371 Chr.1: 153540058-153540058 on Build GRCh38
rs761753792 Chr.1: 153540070-153540070 on Build GRCh38
rs748689002 Chr.1: 153540043-153540043 on Build GRCh38
rs775499138 Chr.1: 153540069-153540069 on Build GRCh38
rs765109640 Chr.1: 153540074-153540074 on Build GRCh38
rs536784065 Chr.1: 153540148-153540148 on Build GRCh38
rs371183110 Chr.1: 153540123-153540123 on Build GRCh38
rs2230870 Chr.1: 153540136-153540136 on Build GRCh38
rs34227468 Chr.1: 153540056-153540056 on Build GRCh38
rs747089931 Chr.1: 153540150-153540150 on Build GRCh38
rs780627233 Chr.1: 153540038-153540038 on Build GRCh38
rs146167533 Chr.1: 153540132-153540132 on Build GRCh38
rs754491244 Chr.1: 153540019-153540019 on Build GRCh38
rs978497388 Chr.1: 153540218-153540218 on Build GRCh38
rs762938935 Chr.1: 153540156-153540156 on Build GRCh38
rs772187575 Chr.1: 153540142-153540142 on Build GRCh38
rs147827125 Chr.1: 153540198-153540198 on Build GRCh38
rs778138440 Chr.1: 153540049-153540049 on Build GRCh38
rs149684046 Chr.1: 153540179-153540179 on Build GRCh38
rs770711130 Chr.1: 153540159-153540159 on Build GRCh38
rs182844223 Chr.1: 153540053-153540053 on Build GRCh38
rs757672688 Chr.1: 153540135-153540135 on Build GRCh38
rs897630189 Chr.1: 153540126-153540126 on Build GRCh38
rs752207756 Chr.1: 153540190-153540190 on Build GRCh38
rs749688258 Chr.1: 153540052-153540052 on Build GRCh38
rs1052670855 Chr.1: 153540040-153540040 on Build GRCh38
rs1045073828 Chr.1: 153540066-153540066 on Build GRCh38
rs531426304 Chr.1: 153540174-153540174 on Build GRCh38
rs772179582 Chr.1: 153540062-153540062 on Build GRCh38
rs747229610 Chr.1: 153540056-153540056 on Build GRCh38
rs148936113 Chr.1: 153540137-153540137 on Build GRCh38
rs751254770 Chr.1: 153540014-153540014 on Build GRCh38
rs768926248 Chr.1: 153540122-153540123 on Build GRCh38
rs756592022 Chr.1: 153540124-153540124 on Build GRCh38
rs756586740 Chr.1: 153540044-153540044 on Build GRCh38
rs41265158 Chr.1: 153540215-153540215 on Build GRCh38
rs923076708 Chr.1: 153540029-153540029 on Build GRCh38
rs1017265766 Chr.1: 153540195-153540195 on Build GRCh38
rs888699134 Chr.1: 153540158-153540158 on Build GRCh38
rs773112909 Chr.1: 153540152-153540152 on Build GRCh38
rs532923942 Chr.1: 153540133-153540133 on Build GRCh38
rs766214350 Chr.1: 153540111-153540111 on Build GRCh38
rs369991347 Chr.1: 153540118-153540118 on Build GRCh38
rs754381664 Chr.1: 153540213-153540213 on Build GRCh38
rs191063318 Chr.1: 153540201-153540201 on Build GRCh38
rs528717159 Chr.1: 153540054-153540054 on Build GRCh38
rs374976569 Chr.1: 153540196-153540196 on Build GRCh38
rs768822698 Chr.1: 153540151-153540151 on Build GRCh38
rs763744728 Chr.1: 153540200-153540200 on Build GRCh38
rs762707297 Chr.1: 153540093-153540093 on Build GRCh38

View Full Product Details