Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 37485973-37486198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR1E
Entrez Gene ID
64425
Gene Name
RNA polymerase I subunit E
Gene Aliases
PAF53, PRAF1
UniGene
Hs.591087
Gene Chromosome Location
Chr.9: 37485935-37503697 on Build GRCh38
Transcripts
NM_001282766.1 NM_022490.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1108994 c.47C>T p.A16V Chr.9: 37486094-37486094 on Build GRCh38
5998247 c.4G>A p.A2T Chr.9: 37486051-37486051 on Build GRCh38
5998432 c.37T>C p.Y13H Chr.9: 37486084-37486084 on Build GRCh38
1462413 c.69T>G p.A23A Chr.9: 37486116-37486116 on Build GRCh38
1108993 c.17T>C p.L6S Chr.9: 37486064-37486064 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs952185055 Chr.9: 37485998-37485998 on Build GRCh38
rs370920219 Chr.9: 37486174-37486174 on Build GRCh38
rs112552905 Chr.9: 37486112-37486112 on Build GRCh38
rs556400767 Chr.9: 37486131-37486131 on Build GRCh38
rs534029120 Chr.9: 37486007-37486007 on Build GRCh38
rs770572709 Chr.9: 37486146-37486146 on Build GRCh38
rs773030513 Chr.9: 37486038-37486038 on Build GRCh38
rs374830865 Chr.9: 37485999-37485999 on Build GRCh38
rs776282343 Chr.9: 37486021-37486021 on Build GRCh38
rs547356580 Chr.9: 37486008-37486008 on Build GRCh38
rs1053050037 Chr.9: 37486165-37486165 on Build GRCh38
rs769414999 Chr.9: 37486138-37486138 on Build GRCh38
rs768091738 Chr.9: 37486010-37486010 on Build GRCh38
rs756604188 Chr.9: 37486004-37486004 on Build GRCh38
rs576305630 Chr.9: 37486168-37486168 on Build GRCh38
rs760785380 Chr.9: 37486172-37486172 on Build GRCh38
rs946646150 Chr.9: 37485996-37485996 on Build GRCh38
rs1040868474 Chr.9: 37486022-37486022 on Build GRCh38
rs373709196 Chr.9: 37486152-37486152 on Build GRCh38
rs772693330 Chr.9: 37486139-37486139 on Build GRCh38
rs769644660 Chr.9: 37486036-37486036 on Build GRCh38
rs1027828606 Chr.9: 37485995-37485995 on Build GRCh38
rs778327268 Chr.9: 37486082-37486082 on Build GRCh38
rs767691203 Chr.9: 37486052-37486052 on Build GRCh38
rs1043648123 Chr.9: 37486001-37486001 on Build GRCh38
rs759673663 Chr.9: 37486049-37486049 on Build GRCh38
rs376455265 Chr.9: 37486155-37486155 on Build GRCh38
rs373462535 Chr.9: 37486006-37486006 on Build GRCh38
rs753034783 Chr.9: 37486055-37486055 on Build GRCh38
rs147983938 Chr.9: 37486045-37486045 on Build GRCh38
rs761446253 Chr.9: 37486033-37486033 on Build GRCh38
rs768216623 Chr.9: 37486117-37486117 on Build GRCh38
rs770823338 Chr.9: 37486143-37486143 on Build GRCh38
rs752799397 Chr.9: 37486167-37486167 on Build GRCh38
rs756369126 Chr.9: 37486068-37486068 on Build GRCh38
rs992446042 Chr.9: 37486039-37486039 on Build GRCh38
rs757520333 Chr.9: 37486102-37486102 on Build GRCh38
rs761755533 Chr.9: 37486112-37486116 on Build GRCh38
rs751384643 Chr.9: 37486046-37486046 on Build GRCh38
rs780677026 Chr.9: 37486118-37486118 on Build GRCh38
rs754249668 Chr.9: 37486094-37486094 on Build GRCh38
rs369689056 Chr.9: 37486005-37486005 on Build GRCh38
rs748571251 Chr.9: 37485994-37485994 on Build GRCh38
rs376139249 Chr.9: 37486043-37486043 on Build GRCh38
rs779481595 Chr.9: 37486108-37486108 on Build GRCh38

View Full Product Details