Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150337106-150337377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC61
Entrez Gene ID
65999
Gene Name
leucine rich repeat containing 61
Gene Aliases
HSPC295
UniGene
Hs.647119
Gene Chromosome Location
Chr.7: 150322639-150338156 on Build GRCh38
Transcripts
NM_001142928.1 NM_023942.2 XM_006716095.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6176671 c.313G>A p.A105T Chr.7: 150337174-150337174 on Build GRCh38
1449147 c.335C>T p.P112L Chr.7: 150337196-150337196 on Build GRCh38
1488355 c.452G>A p.R151Q Chr.7: 150337313-150337313 on Build GRCh38
3879237 c.430C>A p.P144T Chr.7: 150337291-150337291 on Build GRCh38
1213900 c.316G>T p.G106C Chr.7: 150337177-150337177 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969961213 Chr.7: 150337145-150337145 on Build GRCh38
rs765847012 Chr.7: 150337219-150337219 on Build GRCh38
rs769019408 Chr.7: 150337219-150337219 on Build GRCh38
rs139137430 Chr.7: 150337227-150337227 on Build GRCh38
rs773701188 Chr.7: 150337148-150337148 on Build GRCh38
rs759257308 Chr.7: 150337312-150337312 on Build GRCh38
rs766204413 Chr.7: 150337243-150337243 on Build GRCh38
rs752400572 Chr.7: 150337265-150337265 on Build GRCh38
rs750104213 Chr.7: 150337206-150337207 on Build GRCh38
rs538815439 Chr.7: 150337352-150337352 on Build GRCh38
rs751583242 Chr.7: 150337179-150337179 on Build GRCh38
rs779284257 Chr.7: 150337351-150337351 on Build GRCh38
rs761553240 Chr.7: 150337193-150337193 on Build GRCh38
rs761575975 Chr.7: 150337160-150337160 on Build GRCh38
rs770559995 Chr.7: 150337295-150337295 on Build GRCh38
rs753611359 Chr.7: 150337244-150337244 on Build GRCh38
rs761224752 Chr.7: 150337226-150337226 on Build GRCh38
rs776559806 Chr.7: 150337298-150337298 on Build GRCh38
rs142015684 Chr.7: 150337341-150337341 on Build GRCh38
rs148451290 Chr.7: 150337242-150337242 on Build GRCh38
rs200469711 Chr.7: 150337290-150337290 on Build GRCh38
rs781374578 Chr.7: 150337264-150337264 on Build GRCh38
rs767215437 Chr.7: 150337198-150337204 on Build GRCh38
rs961070923 Chr.7: 150337191-150337191 on Build GRCh38
rs760463532 Chr.7: 150337168-150337168 on Build GRCh38
rs779289236 Chr.7: 150337287-150337287 on Build GRCh38
rs746716995 Chr.7: 150337292-150337292 on Build GRCh38
rs775313070 Chr.7: 150337315-150337315 on Build GRCh38
rs781570841 Chr.7: 150337193-150337193 on Build GRCh38
rs750507748 Chr.7: 150337327-150337327 on Build GRCh38
rs754024922 Chr.7: 150337344-150337344 on Build GRCh38
rs780528344 Chr.7: 150337203-150337203 on Build GRCh38
rs772862456 Chr.7: 150337235-150337235 on Build GRCh38
rs200032049 Chr.7: 150337125-150337125 on Build GRCh38
rs371487497 Chr.7: 150337140-150337140 on Build GRCh38
rs750282370 Chr.7: 150337167-150337167 on Build GRCh38
rs374782170 Chr.7: 150337250-150337250 on Build GRCh38
rs754843189 Chr.7: 150337174-150337174 on Build GRCh38
rs746030305 Chr.7: 150337194-150337194 on Build GRCh38
rs530559551 Chr.7: 150337173-150337173 on Build GRCh38
rs755308143 Chr.7: 150337345-150337345 on Build GRCh38
rs893908814 Chr.7: 150337307-150337307 on Build GRCh38
rs773969367 Chr.7: 150337220-150337220 on Build GRCh38
rs571316608 Chr.7: 150337166-150337166 on Build GRCh38
rs766101661 Chr.7: 150337171-150337171 on Build GRCh38
rs755790190 Chr.7: 150337210-150337210 on Build GRCh38
rs759579728 Chr.7: 150337248-150337248 on Build GRCh38
rs751375450 Chr.7: 150337160-150337161 on Build GRCh38
rs777960753 Chr.7: 150337176-150337176 on Build GRCh38
rs202086113 Chr.7: 150337126-150337126 on Build GRCh38
rs772666241 Chr.7: 150337291-150337291 on Build GRCh38
rs765026130 Chr.7: 150337313-150337313 on Build GRCh38
rs757569479 Chr.7: 150337190-150337190 on Build GRCh38
rs149743914 Chr.7: 150337196-150337196 on Build GRCh38
rs767746585 Chr.7: 150337323-150337323 on Build GRCh38
rs374726542 Chr.7: 150337278-150337278 on Build GRCh38
rs372372621 Chr.7: 150337262-150337262 on Build GRCh38
rs923969650 Chr.7: 150337277-150337277 on Build GRCh38
rs765085116 Chr.7: 150337249-150337249 on Build GRCh38
rs769272100 Chr.7: 150337279-150337279 on Build GRCh38
rs566807948 Chr.7: 150337342-150337342 on Build GRCh38
rs749840423 Chr.7: 150337215-150337215 on Build GRCh38
rs3735169 Chr.7: 150337289-150337289 on Build GRCh38
rs147620488 Chr.7: 150337321-150337321 on Build GRCh38
rs753377412 Chr.7: 150337254-150337254 on Build GRCh38
rs768243839 Chr.7: 150337130-150337130 on Build GRCh38

View Full Product Details