Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 65576772-65577019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPWD1
Entrez Gene ID
23398
Gene Name
peptidylprolyl isomerase domain and WD repeat containing 1
Gene Aliases
-
UniGene
Hs.121432
Gene Chromosome Location
Chr.5: 65563236-65587549 on Build GRCh38
Transcripts
NM_001278926.1 NM_001278927.1 NM_001278929.1 NM_015342.3 XM_005248474.3 XM_006714583.3 XM_011543295.1 XM_017009286.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1471814 c.1025G>A p.R342Q Chr.5: 65576934-65576934 on Build GRCh38
3346165 c.1045G>T p.E349* Chr.5: 65576954-65576954 on Build GRCh38
6754706 c.1042C>T p.R348C Chr.5: 65576951-65576951 on Build GRCh38
3617469 c.1094A>G p.E365G Chr.5: 65577003-65577003 on Build GRCh38
345769 c.1057G>C p.V353L Chr.5: 65576966-65576966 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199936033 Chr.5: 65576952-65576952 on Build GRCh38
rs17233185 Chr.5: 65576870-65576870 on Build GRCh38
rs753472003 Chr.5: 65576959-65576959 on Build GRCh38
rs1056055356 Chr.5: 65576830-65576830 on Build GRCh38
rs750982428 Chr.5: 65576855-65576855 on Build GRCh38
rs773833542 Chr.5: 65576829-65576834 on Build GRCh38
rs759904660 Chr.5: 65576838-65576838 on Build GRCh38
rs755653225 Chr.5: 65576933-65576933 on Build GRCh38
rs888116647 Chr.5: 65576944-65576944 on Build GRCh38
rs774338325 Chr.5: 65576864-65576864 on Build GRCh38
rs749843216 Chr.5: 65576908-65576908 on Build GRCh38
rs377508764 Chr.5: 65576842-65576842 on Build GRCh38
rs938911194 Chr.5: 65576806-65576809 on Build GRCh38
rs374361968 Chr.5: 65576821-65576826 on Build GRCh38
rs759007939 Chr.5: 65576866-65576869 on Build GRCh38
rs930263660 Chr.5: 65576845-65576845 on Build GRCh38
rs763968281 Chr.5: 65576841-65576841 on Build GRCh38
rs1011884791 Chr.5: 65576977-65576977 on Build GRCh38
rs754670905 Chr.5: 65576990-65576990 on Build GRCh38
rs907383708 Chr.5: 65576805-65576805 on Build GRCh38
rs779215174 Chr.5: 65576991-65576991 on Build GRCh38

View Full Product Details