Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53872309-53872574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR8
Entrez Gene ID
93973
Gene Name
ARP8 actin-related protein 8 homolog
Gene Aliases
ARP8, INO80N, hArp8
UniGene
Hs.412186
Gene Chromosome Location
Chr.3: 53863096-53882202 on Build GRCh38
Transcripts
NM_022899.4 XM_005265587.4 XM_011534249.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6593143 c.1260C>T p.H420H Chr.3: 53872426-53872426 on Build GRCh38
4584571 c.390G>C p.L130L Chr.3: 53872411-53872411 on Build GRCh38
4755468 c.354G>T p.Q118H Chr.3: 53872447-53872447 on Build GRCh38
4119374 c.306T>G p.T102T Chr.3: 53872495-53872495 on Build GRCh38
5131713 c.1279G>C p.A427P Chr.3: 53872407-53872407 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746741793 Chr.3: 53872442-53872442 on Build GRCh38
rs753127053 Chr.3: 53872379-53872379 on Build GRCh38
rs201796393 Chr.3: 53872524-53872524 on Build GRCh38
rs556414817 Chr.3: 53872419-53872419 on Build GRCh38
rs200750107 Chr.3: 53872444-53872444 on Build GRCh38
rs776785672 Chr.3: 53872445-53872445 on Build GRCh38
rs946965278 Chr.3: 53872355-53872355 on Build GRCh38
rs375742765 Chr.3: 53872371-53872371 on Build GRCh38
rs757896511 Chr.3: 53872352-53872353 on Build GRCh38
rs765203812 Chr.3: 53872453-53872453 on Build GRCh38
rs199805171 Chr.3: 53872543-53872543 on Build GRCh38
rs754699144 Chr.3: 53872518-53872518 on Build GRCh38
rs377647744 Chr.3: 53872497-53872497 on Build GRCh38
rs748976721 Chr.3: 53872426-53872426 on Build GRCh38
rs779763400 Chr.3: 53872537-53872538 on Build GRCh38
rs4687757 Chr.3: 53872486-53872486 on Build GRCh38
rs915417837 Chr.3: 53872345-53872345 on Build GRCh38
rs757707229 Chr.3: 53872411-53872411 on Build GRCh38
rs778534930 Chr.3: 53872429-53872429 on Build GRCh38
rs145834372 Chr.3: 53872407-53872407 on Build GRCh38
rs371924812 Chr.3: 53872521-53872521 on Build GRCh38
rs886846717 Chr.3: 53872463-53872463 on Build GRCh38
rs373126432 Chr.3: 53872469-53872469 on Build GRCh38
rs950300355 Chr.3: 53872545-53872545 on Build GRCh38
rs779782027 Chr.3: 53872425-53872425 on Build GRCh38
rs374523447 Chr.3: 53872501-53872501 on Build GRCh38
rs367777535 Chr.3: 53872380-53872380 on Build GRCh38
rs753518507 Chr.3: 53872509-53872509 on Build GRCh38
rs988976011 Chr.3: 53872523-53872523 on Build GRCh38
rs138389972 Chr.3: 53872527-53872527 on Build GRCh38
rs777649971 Chr.3: 53872544-53872544 on Build GRCh38
rs759854975 Chr.3: 53872449-53872449 on Build GRCh38
rs371132984 Chr.3: 53872500-53872500 on Build GRCh38
rs145231853 Chr.3: 53872434-53872434 on Build GRCh38
rs890373366 Chr.3: 53872364-53872364 on Build GRCh38
rs748003248 Chr.3: 53872432-53872432 on Build GRCh38
rs1008933673 Chr.3: 53872386-53872386 on Build GRCh38

View Full Product Details