Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132883125-132883396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFF4
Entrez Gene ID
27125
Gene Name
AF4/FMR2 family member 4
Gene Aliases
AF5Q31, CHOPS, MCEF
UniGene
Hs.519313
Gene Chromosome Location
Chr.5: 132875379-132963662 on Build GRCh38
Transcripts
NM_014423.3 XM_005271963.4 XM_005271964.4 XM_006714587.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3137669 c.3363A>G p.K1121K Chr.5: 132883341-132883341 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs956783718 Chr.5: 132883203-132883203 on Build GRCh38
rs765528883 Chr.5: 132883309-132883309 on Build GRCh38
rs891038447 Chr.5: 132883276-132883276 on Build GRCh38
rs377180468 Chr.5: 132883312-132883312 on Build GRCh38
rs781454670 Chr.5: 132883362-132883362 on Build GRCh38
rs73788220 Chr.5: 132883258-132883258 on Build GRCh38
rs745455720 Chr.5: 132883234-132883234 on Build GRCh38
rs747309720 Chr.5: 132883344-132883344 on Build GRCh38
rs374222042 Chr.5: 132883346-132883346 on Build GRCh38
rs528233721 Chr.5: 132883162-132883162 on Build GRCh38
rs369966536 Chr.5: 132883159-132883159 on Build GRCh38
rs777846570 Chr.5: 132883335-132883335 on Build GRCh38
rs1043846283 Chr.5: 132883183-132883183 on Build GRCh38
rs186991911 Chr.5: 132883209-132883209 on Build GRCh38
rs758864318 Chr.5: 132883319-132883319 on Build GRCh38
rs540652154 Chr.5: 132883164-132883164 on Build GRCh38
rs916590544 Chr.5: 132883153-132883153 on Build GRCh38
rs975669029 Chr.5: 132883254-132883254 on Build GRCh38
rs757256807 Chr.5: 132883347-132883347 on Build GRCh38
rs753066885 Chr.5: 132883313-132883313 on Build GRCh38
rs755497519 Chr.5: 132883301-132883301 on Build GRCh38
rs912690530 Chr.5: 132883218-132883218 on Build GRCh38
rs56961677 Chr.5: 132883237-132883237 on Build GRCh38
rs925378824 Chr.5: 132883179-132883179 on Build GRCh38
rs930995991 Chr.5: 132883268-132883268 on Build GRCh38

View Full Product Details