Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49412566-49412837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCTA
Entrez Gene ID
6988
Gene Name
T-cell leukemia translocation altered
Gene Aliases
-
UniGene
Hs.517962
Gene Chromosome Location
Chr.3: 49412206-49416476 on Build GRCh38
Transcripts
NM_022171.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6273642 c.197G>T p.G66V Chr.3: 49412623-49412623 on Build GRCh38
5950093 c.160C>T p.Q54* Chr.3: 49412586-49412586 on Build GRCh38
1046147 c.203A>G p.Y68C Chr.3: 49412629-49412629 on Build GRCh38
4432642 c.201G>T p.L67F Chr.3: 49412627-49412627 on Build GRCh38
5671773 c.170delG p.F59fs*6 Chr.3: 49412596-49412596 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781011462 Chr.3: 49412681-49412681 on Build GRCh38
rs936001070 Chr.3: 49412615-49412615 on Build GRCh38
rs201580769 Chr.3: 49412689-49412689 on Build GRCh38
rs140484970 Chr.3: 49412588-49412588 on Build GRCh38
rs762108083 Chr.3: 49412651-49412651 on Build GRCh38
rs754865265 Chr.3: 49412679-49412679 on Build GRCh38
rs765432451 Chr.3: 49412653-49412653 on Build GRCh38
rs769326574 Chr.3: 49412644-49412644 on Build GRCh38
rs752475760 Chr.3: 49412683-49412683 on Build GRCh38
rs1024443752 Chr.3: 49412785-49412785 on Build GRCh38
rs1055704057 Chr.3: 49412652-49412652 on Build GRCh38
rs569051994 Chr.3: 49412802-49412802 on Build GRCh38
rs368595914 Chr.3: 49412657-49412657 on Build GRCh38
rs997520412 Chr.3: 49412706-49412706 on Build GRCh38
rs951171577 Chr.3: 49412811-49412811 on Build GRCh38
rs1007837344 Chr.3: 49412721-49412721 on Build GRCh38
rs145281373 Chr.3: 49412627-49412627 on Build GRCh38
rs375116933 Chr.3: 49412677-49412677 on Build GRCh38
rs747923015 Chr.3: 49412596-49412596 on Build GRCh38
rs375692245 Chr.3: 49412689-49412690 on Build GRCh38
rs372020505 Chr.3: 49412666-49412666 on Build GRCh38
rs763218481 Chr.3: 49412662-49412662 on Build GRCh38
rs755869996 Chr.3: 49412685-49412685 on Build GRCh38
rs145962554 Chr.3: 49412633-49412633 on Build GRCh38
rs754956265 Chr.3: 49412587-49412587 on Build GRCh38
rs762614917 Chr.3: 49412680-49412681 on Build GRCh38
rs546230864 Chr.3: 49412686-49412686 on Build GRCh38
rs539291506 Chr.3: 49412806-49412806 on Build GRCh38
rs879230499 Chr.3: 49412680-49412680 on Build GRCh38
rs777016419 Chr.3: 49412650-49412650 on Build GRCh38
rs550335746 Chr.3: 49412718-49412718 on Build GRCh38
rs774898852 Chr.3: 49412605-49412605 on Build GRCh38
rs761242017 Chr.3: 49412642-49412642 on Build GRCh38
rs972223124 Chr.3: 49412768-49412768 on Build GRCh38
rs992071911 Chr.3: 49412744-49412744 on Build GRCh38
rs748768655 Chr.3: 49412622-49412622 on Build GRCh38
rs1016491562 Chr.3: 49412730-49412730 on Build GRCh38
rs777307615 Chr.3: 49412609-49412609 on Build GRCh38
rs963891523 Chr.3: 49412752-49412752 on Build GRCh38
rs60042123 Chr.3: 49412655-49412655 on Build GRCh38
rs937295798 Chr.3: 49412640-49412640 on Build GRCh38
rs114287800 Chr.3: 49412704-49412704 on Build GRCh38
rs1030370571 Chr.3: 49412707-49412707 on Build GRCh38

View Full Product Details