Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 91000437-91000670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS33B
Entrez Gene ID
26276
Gene Name
VPS33B, late endosome and lysosome associated
Gene Aliases
-
UniGene
Hs.745007
Gene Chromosome Location
Chr.15: 90998416-91022621 on Build GRCh38
Transcripts
NM_001289148.1 NM_001289149.1 NM_018668.4 XM_005254887.1 XM_011521448.1 XM_011521449.2 XM_017022075.1 XM_017022076.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5754751 c.1495G>A p.G499S Chr.15: 91000576-91000576 on Build GRCh38
966634 c.1574T>C p.I525T Chr.15: 91000497-91000497 on Build GRCh38
966635 c.1523A>G p.D508G Chr.15: 91000548-91000548 on Build GRCh38
966636 c.1497C>T p.G499G Chr.15: 91000574-91000574 on Build GRCh38
147985 c.1581+3G>A p.? Chr.15: 91000487-91000487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753782249 Chr.15: 91000469-91000469 on Build GRCh38
rs989449085 Chr.15: 91000585-91000585 on Build GRCh38
rs767914174 Chr.15: 91000574-91000574 on Build GRCh38
rs773182950 Chr.15: 91000632-91000632 on Build GRCh38
rs200348382 Chr.15: 91000611-91000611 on Build GRCh38
rs768250541 Chr.15: 91000537-91000537 on Build GRCh38
rs775501556 Chr.15: 91000565-91000565 on Build GRCh38
rs758590786 Chr.15: 91000500-91000500 on Build GRCh38
rs868286599 Chr.15: 91000645-91000645 on Build GRCh38
rs1007308602 Chr.15: 91000481-91000481 on Build GRCh38
rs781683489 Chr.15: 91000513-91000513 on Build GRCh38
rs774229913 Chr.15: 91000538-91000538 on Build GRCh38
rs11073964 Chr.15: 91000531-91000531 on Build GRCh38
rs778476250 Chr.15: 91000627-91000628 on Build GRCh38
rs746964381 Chr.15: 91000633-91000633 on Build GRCh38
rs200539802 Chr.15: 91000570-91000570 on Build GRCh38
rs977026766 Chr.15: 91000549-91000549 on Build GRCh38
rs745328531 Chr.15: 91000514-91000514 on Build GRCh38
rs764239087 Chr.15: 91000571-91000571 on Build GRCh38
rs1018892338 Chr.15: 91000516-91000516 on Build GRCh38
rs773306000 Chr.15: 91000552-91000552 on Build GRCh38
rs772096296 Chr.15: 91000551-91000551 on Build GRCh38
rs765330198 Chr.15: 91000476-91000476 on Build GRCh38
rs747961052 Chr.15: 91000543-91000543 on Build GRCh38
rs751858602 Chr.15: 91000573-91000573 on Build GRCh38
rs765172002 Chr.15: 91000580-91000580 on Build GRCh38
rs370877739 Chr.15: 91000487-91000487 on Build GRCh38
rs750896440 Chr.15: 91000576-91000576 on Build GRCh38
rs373949995 Chr.15: 91000594-91000594 on Build GRCh38
rs138436850 Chr.15: 91000578-91000578 on Build GRCh38
rs770044991 Chr.15: 91000624-91000625 on Build GRCh38
rs757706978 Chr.15: 91000511-91000511 on Build GRCh38
rs765330988 Chr.15: 91000554-91000554 on Build GRCh38
rs202088691 Chr.15: 91000637-91000637 on Build GRCh38
rs756643532 Chr.15: 91000577-91000577 on Build GRCh38
rs754592554 Chr.15: 91000589-91000589 on Build GRCh38
rs779601858 Chr.15: 91000579-91000579 on Build GRCh38
rs775586418 Chr.15: 91000636-91000636 on Build GRCh38
rs751982861 Chr.15: 91000504-91000504 on Build GRCh38
rs3177428 Chr.15: 91000533-91000533 on Build GRCh38
rs748759914 Chr.15: 91000584-91000584 on Build GRCh38
rs764518291 Chr.15: 91000503-91000503 on Build GRCh38
rs748963280 Chr.15: 91000536-91000536 on Build GRCh38

View Full Product Details